Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 57532525-57532798 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCTN2
Entrez Gene ID
10540
Gene Name
dynactin subunit 2
Gene Aliases
DCTN50, DYNAMITIN, HEL-S-77, RBP50
UniGene
Hs.289123
Gene Chromosome Location
Chr.12: 57530050-57547331 on Build GRCh38
Transcripts
NM_001261412.1 NM_001261413.1 NM_006400.4 XM_011537761.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6641120 c.888C>T p.N296N Chr.12: 57532623-57532623 on Build GRCh38
1241064 c.855G>T p.E285D Chr.12: 57532745-57532745 on Build GRCh38
272570 c.823G>A p.A275T Chr.12: 57532777-57532777 on Build GRCh38
4998497 c.838G>T p.V280F Chr.12: 57532762-57532762 on Build GRCh38
4802646 c.823G>A p.A275T Chr.12: 57532777-57532777 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs703837 Chr.12: 57532689-57532689 on Build GRCh38
rs573070976 Chr.12: 57532657-57532657 on Build GRCh38
rs748530280 Chr.12: 57532567-57532567 on Build GRCh38
rs763008780 Chr.12: 57532669-57532669 on Build GRCh38
rs755230022 Chr.12: 57532559-57532559 on Build GRCh38
rs202123398 Chr.12: 57532703-57532703 on Build GRCh38
rs757824658 Chr.12: 57532751-57532751 on Build GRCh38
rs756484800 Chr.12: 57532701-57532701 on Build GRCh38
rs775130542 Chr.12: 57532638-57532638 on Build GRCh38
rs557813676 Chr.12: 57532654-57532654 on Build GRCh38
rs758908043 Chr.12: 57532713-57532714 on Build GRCh38
rs186544514 Chr.12: 57532578-57532578 on Build GRCh38
rs751707552 Chr.12: 57532691-57532691 on Build GRCh38
rs575590538 Chr.12: 57532705-57532705 on Build GRCh38
rs943000898 Chr.12: 57532674-57532674 on Build GRCh38
rs181473937 Chr.12: 57532757-57532757 on Build GRCh38
rs757335083 Chr.12: 57532656-57532657 on Build GRCh38
rs370959939 Chr.12: 57532628-57532628 on Build GRCh38
rs539406696 Chr.12: 57532589-57532589 on Build GRCh38
rs915957007 Chr.12: 57532575-57532575 on Build GRCh38
rs141248212 Chr.12: 57532777-57532777 on Build GRCh38
rs1038143921 Chr.12: 57532622-57532622 on Build GRCh38
rs749610898 Chr.12: 57532736-57532736 on Build GRCh38
rs753927685 Chr.12: 57532552-57532557 on Build GRCh38
rs890390972 Chr.12: 57532671-57532671 on Build GRCh38
rs542297132 Chr.12: 57532697-57532697 on Build GRCh38
rs778256171 Chr.12: 57532614-57532614 on Build GRCh38
rs118012274 Chr.12: 57532695-57532695 on Build GRCh38
rs973546332 Chr.12: 57532637-57532637 on Build GRCh38
rs188842737 Chr.12: 57532665-57532665 on Build GRCh38
rs765577687 Chr.12: 57532623-57532623 on Build GRCh38
rs750998385 Chr.12: 57532704-57532709 on Build GRCh38
rs765764479 Chr.12: 57532680-57532680 on Build GRCh38
rs991504387 Chr.12: 57532573-57532573 on Build GRCh38
rs186110292 Chr.12: 57532775-57532775 on Build GRCh38
rs748990564 Chr.12: 57532740-57532740 on Build GRCh38
rs781367587 Chr.12: 57532561-57532561 on Build GRCh38
rs752390584 Chr.12: 57532560-57532560 on Build GRCh38
rs201411813 Chr.12: 57532778-57532778 on Build GRCh38
rs752849976 Chr.12: 57532699-57532699 on Build GRCh38
rs374575250 Chr.12: 57532664-57532664 on Build GRCh38
rs1042164454 Chr.12: 57532722-57532722 on Build GRCh38
rs200231991 Chr.12: 57532554-57532554 on Build GRCh38
rs1003936855 Chr.12: 57532610-57532610 on Build GRCh38
rs375551214 Chr.12: 57532653-57532653 on Build GRCh38

View Full Product Details