Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 28933177-28933408 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD19
Entrez Gene ID
930
Gene Name
CD19 molecule
Gene Aliases
B4, CVID3
UniGene
Hs.652262
Gene Chromosome Location
Chr.16: 28931735-28939347 on Build GRCh38
Transcripts
NM_001178098.1 NM_001770.5 XM_006721103.3 XM_011545981.1 XM_017023893.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4554706 c.628G>A p.G210S Chr.16: 28933302-28933302 on Build GRCh38
4567315 c.661_662GG>AA p.G221K Chr.16: 28933335-28933336 on Build GRCh38
4059903 c.731C>T p.T244M Chr.16: 28933405-28933405 on Build GRCh38
6623507 c.696C>T p.D232D Chr.16: 28933370-28933370 on Build GRCh38
3754835 c.705G>T p.P235P Chr.16: 28933379-28933379 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776701989 Chr.16: 28933289-28933289 on Build GRCh38
rs773781983 Chr.16: 28933361-28933361 on Build GRCh38
rs992783874 Chr.16: 28933301-28933301 on Build GRCh38
rs763327569 Chr.16: 28933352-28933352 on Build GRCh38
rs747991749 Chr.16: 28933228-28933229 on Build GRCh38
rs748449748 Chr.16: 28933302-28933302 on Build GRCh38
rs376395490 Chr.16: 28933374-28933374 on Build GRCh38
rs752270549 Chr.16: 28933371-28933371 on Build GRCh38
rs779384297 Chr.16: 28933245-28933245 on Build GRCh38
rs200751866 Chr.16: 28933266-28933266 on Build GRCh38
rs773089937 Chr.16: 28933322-28933322 on Build GRCh38
rs766314772 Chr.16: 28933336-28933336 on Build GRCh38
rs746331250 Chr.16: 28933247-28933247 on Build GRCh38
rs968233112 Chr.16: 28933211-28933211 on Build GRCh38
rs768936030 Chr.16: 28933284-28933284 on Build GRCh38
rs757550516 Chr.16: 28933226-28933226 on Build GRCh38
rs917602503 Chr.16: 28933321-28933321 on Build GRCh38
rs35979293 Chr.16: 28933379-28933379 on Build GRCh38
rs762828020 Chr.16: 28933323-28933323 on Build GRCh38
rs769440514 Chr.16: 28933318-28933318 on Build GRCh38
rs935597322 Chr.16: 28933292-28933292 on Build GRCh38
rs1018349529 Chr.16: 28933202-28933202 on Build GRCh38
rs759044816 Chr.16: 28933366-28933366 on Build GRCh38
rs759009589 Chr.16: 28933259-28933259 on Build GRCh38
rs752463238 Chr.16: 28933212-28933212 on Build GRCh38
rs924224771 Chr.16: 28933283-28933283 on Build GRCh38
rs780236424 Chr.16: 28933277-28933277 on Build GRCh38
rs979613327 Chr.16: 28933246-28933246 on Build GRCh38
rs373234931 Chr.16: 28933370-28933370 on Build GRCh38
rs750825735 Chr.16: 28933375-28933375 on Build GRCh38
rs776423905 Chr.16: 28933211-28933212 on Build GRCh38
rs758919478 Chr.16: 28933378-28933378 on Build GRCh38

View Full Product Details