Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 89680522-89680783 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOLH1B
Entrez Gene ID
219595
Gene Name
folate hydrolase 1B
Gene Aliases
FOLH2, FOLHP, GCP3, GCPIII, PSM, PSMA-LIKE, PSMAL
UniGene
Hs.645352
Gene Chromosome Location
Chr.11: 89637575-89698718 on Build GRCh38
Transcripts
NM_153696.2 XM_017017319.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4592744 c.464G>A p.R155K Chr.11: 89680624-89680624 on Build GRCh38
1357666 c.466G>T p.V156F Chr.11: 89680626-89680626 on Build GRCh38
5673474 c.464G>T p.R155I Chr.11: 89680624-89680624 on Build GRCh38
7273990 c.503A>G p.N168S Chr.11: 89680663-89680663 on Build GRCh38
3953914 c.473G>A p.C158Y Chr.11: 89680633-89680633 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777426370 Chr.11: 89680611-89680611 on Build GRCh38
rs759326860 Chr.11: 89680665-89680665 on Build GRCh38
rs774627434 Chr.11: 89680561-89680561 on Build GRCh38
rs10830340 Chr.11: 89680682-89680682 on Build GRCh38
rs760876958 Chr.11: 89680716-89680716 on Build GRCh38
rs768992761 Chr.11: 89680689-89680689 on Build GRCh38
rs745394773 Chr.11: 89680685-89680685 on Build GRCh38
rs778642955 Chr.11: 89680593-89680593 on Build GRCh38
rs10830339 Chr.11: 89680613-89680613 on Build GRCh38
rs547719451 Chr.11: 89680592-89680592 on Build GRCh38
rs776518211 Chr.11: 89680722-89680722 on Build GRCh38
rs762288383 Chr.11: 89680629-89680629 on Build GRCh38
rs766305695 Chr.11: 89680585-89680585 on Build GRCh38
rs753813327 Chr.11: 89680651-89680651 on Build GRCh38
rs760811901 Chr.11: 89680641-89680641 on Build GRCh38
rs756289465 Chr.11: 89680578-89680578 on Build GRCh38
rs758758904 Chr.11: 89680621-89680622 on Build GRCh38
rs202238498 Chr.11: 89680587-89680587 on Build GRCh38
rs527444398 Chr.11: 89680559-89680559 on Build GRCh38
rs7128813 Chr.11: 89680699-89680699 on Build GRCh38
rs145858863 Chr.11: 89680627-89680627 on Build GRCh38
rs759685900 Chr.11: 89680723-89680723 on Build GRCh38
rs571515964 Chr.11: 89680679-89680679 on Build GRCh38
rs767863363 Chr.11: 89680569-89680569 on Build GRCh38
rs561142105 Chr.11: 89680594-89680594 on Build GRCh38
rs551577221 Chr.11: 89680669-89680669 on Build GRCh38
rs913035399 Chr.11: 89680631-89680631 on Build GRCh38
rs771192503 Chr.11: 89680721-89680721 on Build GRCh38
rs374222504 Chr.11: 89680687-89680687 on Build GRCh38
rs772609954 Chr.11: 89680710-89680710 on Build GRCh38
rs766736005 Chr.11: 89680637-89680638 on Build GRCh38
rs374543265 Chr.11: 89680724-89680724 on Build GRCh38
rs534078857 Chr.11: 89680681-89680681 on Build GRCh38
rs773559406 Chr.11: 89680715-89680715 on Build GRCh38
rs750704379 Chr.11: 89680568-89680569 on Build GRCh38
rs762057668 Chr.11: 89680563-89680563 on Build GRCh38
rs767805500 Chr.11: 89680640-89680640 on Build GRCh38
rs757990660 Chr.11: 89680606-89680606 on Build GRCh38
rs371601359 Chr.11: 89680709-89680709 on Build GRCh38
rs766431379 Chr.11: 89680645-89680645 on Build GRCh38
rs776050875 Chr.11: 89680619-89680619 on Build GRCh38
rs140720479 Chr.11: 89680624-89680624 on Build GRCh38
rs146604247 Chr.11: 89680626-89680626 on Build GRCh38
rs867806218 Chr.11: 89680608-89680608 on Build GRCh38
rs765236630 Chr.11: 89680667-89680667 on Build GRCh38
rs770388574 Chr.11: 89680618-89680618 on Build GRCh38

View Full Product Details