Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 143511234-143511491 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FUCA2
Entrez Gene ID
2519
Gene Name
fucosidase, alpha-L- 2, plasma
Gene Aliases
dJ20N2.5
UniGene
Hs.17680
Gene Chromosome Location
Chr.6: 143494812-143511883 on Build GRCh38
Transcripts
NM_032020.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7082964 c.189G>T p.V63V Chr.6: 143511446-143511446 on Build GRCh38
5024325 c.212G>A p.S71N Chr.6: 143511423-143511423 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs545893870 Chr.6: 143511328-143511328 on Build GRCh38
rs761024545 Chr.6: 143511444-143511444 on Build GRCh38
rs558149840 Chr.6: 143511337-143511337 on Build GRCh38
rs540133278 Chr.6: 143511370-143511370 on Build GRCh38
rs748797282 Chr.6: 143511428-143511428 on Build GRCh38
rs985295699 Chr.6: 143511325-143511325 on Build GRCh38
rs770304902 Chr.6: 143511435-143511435 on Build GRCh38
rs549302348 Chr.6: 143511326-143511326 on Build GRCh38
rs572818814 Chr.6: 143511338-143511338 on Build GRCh38
rs879839591 Chr.6: 143511306-143511306 on Build GRCh38
rs780896097 Chr.6: 143511411-143511411 on Build GRCh38
rs1012286121 Chr.6: 143511287-143511287 on Build GRCh38
rs771567042 Chr.6: 143511278-143511278 on Build GRCh38
rs1009866121 Chr.6: 143511314-143511314 on Build GRCh38
rs1021346163 Chr.6: 143511316-143511316 on Build GRCh38
rs965283643 Chr.6: 143511340-143511340 on Build GRCh38
rs557093449 Chr.6: 143511257-143511257 on Build GRCh38
rs919705872 Chr.6: 143511380-143511380 on Build GRCh38
rs976623907 Chr.6: 143511343-143511343 on Build GRCh38
rs777309409 Chr.6: 143511427-143511427 on Build GRCh38
rs777126432 Chr.6: 143511460-143511460 on Build GRCh38
rs955667690 Chr.6: 143511292-143511292 on Build GRCh38
rs751405206 Chr.6: 143511381-143511381 on Build GRCh38
rs529032539 Chr.6: 143511268-143511268 on Build GRCh38
rs762179871 Chr.6: 143511466-143511466 on Build GRCh38
rs866796659 Chr.6: 143511453-143511453 on Build GRCh38
rs749272547 Chr.6: 143511263-143511263 on Build GRCh38
rs750571718 Chr.6: 143511470-143511470 on Build GRCh38
rs769422810 Chr.6: 143511426-143511426 on Build GRCh38
rs983689395 Chr.6: 143511304-143511304 on Build GRCh38
rs192285682 Chr.6: 143511290-143511290 on Build GRCh38
rs1024976859 Chr.6: 143511297-143511297 on Build GRCh38
rs369061103 Chr.6: 143511368-143511368 on Build GRCh38
rs970991226 Chr.6: 143511303-143511303 on Build GRCh38
rs758308714 Chr.6: 143511363-143511363 on Build GRCh38
rs776083578 Chr.6: 143511440-143511440 on Build GRCh38
rs747863648 Chr.6: 143511421-143511421 on Build GRCh38
rs769212130 Chr.6: 143511458-143511458 on Build GRCh38
rs754813697 Chr.6: 143511397-143511397 on Build GRCh38
rs968893282 Chr.6: 143511319-143511319 on Build GRCh38
rs765526154 Chr.6: 143511469-143511469 on Build GRCh38

View Full Product Details