Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 74561950-74562211 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNDP1
Entrez Gene ID
84735
Gene Name
carnosine dipeptidase 1
Gene Aliases
CN1, CPGL2, HsT2308
UniGene
Hs.400613
Gene Chromosome Location
Chr.18: 74534457-74585025 on Build GRCh38
Transcripts
NM_032649.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3146331 c.467-10C>T p.? Chr.18: 74562037-74562037 on Build GRCh38
3146332 c.473T>G p.L158R Chr.18: 74562053-74562053 on Build GRCh38
5434964 c.505C>T p.P169S Chr.18: 74562085-74562085 on Build GRCh38
4073228 c.493G>A p.D165N Chr.18: 74562073-74562073 on Build GRCh38
4797418 c.533G>A p.S178N Chr.18: 74562113-74562113 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1051231478 Chr.18: 74562012-74562012 on Build GRCh38
rs753753972 Chr.18: 74562041-74562041 on Build GRCh38
rs773993247 Chr.18: 74562180-74562182 on Build GRCh38
rs374853002 Chr.18: 74562061-74562061 on Build GRCh38
rs9967490 Chr.18: 74561977-74561977 on Build GRCh38
rs747787545 Chr.18: 74562060-74562060 on Build GRCh38
rs769197233 Chr.18: 74562076-74562076 on Build GRCh38
rs777300476 Chr.18: 74562026-74562026 on Build GRCh38
rs918724438 Chr.18: 74562000-74562000 on Build GRCh38
rs745492929 Chr.18: 74562124-74562124 on Build GRCh38
rs890789122 Chr.18: 74562016-74562016 on Build GRCh38
rs377276019 Chr.18: 74561994-74561994 on Build GRCh38
rs368206421 Chr.18: 74562172-74562172 on Build GRCh38
rs550763706 Chr.18: 74562114-74562114 on Build GRCh38
rs987005192 Chr.18: 74562040-74562040 on Build GRCh38
rs753564348 Chr.18: 74561996-74561996 on Build GRCh38
rs768391446 Chr.18: 74562161-74562163 on Build GRCh38
rs770554951 Chr.18: 74562037-74562037 on Build GRCh38
rs757889546 Chr.18: 74562025-74562025 on Build GRCh38
rs769181262 Chr.18: 74562127-74562127 on Build GRCh38
rs758303238 Chr.18: 74562097-74562097 on Build GRCh38
rs765041829 Chr.18: 74562182-74562182 on Build GRCh38
rs775155013 Chr.18: 74562086-74562087 on Build GRCh38
rs752613079 Chr.18: 74562018-74562018 on Build GRCh38
rs867186853 Chr.18: 74562042-74562042 on Build GRCh38
rs773287080 Chr.18: 74562068-74562068 on Build GRCh38
rs112761183 Chr.18: 74562019-74562019 on Build GRCh38
rs754764835 Chr.18: 74561997-74561997 on Build GRCh38
rs369494548 Chr.18: 74562043-74562043 on Build GRCh38
rs746660500 Chr.18: 74562035-74562035 on Build GRCh38
rs760867820 Chr.18: 74562170-74562170 on Build GRCh38
rs748618442 Chr.18: 74562158-74562158 on Build GRCh38
rs752666050 Chr.18: 74562077-74562077 on Build GRCh38
rs745370800 Chr.18: 74562045-74562046 on Build GRCh38
rs372520573 Chr.18: 74562177-74562177 on Build GRCh38
rs771780063 Chr.18: 74562074-74562074 on Build GRCh38
rs142662230 Chr.18: 74562062-74562062 on Build GRCh38
rs773917466 Chr.18: 74562001-74562002 on Build GRCh38
rs779379707 Chr.18: 74562140-74562140 on Build GRCh38
rs781047897 Chr.18: 74562115-74562115 on Build GRCh38
rs773588613 Chr.18: 74562166-74562166 on Build GRCh38
rs759717511 Chr.18: 74562181-74562181 on Build GRCh38
rs146707393 Chr.18: 74562073-74562073 on Build GRCh38
rs762541533 Chr.18: 74562107-74562108 on Build GRCh38
rs778695428 Chr.18: 74562048-74562048 on Build GRCh38
rs754049170 Chr.18: 74562072-74562072 on Build GRCh38
rs1025432950 Chr.18: 74562087-74562087 on Build GRCh38
rs202064457 Chr.18: 74562049-74562049 on Build GRCh38
rs570510264 Chr.18: 74562164-74562164 on Build GRCh38
rs562967819 Chr.18: 74562069-74562069 on Build GRCh38
rs529290100 Chr.18: 74562003-74562003 on Build GRCh38
rs778883603 Chr.18: 74562015-74562015 on Build GRCh38
rs376790102 Chr.18: 74562185-74562185 on Build GRCh38

View Full Product Details