Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 108657225-108657498 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CORO1C
Entrez Gene ID
23603
Gene Name
coronin 1C
Gene Aliases
HCRNN4
UniGene
Hs.330384
Gene Chromosome Location
Chr.12: 108645109-108731550 on Build GRCh38
Transcripts
NM_001105237.2 NM_001276471.1 NM_014325.3 XM_011538125.1 XM_017019121.1 XM_017019122.1 XM_017019123.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5882176 c.721A>C p.S241R Chr.12: 108657333-108657333 on Build GRCh38
6452063 c.875G>A p.R292H Chr.12: 108657338-108657338 on Build GRCh38
369880 c.739C>T p.L247F Chr.12: 108657315-108657315 on Build GRCh38
5916198 c.683C>T p.A228V Chr.12: 108657371-108657371 on Build GRCh38
4831725 c.631G>A p.E211K Chr.12: 108657423-108657423 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368890006 Chr.12: 108657413-108657413 on Build GRCh38
rs751537384 Chr.12: 108657307-108657307 on Build GRCh38
rs746017615 Chr.12: 108657257-108657257 on Build GRCh38
rs773011352 Chr.12: 108657423-108657423 on Build GRCh38
rs772502968 Chr.12: 108657265-108657265 on Build GRCh38
rs200384833 Chr.12: 108657297-108657297 on Build GRCh38
rs1034454246 Chr.12: 108657479-108657479 on Build GRCh38
rs762555895 Chr.12: 108657304-108657304 on Build GRCh38
rs372573155 Chr.12: 108657443-108657443 on Build GRCh38
rs557390202 Chr.12: 108657336-108657336 on Build GRCh38
rs770444434 Chr.12: 108657452-108657452 on Build GRCh38
rs769092433 Chr.12: 108657277-108657277 on Build GRCh38
rs776896693 Chr.12: 108657289-108657289 on Build GRCh38
rs754817920 Chr.12: 108657477-108657477 on Build GRCh38
rs899312118 Chr.12: 108657306-108657306 on Build GRCh38
rs747515987 Chr.12: 108657350-108657350 on Build GRCh38
rs371061578 Chr.12: 108657417-108657417 on Build GRCh38
rs188618736 Chr.12: 108657346-108657346 on Build GRCh38
rs761532317 Chr.12: 108657291-108657291 on Build GRCh38
rs757467310 Chr.12: 108657331-108657331 on Build GRCh38
rs192795242 Chr.12: 108657370-108657370 on Build GRCh38
rs535985851 Chr.12: 108657313-108657313 on Build GRCh38
rs752547940 Chr.12: 108657328-108657328 on Build GRCh38
rs759505954 Chr.12: 108657468-108657468 on Build GRCh38
rs377185786 Chr.12: 108657390-108657390 on Build GRCh38
rs769089003 Chr.12: 108657362-108657362 on Build GRCh38
rs767321878 Chr.12: 108657326-108657326 on Build GRCh38
rs772843096 Chr.12: 108657300-108657300 on Build GRCh38
rs372988394 Chr.12: 108657276-108657276 on Build GRCh38
rs1015544883 Chr.12: 108657299-108657299 on Build GRCh38
rs376380472 Chr.12: 108657460-108657460 on Build GRCh38
rs1005124973 Chr.12: 108657258-108657258 on Build GRCh38
rs750447950 Chr.12: 108657338-108657338 on Build GRCh38
rs766170351 Chr.12: 108657305-108657305 on Build GRCh38
rs769625321 Chr.12: 108657295-108657295 on Build GRCh38

View Full Product Details