Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001001391.1 | NP_001001391.1 | 6 - 7 | 1169 | 102 |
NM_001202557.1 | NP_001189486.1 | 6 - 7 | 1169 | 102 | |
XM_017018584.1 | XP_016874073.1 | 8 - 9 | 1151 | 102 | |
XM_017018585.1 | XP_016874074.1 | 7 - 8 | 1022 | 102 | |
GenBank mRNA | AK129808.1 | - | 5 - 6 | 564 | 102 |
AK290424.1 | - | 6 - 7 | 735 | 102 | |
AK297776.1 | - | 6 - 7 | 775 | 102 | |
AL832642.2 | - | 6 - 7 | 1169 | 102 | |
AY101193.1 | - | 6 - 7 | 852 | 102 | |
BC052287.1 | - | 5 - 6 | 723 | 102 | |
BC067348.1 | - | 6 - 7 | 852 | 102 | |
M24915.1 | - | 6 - 7 | 850 | 102 | |
M25078.1 | - | 6 - 7 | 733 | 102 | |
M59040.1 | - | 6 - 7 | 763 | 102 | |
U40373.1 | - | 6 - 7 | 799 | 102 |