Assay Design



Gene Transcripts

Gene Symbol
MARCKS
Entrez Gene ID
102139983
Gene Name
myristoylated alanine rich protein kinase C substrate
Gene Aliases
-
Chromosome Location
Chr.4: 17842467 - 17849220 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005551628.2 XP_005551685.1 1 - 2 560 61

View Full Product Details