Assay Design



Gene Transcripts

Gene Symbol
KRAS
Entrez Gene ID
101114005
Gene Name
KRAS proto-oncogene, GTPase
Gene Aliases
-
Chromosome Location
Chr.3: 203572870 - 203617003 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027967805.1 XP_027823606.1 4 - 5 638 100
XM_027967806.1 XP_027823607.1 4 - 5 639 100

View Full Product Details