Assay Design



Gene Transcripts

Gene Symbol
FTO
Entrez Gene ID
100125353
Gene Name
FTO alpha-ketoglutarate dependent dioxygenase
Gene Aliases
-
Chromosome Location
Chr.14: 23529695 - 23956080 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001104931.1 NP_001098401.1 6 - 7 1134 102
XM_027977360.1 XP_027833161.1 6 - 7 1231 102
GenBank mRNA EU072419.1 - 6 - 7 1134 102

View Full Product Details