Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022205.3 | NP_071541.2 | 2 - 2 | 1067 | 58 |
GenBank mRNA | AF452185.1 | - | 1 - 1 | 992 | 58 |
BC089804.1 | - | 2 - 2 | 1047 | 58 | |
U54791.1 | - | 3 - 3 | 984 | 58 | |
U90610.1 | - | 1 - 1 | 992 | 58 |