Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001107202.2 | NP_001100672.1 | 1 - 2 | 78 | 60 |
XM_008769857.1 | XP_008768079.1 | 1 - 2 | 368 | 60 | |
GenBank mRNA | EU448292.1 | - | 1 - 2 | 78 | 60 |
EU448293.1 | - | 1 - 2 | 78 | 60 |