Assay Design



Gene Transcripts

Gene Symbol
ak2
Entrez Gene ID
448011
Gene Name
adenylate kinase 2
Gene Aliases
adk2
Chromosome Location
Chr.2: 86231354 - 86242340 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004791.1 NP_001004791.1 1 - 2 138 85
XM_012956698.2 XP_012812152.1 1 - 2 259 85
GenBank mRNA BC074526.1 - 1 - 2 138 85
CR762157.2 - 1 - 2 109 85

View Full Product Details