Assay Design



Gene Transcripts

Gene Symbol
vhl
Entrez Gene ID
549121
Gene Name
von Hippel-Lindau tumor suppressor
Gene Aliases
hrca1, rca1, vhl1
Chromosome Location
Chr.4: 121310136 - 121313651 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016367.2 NP_001016367.1 2 - 3 366 64
GenBank mRNA BC157329.1 - 2 - 3 325 64
CR761285.2 - 2 - 3 366 64

View Full Product Details