Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 100992379-100992879 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGAP42
Entrez Gene ID
143872
Gene Name
Rho GTPase activating protein 42
Gene Aliases
GRAF3
UniGene
Hs.741465
Gene Chromosome Location
Chr.11: 100687676-100993941 on Build GRCh38
Transcripts
XM_011542615.2 XM_011542616.2 XM_011542617.2 XM_017017237.1 XM_017017238.1 NM_032021.2

Gene Symbol
TMEM133
Entrez Gene ID
83935
Gene Name
transmembrane protein 133
Gene Aliases
AD031
UniGene
Hs.44004
Gene Chromosome Location
Chr.11: 100992080-100993935 on Build GRCh38
Transcripts
XM_011542615.2 XM_011542616.2 XM_011542617.2 XM_017017237.1 XM_017017238.1 NM_032021.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3868244 c.223C>T p.P75S Chr.11: 100992531-100992531 on Build GRCh38
7394140 c.364G>T p.V122F Chr.11: 100992672-100992672 on Build GRCh38
3985848 c.353T>A p.L118H Chr.11: 100992661-100992661 on Build GRCh38
318128 c.77A>G p.Y26C Chr.11: 100992385-100992385 on Build GRCh38
7221519 c.157T>C p.F53L Chr.11: 100992465-100992465 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750148725 Chr.11: 100992653-100992653 on Build GRCh38
rs769459867 Chr.11: 100992601-100992601 on Build GRCh38
rs182992679 Chr.11: 100992749-100992749 on Build GRCh38
rs763212021 Chr.11: 100992621-100992621 on Build GRCh38
rs776205705 Chr.11: 100992710-100992710 on Build GRCh38
rs760216388 Chr.11: 100992438-100992438 on Build GRCh38
rs574827948 Chr.11: 100992594-100992594 on Build GRCh38
rs544581252 Chr.11: 100992566-100992566 on Build GRCh38
rs959616350 Chr.11: 100992684-100992684 on Build GRCh38
rs750829605 Chr.11: 100992418-100992418 on Build GRCh38
rs927935401 Chr.11: 100992438-100992438 on Build GRCh38
rs946860657 Chr.11: 100992489-100992489 on Build GRCh38
rs758280743 Chr.11: 100992564-100992564 on Build GRCh38
rs753439434 Chr.11: 100992526-100992526 on Build GRCh38
rs752162993 Chr.11: 100992534-100992534 on Build GRCh38
rs763623801 Chr.11: 100992508-100992509 on Build GRCh38
rs766787801 Chr.11: 100992530-100992530 on Build GRCh38
rs751287706 Chr.11: 100992745-100992745 on Build GRCh38
rs471933 Chr.11: 100992623-100992623 on Build GRCh38
rs760082580 Chr.11: 100992627-100992627 on Build GRCh38
rs755749939 Chr.11: 100992435-100992435 on Build GRCh38
rs755660357 Chr.11: 100992536-100992536 on Build GRCh38
rs943018105 Chr.11: 100992855-100992855 on Build GRCh38
rs149959826 Chr.11: 100992433-100992433 on Build GRCh38
rs763740402 Chr.11: 100992552-100992552 on Build GRCh38
rs201854990 Chr.11: 100992425-100992425 on Build GRCh38
rs772644869 Chr.11: 100992709-100992709 on Build GRCh38
rs776801663 Chr.11: 100992509-100992509 on Build GRCh38
rs748838150 Chr.11: 100992736-100992736 on Build GRCh38
rs148668544 Chr.11: 100992515-100992515 on Build GRCh38
rs766919060 Chr.11: 100992431-100992431 on Build GRCh38
rs761395129 Chr.11: 100992711-100992711 on Build GRCh38
rs774335734 Chr.11: 100992755-100992755 on Build GRCh38
rs890069113 Chr.11: 100992818-100992818 on Build GRCh38
rs368122394 Chr.11: 100992671-100992671 on Build GRCh38
rs771092001 Chr.11: 100992681-100992681 on Build GRCh38
rs777551663 Chr.11: 100992443-100992443 on Build GRCh38
rs763375873 Chr.11: 100992528-100992528 on Build GRCh38
rs990863129 Chr.11: 100992689-100992689 on Build GRCh38
rs765628634 Chr.11: 100992413-100992413 on Build GRCh38
rs185793193 Chr.11: 100992562-100992562 on Build GRCh38
rs779671371 Chr.11: 100992672-100992672 on Build GRCh38
rs77738491 Chr.11: 100992465-100992465 on Build GRCh38
rs773546322 Chr.11: 100992614-100992614 on Build GRCh38
rs771846350 Chr.11: 100992470-100992470 on Build GRCh38
rs768783424 Chr.11: 100992505-100992505 on Build GRCh38
rs751375719 Chr.11: 100992660-100992660 on Build GRCh38
rs774691586 Chr.11: 100992626-100992626 on Build GRCh38
rs950561025 Chr.11: 100992662-100992662 on Build GRCh38
rs1051347465 Chr.11: 100992806-100992806 on Build GRCh38
rs758108330 Chr.11: 100992655-100992655 on Build GRCh38
rs200387604 Chr.11: 100992567-100992567 on Build GRCh38
rs765077388 Chr.11: 100992733-100992734 on Build GRCh38
rs768609521 Chr.11: 100992587-100992587 on Build GRCh38
rs191585811 Chr.11: 100992746-100992746 on Build GRCh38
rs759445076 Chr.11: 100992765-100992765 on Build GRCh38
rs934183138 Chr.11: 100992787-100992787 on Build GRCh38
rs753344919 Chr.11: 100992635-100992635 on Build GRCh38
rs764852263 Chr.11: 100992724-100992724 on Build GRCh38
rs776906582 Chr.11: 100992410-100992410 on Build GRCh38
rs774810548 Chr.11: 100992696-100992696 on Build GRCh38
rs114921248 Chr.11: 100992563-100992563 on Build GRCh38
rs756907650 Chr.11: 100992609-100992610 on Build GRCh38
rs749001162 Chr.11: 100992445-100992445 on Build GRCh38
rs775502896 Chr.11: 100992487-100992487 on Build GRCh38
rs779856225 Chr.11: 100992658-100992658 on Build GRCh38
rs200190230 Chr.11: 100992434-100992434 on Build GRCh38
rs757090173 Chr.11: 100992452-100992452 on Build GRCh38
rs761412236 Chr.11: 100992637-100992637 on Build GRCh38
rs750014737 Chr.11: 100992726-100992726 on Build GRCh38
rs762154278 Chr.11: 100992412-100992412 on Build GRCh38
rs529720630 Chr.11: 100992422-100992422 on Build GRCh38
rs202128087 Chr.11: 100992407-100992407 on Build GRCh38
rs573469870 Chr.11: 100992457-100992457 on Build GRCh38
rs749243358 Chr.11: 100992405-100992405 on Build GRCh38
rs550953805 Chr.11: 100992760-100992763 on Build GRCh38
rs938059271 Chr.11: 100992459-100992459 on Build GRCh38
rs200444776 Chr.11: 100992675-100992675 on Build GRCh38
rs764794139 Chr.11: 100992648-100992648 on Build GRCh38
rs140313796 Chr.11: 100992667-100992667 on Build GRCh38
rs559489662 Chr.11: 100992676-100992676 on Build GRCh38
rs748239838 Chr.11: 100992593-100992593 on Build GRCh38
rs867337002 Chr.11: 100992537-100992537 on Build GRCh38
rs747056949 Chr.11: 100992503-100992503 on Build GRCh38
rs61995873 Chr.11: 100992588-100992588 on Build GRCh38
rs939795427 Chr.11: 100992632-100992632 on Build GRCh38
rs924074951 Chr.11: 100992781-100992781 on Build GRCh38
rs998397516 Chr.11: 100992542-100992542 on Build GRCh38
rs759996513 Chr.11: 100992708-100992708 on Build GRCh38
rs752062401 Chr.11: 100992473-100992473 on Build GRCh38
rs781576296 Chr.11: 100992553-100992553 on Build GRCh38

View Full Product Details