Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 114353789-114354294 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TBX5
Entrez Gene ID
6910
Gene Name
T-box 5
Gene Aliases
HOS
UniGene
Hs.381715
Gene Chromosome Location
Chr.12: 114353911-114408708 on Build GRCh38
Transcripts
NM_000192.3 NM_080717.2 NM_181486.2 XM_017019912.1

SNP Detail

SNP ID Chromosome Location
rs745499257 Chr.12: 114353915-114353915 on Build GRCh38
rs776263824 Chr.12: 114354207-114354214 on Build GRCh38
rs548364757 Chr.12: 114353910-114353910 on Build GRCh38
rs1017748384 Chr.12: 114353812-114353812 on Build GRCh38
rs747205317 Chr.12: 114353951-114353951 on Build GRCh38
rs75334514 Chr.12: 114354156-114354156 on Build GRCh38
rs949345090 Chr.12: 114354021-114354021 on Build GRCh38
rs1007498479 Chr.12: 114354204-114354204 on Build GRCh38
rs34050413 Chr.12: 114354000-114354001 on Build GRCh38
rs917558766 Chr.12: 114353987-114353987 on Build GRCh38
rs1031800606 Chr.12: 114354242-114354242 on Build GRCh38
rs946579484 Chr.12: 114354024-114354025 on Build GRCh38
rs953598474 Chr.12: 114353850-114353850 on Build GRCh38
rs925067842 Chr.12: 114353867-114353867 on Build GRCh38
rs542952088 Chr.12: 114354220-114354220 on Build GRCh38
rs935092424 Chr.12: 114353883-114353883 on Build GRCh38
rs576867150 Chr.12: 114354086-114354086 on Build GRCh38
rs202095845 Chr.12: 114354145-114354145 on Build GRCh38
rs905097059 Chr.12: 114354044-114354044 on Build GRCh38
rs201166554 Chr.12: 114354197-114354197 on Build GRCh38
rs187379531 Chr.12: 114354037-114354037 on Build GRCh38
rs895789702 Chr.12: 114354152-114354152 on Build GRCh38
rs147774482 Chr.12: 114354175-114354175 on Build GRCh38
rs183595902 Chr.12: 114354012-114354012 on Build GRCh38
rs373833293 Chr.12: 114354203-114354204 on Build GRCh38
rs935070500 Chr.12: 114353980-114353980 on Build GRCh38
rs558688524 Chr.12: 114354065-114354065 on Build GRCh38
rs879491404 Chr.12: 114353969-114353969 on Build GRCh38
rs886048983 Chr.12: 114354021-114354021 on Build GRCh38
rs576217372 Chr.12: 114353898-114353898 on Build GRCh38
rs968936964 Chr.12: 114354214-114354214 on Build GRCh38
rs939250279 Chr.12: 114354137-114354137 on Build GRCh38
rs201799304 Chr.12: 114354204-114354205 on Build GRCh38
rs59746696 Chr.12: 114353909-114353910 on Build GRCh38
rs890439813 Chr.12: 114354203-114354203 on Build GRCh38
rs978888146 Chr.12: 114353842-114353842 on Build GRCh38
rs987147790 Chr.12: 114353905-114353905 on Build GRCh38
rs767576629 Chr.12: 114353898-114353899 on Build GRCh38
rs192527148 Chr.12: 114354159-114354159 on Build GRCh38
rs199773239 Chr.12: 114354157-114354157 on Build GRCh38
rs1057065756 Chr.12: 114354138-114354138 on Build GRCh38
rs145772162 Chr.12: 114353899-114353899 on Build GRCh38
rs987881371 Chr.12: 114354269-114354269 on Build GRCh38
rs956668424 Chr.12: 114354246-114354246 on Build GRCh38
rs564562667 Chr.12: 114354052-114354055 on Build GRCh38
rs988216722 Chr.12: 114353907-114353907 on Build GRCh38
rs535072534 Chr.12: 114354201-114354201 on Build GRCh38
rs200407853 Chr.12: 114354147-114354147 on Build GRCh38
rs35534655 Chr.12: 114354147-114354147 on Build GRCh38
rs866303150 Chr.12: 114354213-114354213 on Build GRCh38
rs773894324 Chr.12: 114353826-114353826 on Build GRCh38

View Full Product Details