Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 114354172-114354686 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TBX5
Entrez Gene ID
6910
Gene Name
T-box 5
Gene Aliases
HOS
UniGene
Hs.381715
Gene Chromosome Location
Chr.12: 114353911-114408708 on Build GRCh38
Transcripts
NM_000192.3 NM_080717.2 NM_181486.2 XM_017019912.1

SNP Detail

SNP ID Chromosome Location
rs776263824 Chr.12: 114354207-114354214 on Build GRCh38
rs1000371364 Chr.12: 114354547-114354547 on Build GRCh38
rs6489956 Chr.12: 114354431-114354431 on Build GRCh38
rs1039008198 Chr.12: 114354477-114354477 on Build GRCh38
rs764387876 Chr.12: 114354513-114354513 on Build GRCh38
rs547907580 Chr.12: 114354567-114354567 on Build GRCh38
rs188337192 Chr.12: 114354466-114354466 on Build GRCh38
rs549280585 Chr.12: 114354447-114354447 on Build GRCh38
rs939281382 Chr.12: 114354556-114354556 on Build GRCh38
rs1009061481 Chr.12: 114354644-114354644 on Build GRCh38
rs987820559 Chr.12: 114354446-114354446 on Build GRCh38
rs1007498479 Chr.12: 114354204-114354204 on Build GRCh38
rs1031800606 Chr.12: 114354242-114354242 on Build GRCh38
rs561439634 Chr.12: 114354279-114354279 on Build GRCh38
rs917639246 Chr.12: 114354272-114354272 on Build GRCh38
rs940711927 Chr.12: 114354576-114354576 on Build GRCh38
rs557604192 Chr.12: 114354334-114354334 on Build GRCh38
rs542952088 Chr.12: 114354220-114354220 on Build GRCh38
rs926421576 Chr.12: 114354490-114354490 on Build GRCh38
rs184793497 Chr.12: 114354438-114354438 on Build GRCh38
rs904787360 Chr.12: 114354486-114354486 on Build GRCh38
rs201166554 Chr.12: 114354197-114354197 on Build GRCh38
rs1051881489 Chr.12: 114354460-114354460 on Build GRCh38
rs373833293 Chr.12: 114354203-114354204 on Build GRCh38
rs890525823 Chr.12: 114354461-114354461 on Build GRCh38
rs1018629580 Chr.12: 114354359-114354359 on Build GRCh38
rs141223729 Chr.12: 114354344-114354344 on Build GRCh38
rs536633001 Chr.12: 114354588-114354588 on Build GRCh38
rs773180889 Chr.12: 114354632-114354632 on Build GRCh38
rs968936964 Chr.12: 114354214-114354214 on Build GRCh38
rs201799304 Chr.12: 114354204-114354205 on Build GRCh38
rs184360838 Chr.12: 114354623-114354623 on Build GRCh38
rs886048984 Chr.12: 114354336-114354336 on Build GRCh38
rs890439813 Chr.12: 114354203-114354203 on Build GRCh38
rs571557978 Chr.12: 114354645-114354645 on Build GRCh38
rs946472215 Chr.12: 114354479-114354479 on Build GRCh38
rs970462931 Chr.12: 114354657-114354657 on Build GRCh38
rs777236533 Chr.12: 114354561-114354561 on Build GRCh38
rs886048985 Chr.12: 114354421-114354421 on Build GRCh38
rs532005395 Chr.12: 114354321-114354321 on Build GRCh38
rs1036352891 Chr.12: 114354579-114354580 on Build GRCh38
rs1007552123 Chr.12: 114354470-114354470 on Build GRCh38
rs886048986 Chr.12: 114354606-114354606 on Build GRCh38
rs987881371 Chr.12: 114354269-114354269 on Build GRCh38
rs956668424 Chr.12: 114354246-114354246 on Build GRCh38
rs1037545131 Chr.12: 114354352-114354352 on Build GRCh38
rs192445542 Chr.12: 114354469-114354469 on Build GRCh38
rs956254515 Chr.12: 114354587-114354587 on Build GRCh38
rs1029578483 Chr.12: 114354612-114354612 on Build GRCh38
rs926566163 Chr.12: 114354338-114354338 on Build GRCh38
rs899259963 Chr.12: 114354580-114354580 on Build GRCh38
rs535072534 Chr.12: 114354201-114354201 on Build GRCh38
rs886048987 Chr.12: 114354657-114354657 on Build GRCh38
rs566107581 Chr.12: 114354572-114354572 on Build GRCh38
rs866303150 Chr.12: 114354213-114354213 on Build GRCh38

View Full Product Details