Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 24267791-24268292 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RABGGTA
Entrez Gene ID
5875
Gene Name
Rab geranylgeranyltransferase alpha subunit
Gene Aliases
PTAR3
UniGene
Hs.377992
Gene Chromosome Location
Chr.14: 24265538-24271627 on Build GRCh38
Transcripts
NM_004581.5 NM_182836.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6140397 c.1223T>C p.F408S Chr.14: 24267883-24267883 on Build GRCh38
5562305 c.1135C>T p.P379S Chr.14: 24268122-24268122 on Build GRCh38
3983209 c.1059-5C>A p.? Chr.14: 24268203-24268203 on Build GRCh38
3983208 c.1147+1G>A p.? Chr.14: 24268109-24268109 on Build GRCh38
1369354 c.1178G>A p.R393Q Chr.14: 24267928-24267928 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs919824830 Chr.14: 24267909-24267909 on Build GRCh38
rs780777418 Chr.14: 24268081-24268081 on Build GRCh38
rs1019998418 Chr.14: 24268085-24268085 on Build GRCh38
rs750056428 Chr.14: 24268166-24268166 on Build GRCh38
rs369132931 Chr.14: 24267864-24267864 on Build GRCh38
rs745370644 Chr.14: 24267928-24267928 on Build GRCh38
rs779517321 Chr.14: 24267941-24267941 on Build GRCh38
rs760415836 Chr.14: 24267977-24267977 on Build GRCh38
rs763857742 Chr.14: 24267906-24267906 on Build GRCh38
rs982803784 Chr.14: 24267958-24267958 on Build GRCh38
rs369902107 Chr.14: 24267968-24267968 on Build GRCh38
rs565242422 Chr.14: 24268090-24268090 on Build GRCh38
rs773362886 Chr.14: 24268104-24268104 on Build GRCh38
rs374908021 Chr.14: 24268208-24268208 on Build GRCh38
rs917679312 Chr.14: 24267952-24267952 on Build GRCh38
rs1009758391 Chr.14: 24268048-24268048 on Build GRCh38
rs370695817 Chr.14: 24268074-24268074 on Build GRCh38
rs781610962 Chr.14: 24268207-24268207 on Build GRCh38
rs374107829 Chr.14: 24268094-24268094 on Build GRCh38
rs776574597 Chr.14: 24268145-24268145 on Build GRCh38
rs376355848 Chr.14: 24267967-24267967 on Build GRCh38
rs764918745 Chr.14: 24268060-24268060 on Build GRCh38
rs757025194 Chr.14: 24268007-24268007 on Build GRCh38
rs747593332 Chr.14: 24268080-24268080 on Build GRCh38
rs372539683 Chr.14: 24267902-24267902 on Build GRCh38
rs947761047 Chr.14: 24267969-24267969 on Build GRCh38
rs549921741 Chr.14: 24268034-24268034 on Build GRCh38
rs1027214823 Chr.14: 24268143-24268143 on Build GRCh38
rs768778011 Chr.14: 24267827-24267827 on Build GRCh38
rs769991394 Chr.14: 24267963-24267963 on Build GRCh38
rs776454625 Chr.14: 24267881-24267881 on Build GRCh38
rs1054461612 Chr.14: 24268018-24268018 on Build GRCh38
rs532947880 Chr.14: 24267845-24267845 on Build GRCh38
rs917565940 Chr.14: 24268226-24268226 on Build GRCh38
rs779105303 Chr.14: 24268231-24268231 on Build GRCh38
rs772140705 Chr.14: 24267844-24267844 on Build GRCh38
rs749916809 Chr.14: 24268071-24268071 on Build GRCh38
rs752438111 Chr.14: 24267893-24267893 on Build GRCh38
rs753497982 Chr.14: 24267917-24267917 on Build GRCh38
rs762482261 Chr.14: 24268167-24268167 on Build GRCh38
rs528953339 Chr.14: 24268095-24268095 on Build GRCh38
rs761644738 Chr.14: 24268150-24268150 on Build GRCh38
rs761736359 Chr.14: 24268248-24268248 on Build GRCh38
rs749349356 Chr.14: 24268108-24268108 on Build GRCh38
rs941502 Chr.14: 24267997-24267997 on Build GRCh38
rs778613072 Chr.14: 24267926-24267926 on Build GRCh38
rs377178792 Chr.14: 24268109-24268109 on Build GRCh38
rs984593427 Chr.14: 24268160-24268160 on Build GRCh38
rs747920468 Chr.14: 24267914-24267914 on Build GRCh38
rs565706719 Chr.14: 24268173-24268173 on Build GRCh38
rs764017079 Chr.14: 24267983-24267983 on Build GRCh38
rs549076050 Chr.14: 24268125-24268125 on Build GRCh38
rs201877221 Chr.14: 24268216-24268216 on Build GRCh38
rs756937084 Chr.14: 24267920-24267920 on Build GRCh38
rs745936154 Chr.14: 24268235-24268235 on Build GRCh38
rs969206244 Chr.14: 24268260-24268260 on Build GRCh38
rs12886219 Chr.14: 24268059-24268059 on Build GRCh38
rs748419997 Chr.14: 24267950-24267950 on Build GRCh38
rs991823286 Chr.14: 24267854-24267854 on Build GRCh38
rs765341896 Chr.14: 24267851-24267851 on Build GRCh38
rs755548071 Chr.14: 24268204-24268204 on Build GRCh38
rs767369266 Chr.14: 24267970-24267970 on Build GRCh38
rs770007363 Chr.14: 24268099-24268099 on Build GRCh38
rs760713919 Chr.14: 24268140-24268140 on Build GRCh38
rs776664277 Chr.14: 24268239-24268239 on Build GRCh38
rs904019268 Chr.14: 24267994-24267994 on Build GRCh38
rs750482140 Chr.14: 24267860-24267860 on Build GRCh38
rs771032580 Chr.14: 24268225-24268225 on Build GRCh38
rs755326489 Chr.14: 24267815-24267815 on Build GRCh38
rs769026751 Chr.14: 24267944-24267944 on Build GRCh38
rs768731521 Chr.14: 24268237-24268237 on Build GRCh38
rs749561397 Chr.14: 24268219-24268219 on Build GRCh38
rs370077003 Chr.14: 24268165-24268165 on Build GRCh38
rs775447807 Chr.14: 24267971-24267971 on Build GRCh38
rs776949619 Chr.14: 24267946-24267946 on Build GRCh38
rs921432911 Chr.14: 24267982-24267982 on Build GRCh38
rs1052625974 Chr.14: 24267998-24267998 on Build GRCh38
rs564834697 Chr.14: 24267839-24267839 on Build GRCh38
rs751022237 Chr.14: 24268191-24268191 on Build GRCh38
rs758836110 Chr.14: 24267929-24267929 on Build GRCh38
rs746119867 Chr.14: 24267837-24267838 on Build GRCh38
rs756421546 Chr.14: 24268215-24268215 on Build GRCh38

View Full Product Details