Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 60645623-60646167 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SIX1
Entrez Gene ID
6495
Gene Name
SIX homeobox 1
Gene Aliases
BOS3, DFNA23, TIP39
UniGene
Hs.633506
Gene Chromosome Location
Chr.14: 60644698-60649489 on Build GRCh38
Transcripts
NM_005982.3 XM_017021602.1

SNP Detail

SNP ID Chromosome Location
rs966192118 Chr.14: 60645718-60645718 on Build GRCh38
rs57517528 Chr.14: 60645794-60645794 on Build GRCh38
rs766323925 Chr.14: 60646115-60646115 on Build GRCh38
rs527420447 Chr.14: 60645814-60645814 on Build GRCh38
rs915931771 Chr.14: 60645920-60645920 on Build GRCh38
rs975898177 Chr.14: 60645751-60645751 on Build GRCh38
rs886050570 Chr.14: 60646004-60646007 on Build GRCh38
rs147081368 Chr.14: 60645986-60645986 on Build GRCh38
rs991454260 Chr.14: 60645881-60645881 on Build GRCh38
rs976613412 Chr.14: 60645970-60645970 on Build GRCh38
rs191393983 Chr.14: 60645880-60645880 on Build GRCh38
rs1049391881 Chr.14: 60645684-60645684 on Build GRCh38
rs1010631639 Chr.14: 60645737-60645737 on Build GRCh38
rs556041829 Chr.14: 60645702-60645705 on Build GRCh38
rs78909843 Chr.14: 60645681-60645681 on Build GRCh38
rs182923762 Chr.14: 60645918-60645918 on Build GRCh38
rs921680501 Chr.14: 60645768-60645768 on Build GRCh38
rs879084294 Chr.14: 60645964-60645964 on Build GRCh38
rs1012240423 Chr.14: 60645669-60645673 on Build GRCh38
rs186530769 Chr.14: 60645840-60645840 on Build GRCh38
rs895211253 Chr.14: 60645926-60645926 on Build GRCh38
rs548655754 Chr.14: 60646104-60646104 on Build GRCh38
rs751366842 Chr.14: 60646118-60646118 on Build GRCh38
rs61991660 Chr.14: 60646037-60646037 on Build GRCh38
rs962011450 Chr.14: 60646072-60646073 on Build GRCh38
rs557193382 Chr.14: 60645673-60645673 on Build GRCh38
rs1049363176 Chr.14: 60646017-60646017 on Build GRCh38
rs80194537 Chr.14: 60646018-60646018 on Build GRCh38
rs946416352 Chr.14: 60646082-60646082 on Build GRCh38
rs538935374 Chr.14: 60645912-60645912 on Build GRCh38
rs564879010 Chr.14: 60645803-60645803 on Build GRCh38
rs918820750 Chr.14: 60646145-60646145 on Build GRCh38
rs576067443 Chr.14: 60646094-60646094 on Build GRCh38
rs886050571 Chr.14: 60646122-60646122 on Build GRCh38
rs910154027 Chr.14: 60645833-60645838 on Build GRCh38
rs113680815 Chr.14: 60646139-60646139 on Build GRCh38
rs185694350 Chr.14: 60646005-60646005 on Build GRCh38
rs1022274842 Chr.14: 60645941-60645944 on Build GRCh38
rs893527067 Chr.14: 60645702-60645702 on Build GRCh38
rs999067967 Chr.14: 60645864-60645864 on Build GRCh38
rs908658203 Chr.14: 60645936-60645936 on Build GRCh38
rs869276383 Chr.14: 60646031-60646034 on Build GRCh38
rs984402964 Chr.14: 60645774-60645774 on Build GRCh38
rs986710111 Chr.14: 60646007-60646007 on Build GRCh38
rs908849819 Chr.14: 60646066-60646066 on Build GRCh38
rs544567737 Chr.14: 60645770-60645770 on Build GRCh38
rs76116881 Chr.14: 60645879-60645879 on Build GRCh38
rs367844913 Chr.14: 60646077-60646077 on Build GRCh38
rs984435706 Chr.14: 60646024-60646024 on Build GRCh38
rs766038211 Chr.14: 60646018-60646019 on Build GRCh38
rs976309744 Chr.14: 60646004-60646007 on Build GRCh38
rs568572700 Chr.14: 60646110-60646110 on Build GRCh38
rs1012269705 Chr.14: 60645933-60645933 on Build GRCh38
rs952712736 Chr.14: 60645921-60645921 on Build GRCh38
rs965942520 Chr.14: 60645997-60645997 on Build GRCh38
rs10144415 Chr.14: 60645949-60645949 on Build GRCh38
rs112733948 Chr.14: 60646018-60646018 on Build GRCh38
rs901127505 Chr.14: 60645658-60645658 on Build GRCh38
rs893455823 Chr.14: 60646069-60646069 on Build GRCh38
rs984599493 Chr.14: 60645924-60645924 on Build GRCh38
rs867880214 Chr.14: 60646034-60646034 on Build GRCh38

View Full Product Details