Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 60648850-60649340 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SIX1
Entrez Gene ID
6495
Gene Name
SIX homeobox 1
Gene Aliases
BOS3, DFNA23, TIP39
UniGene
Hs.633506
Gene Chromosome Location
Chr.14: 60644698-60649489 on Build GRCh38
Transcripts
NM_005982.3 XM_017021602.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1287939 c.100C>A p.L34M Chr.14: 60649090-60649090 on Build GRCh38
1300718 c.328C>T p.R110W Chr.14: 60648862-60648862 on Build GRCh38
4051585 c.313G>A p.A105T Chr.14: 60648877-60648877 on Build GRCh38
6554437 c.286G>A p.E96K Chr.14: 60648904-60648904 on Build GRCh38
4051586 c.87C>T p.R29R Chr.14: 60649103-60649103 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs564123011 Chr.14: 60649202-60649202 on Build GRCh38
rs911848334 Chr.14: 60649262-60649262 on Build GRCh38
rs760736490 Chr.14: 60649078-60649078 on Build GRCh38
rs763122547 Chr.14: 60648934-60648934 on Build GRCh38
rs748718182 Chr.14: 60649024-60649024 on Build GRCh38
rs751171137 Chr.14: 60649214-60649214 on Build GRCh38
rs1009804624 Chr.14: 60649117-60649117 on Build GRCh38
rs146357380 Chr.14: 60649106-60649106 on Build GRCh38
rs867031506 Chr.14: 60649116-60649116 on Build GRCh38
rs886050572 Chr.14: 60649273-60649273 on Build GRCh38
rs374228886 Chr.14: 60649203-60649203 on Build GRCh38
rs397515562 Chr.14: 60649140-60649140 on Build GRCh38
rs758038091 Chr.14: 60649115-60649115 on Build GRCh38
rs771057416 Chr.14: 60649176-60649176 on Build GRCh38
rs781574148 Chr.14: 60649122-60649122 on Build GRCh38
rs767354178 Chr.14: 60648954-60648954 on Build GRCh38
rs773333781 Chr.14: 60648908-60648908 on Build GRCh38
rs778375446 Chr.14: 60649167-60649167 on Build GRCh38
rs770875229 Chr.14: 60649046-60649046 on Build GRCh38
rs149166341 Chr.14: 60649058-60649058 on Build GRCh38
rs1045845044 Chr.14: 60648913-60648913 on Build GRCh38
rs904243084 Chr.14: 60649299-60649299 on Build GRCh38
rs749690789 Chr.14: 60649174-60649174 on Build GRCh38
rs777941410 Chr.14: 60649219-60649219 on Build GRCh38
rs1020503352 Chr.14: 60649034-60649034 on Build GRCh38
rs775477360 Chr.14: 60649192-60649192 on Build GRCh38
rs757701604 Chr.14: 60649225-60649225 on Build GRCh38
rs754869969 Chr.14: 60649120-60649120 on Build GRCh38
rs929170580 Chr.14: 60649242-60649242 on Build GRCh38
rs779091171 Chr.14: 60649226-60649226 on Build GRCh38
rs749329131 Chr.14: 60648884-60648884 on Build GRCh38
rs766600023 Chr.14: 60648945-60648945 on Build GRCh38
rs201700039 Chr.14: 60649213-60649213 on Build GRCh38
rs759906473 Chr.14: 60649177-60649177 on Build GRCh38
rs756474637 Chr.14: 60649215-60649215 on Build GRCh38
rs774637636 Chr.14: 60649047-60649047 on Build GRCh38
rs762246254 Chr.14: 60648899-60648899 on Build GRCh38
rs987341411 Chr.14: 60649256-60649256 on Build GRCh38
rs752537825 Chr.14: 60649076-60649076 on Build GRCh38
rs779056573 Chr.14: 60649206-60649218 on Build GRCh38
rs17850414 Chr.14: 60648895-60648895 on Build GRCh38
rs751225628 Chr.14: 60648984-60648984 on Build GRCh38
rs890107472 Chr.14: 60649007-60649007 on Build GRCh38
rs1032525136 Chr.14: 60649086-60649086 on Build GRCh38
rs941857857 Chr.14: 60648930-60648930 on Build GRCh38
rs981551544 Chr.14: 60649264-60649264 on Build GRCh38
rs200835641 Chr.14: 60648906-60648906 on Build GRCh38
rs973266865 Chr.14: 60648938-60648938 on Build GRCh38
rs756912951 Chr.14: 60648963-60648963 on Build GRCh38
rs1051653507 Chr.14: 60648999-60648999 on Build GRCh38
rs973618825 Chr.14: 60649217-60649217 on Build GRCh38
rs772231645 Chr.14: 60649228-60649228 on Build GRCh38
rs927343418 Chr.14: 60649270-60649270 on Build GRCh38
rs919138620 Chr.14: 60648977-60648977 on Build GRCh38
rs756109319 Chr.14: 60648959-60648959 on Build GRCh38
rs560779340 Chr.14: 60649237-60649237 on Build GRCh38
rs752695594 Chr.14: 60648955-60648955 on Build GRCh38
rs747379649 Chr.14: 60648869-60648869 on Build GRCh38
rs375002099 Chr.14: 60649310-60649310 on Build GRCh38
rs764103068 Chr.14: 60649091-60649091 on Build GRCh38
rs942997858 Chr.14: 60649273-60649273 on Build GRCh38
rs749901536 Chr.14: 60649207-60649207 on Build GRCh38
rs369025163 Chr.14: 60648947-60648947 on Build GRCh38
rs1001409064 Chr.14: 60649081-60649081 on Build GRCh38
rs544164466 Chr.14: 60649111-60649111 on Build GRCh38
rs770253091 Chr.14: 60649027-60649027 on Build GRCh38
rs529427457 Chr.14: 60649268-60649268 on Build GRCh38
rs781519092 Chr.14: 60649010-60649010 on Build GRCh38
rs904273350 Chr.14: 60649045-60649045 on Build GRCh38
rs893019043 Chr.14: 60649011-60649011 on Build GRCh38
rs753360604 Chr.14: 60648962-60648962 on Build GRCh38
rs893062393 Chr.14: 60649285-60649285 on Build GRCh38
rs937375427 Chr.14: 60649271-60649272 on Build GRCh38
rs767478646 Chr.14: 60649072-60649072 on Build GRCh38
rs533496677 Chr.14: 60649210-60649210 on Build GRCh38
rs756826384 Chr.14: 60649097-60649097 on Build GRCh38
rs761639693 Chr.14: 60649204-60649204 on Build GRCh38
rs769018187 Chr.14: 60649001-60649001 on Build GRCh38
rs777157929 Chr.14: 60648893-60648893 on Build GRCh38
rs1038685374 Chr.14: 60649294-60649294 on Build GRCh38
rs397515560 Chr.14: 60648873-60648873 on Build GRCh38
rs763114674 Chr.14: 60649040-60649040 on Build GRCh38
rs770093951 Chr.14: 60649166-60649166 on Build GRCh38
rs372978267 Chr.14: 60648982-60648982 on Build GRCh38
rs150550985 Chr.14: 60649028-60649028 on Build GRCh38
rs759405851 Chr.14: 60648949-60648949 on Build GRCh38
rs748487541 Chr.14: 60649129-60649129 on Build GRCh38
rs764866193 Chr.14: 60649206-60649206 on Build GRCh38
rs564171668 Chr.14: 60649310-60649310 on Build GRCh38
rs772394070 Chr.14: 60649178-60649178 on Build GRCh38
rs778653697 Chr.14: 60648974-60648974 on Build GRCh38

View Full Product Details