Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 151812265-151812765 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARL5A
Entrez Gene ID
26225
Gene Name
ADP ribosylation factor like GTPase 5A
Gene Aliases
ARFLP5, ARL5
UniGene
Hs.470233
Gene Chromosome Location
Chr.2: 151800966-151828495 on Build GRCh38
Transcripts
NM_001037174.1 NM_012097.3 NM_177985.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
441261 c.286A>G p.R96G Chr.2: 151812410-151812410 on Build GRCh38
1631339 c.292_293delAG p.R98fs*5 Chr.2: 151812403-151812404 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs774726314 Chr.2: 151812386-151812386 on Build GRCh38
rs1042035905 Chr.2: 151812675-151812676 on Build GRCh38
rs1028477472 Chr.2: 151812348-151812348 on Build GRCh38
rs776378418 Chr.2: 151812349-151812349 on Build GRCh38
rs72866497 Chr.2: 151812704-151812704 on Build GRCh38
rs141663338 Chr.2: 151812364-151812364 on Build GRCh38
rs186511618 Chr.2: 151812612-151812612 on Build GRCh38
rs781176772 Chr.2: 151812473-151812473 on Build GRCh38
rs902320822 Chr.2: 151812627-151812627 on Build GRCh38
rs764155367 Chr.2: 151812434-151812434 on Build GRCh38
rs757293540 Chr.2: 151812444-151812444 on Build GRCh38
rs761210059 Chr.2: 151812474-151812474 on Build GRCh38
rs952948687 Chr.2: 151812404-151812404 on Build GRCh38
rs2030589 Chr.2: 151812713-151812713 on Build GRCh38
rs914994247 Chr.2: 151812569-151812569 on Build GRCh38
rs143037153 Chr.2: 151812679-151812679 on Build GRCh38
rs976946459 Chr.2: 151812510-151812510 on Build GRCh38
rs1042153008 Chr.2: 151812588-151812588 on Build GRCh38
rs752893124 Chr.2: 151812426-151812426 on Build GRCh38
rs181068451 Chr.2: 151812549-151812549 on Build GRCh38
rs759905887 Chr.2: 151812389-151812389 on Build GRCh38
rs777879057 Chr.2: 151812326-151812326 on Build GRCh38
rs371386094 Chr.2: 151812562-151812562 on Build GRCh38
rs774540160 Chr.2: 151812641-151812641 on Build GRCh38
rs201148768 Chr.2: 151812363-151812363 on Build GRCh38
rs112757227 Chr.2: 151812602-151812602 on Build GRCh38
rs908731108 Chr.2: 151812451-151812451 on Build GRCh38
rs972736043 Chr.2: 151812520-151812520 on Build GRCh38
rs113804293 Chr.2: 151812736-151812736 on Build GRCh38
rs763359699 Chr.2: 151812377-151812377 on Build GRCh38
rs746757645 Chr.2: 151812346-151812346 on Build GRCh38
rs745773568 Chr.2: 151812331-151812331 on Build GRCh38
rs769633262 Chr.2: 151812487-151812487 on Build GRCh38
rs773494531 Chr.2: 151812415-151812415 on Build GRCh38
rs769912014 Chr.2: 151812403-151812406 on Build GRCh38
rs540461084 Chr.2: 151812342-151812342 on Build GRCh38
rs190949041 Chr.2: 151812548-151812548 on Build GRCh38
rs781410242 Chr.2: 151812313-151812313 on Build GRCh38
rs946487928 Chr.2: 151812575-151812575 on Build GRCh38
rs758945531 Chr.2: 151812584-151812584 on Build GRCh38
rs755164845 Chr.2: 151812315-151812316 on Build GRCh38
rs763559794 Chr.2: 151812306-151812306 on Build GRCh38
rs778209563 Chr.2: 151812674-151812674 on Build GRCh38
rs772467794 Chr.2: 151812307-151812307 on Build GRCh38
rs984285929 Chr.2: 151812438-151812438 on Build GRCh38
rs773015438 Chr.2: 151812489-151812489 on Build GRCh38
rs781351454 Chr.2: 151812346-151812349 on Build GRCh38
rs1012921891 Chr.2: 151812682-151812682 on Build GRCh38
rs772112525 Chr.2: 151812332-151812332 on Build GRCh38
rs550468173 Chr.2: 151812729-151812729 on Build GRCh38
rs1008322808 Chr.2: 151812723-151812723 on Build GRCh38
rs533011589 Chr.2: 151812627-151812627 on Build GRCh38
rs902415281 Chr.2: 151812303-151812303 on Build GRCh38
rs140774052 Chr.2: 151812511-151812511 on Build GRCh38
rs747821563 Chr.2: 151812356-151812356 on Build GRCh38
rs904131563 Chr.2: 151812680-151812680 on Build GRCh38
rs111462761 Chr.2: 151812324-151812324 on Build GRCh38
rs756139393 Chr.2: 151812321-151812321 on Build GRCh38
rs532438026 Chr.2: 151812373-151812373 on Build GRCh38
rs752948298 Chr.2: 151812320-151812320 on Build GRCh38
rs774931645 Chr.2: 151812664-151812664 on Build GRCh38
rs143420605 Chr.2: 151812358-151812358 on Build GRCh38
rs756366503 Chr.2: 151812430-151812430 on Build GRCh38
rs1034953760 Chr.2: 151812344-151812344 on Build GRCh38
rs575123700 Chr.2: 151812701-151812701 on Build GRCh38
rs904080137 Chr.2: 151812319-151812319 on Build GRCh38
rs779970615 Chr.2: 151812447-151812447 on Build GRCh38
rs755312411 Chr.2: 151812308-151812308 on Build GRCh38
rs200412626 Chr.2: 151812478-151812478 on Build GRCh38
rs932285014 Chr.2: 151812519-151812519 on Build GRCh38
rs767891575 Chr.2: 151812403-151812403 on Build GRCh38
rs930565246 Chr.2: 151812567-151812567 on Build GRCh38
rs777801328 Chr.2: 151812453-151812454 on Build GRCh38
rs556499378 Chr.2: 151812292-151812292 on Build GRCh38
rs754750157 Chr.2: 151812471-151812471 on Build GRCh38
rs746948665 Chr.2: 151812464-151812464 on Build GRCh38
rs961577642 Chr.2: 151812496-151812496 on Build GRCh38
rs917642128 Chr.2: 151812667-151812668 on Build GRCh38
rs748250229 Chr.2: 151812374-151812375 on Build GRCh38
rs766851868 Chr.2: 151812378-151812378 on Build GRCh38
rs766825561 Chr.2: 151812607-151812607 on Build GRCh38

View Full Product Details