Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 62818433-62818936 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COL9A3
Entrez Gene ID
1299
Gene Name
collagen type IX alpha 3
Gene Aliases
DJ885L7.4.1, EDM3, IDD, MED
UniGene
Hs.716639
Gene Chromosome Location
Chr.20: 62817062-62841159 on Build GRCh38
Transcripts
NM_001853.3 XM_011528545.1 XM_017027666.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3548755 c.167G>A p.G56D Chr.20: 62818537-62818537 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs116110757 Chr.20: 62818817-62818817 on Build GRCh38
rs918003200 Chr.20: 62818698-62818698 on Build GRCh38
rs551197936 Chr.20: 62818608-62818608 on Build GRCh38
rs201752661 Chr.20: 62818503-62818503 on Build GRCh38
rs2734537 Chr.20: 62818813-62818813 on Build GRCh38
rs368800943 Chr.20: 62818455-62818455 on Build GRCh38
rs190016881 Chr.20: 62818464-62818464 on Build GRCh38
rs773812813 Chr.20: 62818490-62818490 on Build GRCh38
rs879578267 Chr.20: 62818633-62818633 on Build GRCh38
rs570174430 Chr.20: 62818670-62818670 on Build GRCh38
rs149582526 Chr.20: 62818552-62818552 on Build GRCh38
rs760414801 Chr.20: 62818527-62818527 on Build GRCh38
rs1046829213 Chr.20: 62818791-62818791 on Build GRCh38
rs886056907 Chr.20: 62818513-62818513 on Build GRCh38
rs978212094 Chr.20: 62818708-62818708 on Build GRCh38
rs944323095 Chr.20: 62818770-62818781 on Build GRCh38
rs199641605 Chr.20: 62818479-62818479 on Build GRCh38
rs1026561603 Chr.20: 62818452-62818452 on Build GRCh38
rs748832180 Chr.20: 62818497-62818497 on Build GRCh38
rs113675030 Chr.20: 62818700-62818700 on Build GRCh38
rs567371176 Chr.20: 62818553-62818553 on Build GRCh38
rs1041112800 Chr.20: 62818799-62818799 on Build GRCh38
rs758518434 Chr.20: 62818537-62818537 on Build GRCh38
rs1020139754 Chr.20: 62818482-62818482 on Build GRCh38
rs563109268 Chr.20: 62818878-62818878 on Build GRCh38
rs1019302727 Chr.20: 62818622-62818622 on Build GRCh38
rs768884138 Chr.20: 62818623-62818623 on Build GRCh38
rs893071184 Chr.20: 62818461-62818461 on Build GRCh38
rs555983196 Chr.20: 62818714-62818714 on Build GRCh38
rs779101349 Chr.20: 62818576-62818576 on Build GRCh38
rs770284701 Chr.20: 62818501-62818501 on Build GRCh38
rs1040718519 Chr.20: 62818750-62818750 on Build GRCh38
rs764051012 Chr.20: 62818528-62818528 on Build GRCh38
rs533541118 Chr.20: 62818895-62818895 on Build GRCh38
rs558413179 Chr.20: 62818642-62818642 on Build GRCh38
rs936538399 Chr.20: 62818713-62818713 on Build GRCh38
rs755347516 Chr.20: 62818758-62818758 on Build GRCh38
rs781734370 Chr.20: 62818476-62818476 on Build GRCh38
rs1883850 Chr.20: 62818872-62818872 on Build GRCh38
rs961539779 Chr.20: 62818519-62818519 on Build GRCh38
rs115714551 Chr.20: 62818651-62818651 on Build GRCh38
rs757442656 Chr.20: 62818843-62818843 on Build GRCh38
rs766431560 Chr.20: 62818548-62818548 on Build GRCh38
rs775086653 Chr.20: 62818604-62818604 on Build GRCh38
rs771464531 Chr.20: 62818576-62818576 on Build GRCh38
rs751658593 Chr.20: 62818549-62818549 on Build GRCh38
rs544932957 Chr.20: 62818775-62818775 on Build GRCh38
rs747720580 Chr.20: 62818754-62818754 on Build GRCh38
rs118107952 Chr.20: 62818852-62818852 on Build GRCh38
rs994314447 Chr.20: 62818811-62818811 on Build GRCh38
rs551652191 Chr.20: 62818911-62818911 on Build GRCh38
rs374073790 Chr.20: 62818586-62818586 on Build GRCh38
rs572385789 Chr.20: 62818806-62818806 on Build GRCh38
rs773772804 Chr.20: 62818578-62818578 on Build GRCh38
rs1006539495 Chr.20: 62818608-62818608 on Build GRCh38
rs139504978 Chr.20: 62818811-62818812 on Build GRCh38
rs606231367 Chr.20: 62818517-62818517 on Build GRCh38
rs182337989 Chr.20: 62818617-62818617 on Build GRCh38
rs775311995 Chr.20: 62818521-62818521 on Build GRCh38
rs1882742 Chr.20: 62818876-62818876 on Build GRCh38
rs1049724877 Chr.20: 62818472-62818472 on Build GRCh38
rs1006419090 Chr.20: 62818805-62818805 on Build GRCh38
rs1049671954 Chr.20: 62818753-62818753 on Build GRCh38
rs540176484 Chr.20: 62818635-62818635 on Build GRCh38
rs767327106 Chr.20: 62818650-62818650 on Build GRCh38
rs753652134 Chr.20: 62818531-62818531 on Build GRCh38
rs559017038 Chr.20: 62818792-62818792 on Build GRCh38
rs748496265 Chr.20: 62818477-62818477 on Build GRCh38
rs942366191 Chr.20: 62818719-62818719 on Build GRCh38
rs990843743 Chr.20: 62818692-62818692 on Build GRCh38
rs112169318 Chr.20: 62818787-62818787 on Build GRCh38
rs771733147 Chr.20: 62818603-62818603 on Build GRCh38
rs959348363 Chr.20: 62818685-62818685 on Build GRCh38
rs555439610 Chr.20: 62818655-62818655 on Build GRCh38
rs763592460 Chr.20: 62818564-62818564 on Build GRCh38
rs556748862 Chr.20: 62818645-62818645 on Build GRCh38
rs2294987 Chr.20: 62818737-62818737 on Build GRCh38
rs767124418 Chr.20: 62818515-62818515 on Build GRCh38
rs928386093 Chr.20: 62818694-62818694 on Build GRCh38
rs773898020 Chr.20: 62818502-62818502 on Build GRCh38
rs949229114 Chr.20: 62818511-62818511 on Build GRCh38
rs989051646 Chr.20: 62818709-62818709 on Build GRCh38
rs144876240 Chr.20: 62818573-62818573 on Build GRCh38
rs536836788 Chr.20: 62818460-62818460 on Build GRCh38

View Full Product Details