Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 107824378-107824866 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SGMS2
Entrez Gene ID
166929
Gene Name
sphingomyelin synthase 2
Gene Aliases
SMS2
UniGene
Hs.595423
Gene Chromosome Location
Chr.4: 107823645-107915047 on Build GRCh38
Transcripts
NM_001136258.1 XM_011531701.2 XM_011531702.2

SNP Detail

SNP ID Chromosome Location
rs541832977 Chr.4: 107824640-107824640 on Build GRCh38
rs987565105 Chr.4: 107824580-107824580 on Build GRCh38
rs1036287265 Chr.4: 107824477-107824477 on Build GRCh38
rs972341655 Chr.4: 107824700-107824700 on Build GRCh38
rs1018111474 Chr.4: 107824784-107824784 on Build GRCh38
rs561801476 Chr.4: 107824646-107824646 on Build GRCh38
rs763179141 Chr.4: 107824587-107824587 on Build GRCh38
rs1026538193 Chr.4: 107824746-107824746 on Build GRCh38
rs893953693 Chr.4: 107824827-107824827 on Build GRCh38
rs1052250841 Chr.4: 107824662-107824662 on Build GRCh38
rs146873588 Chr.4: 107824573-107824573 on Build GRCh38
rs925223439 Chr.4: 107824617-107824617 on Build GRCh38
rs541182266 Chr.4: 107824848-107824848 on Build GRCh38
rs1013277596 Chr.4: 107824460-107824460 on Build GRCh38
rs944332698 Chr.4: 107824781-107824781 on Build GRCh38
rs1046576475 Chr.4: 107824684-107824684 on Build GRCh38
rs901343621 Chr.4: 107824576-107824576 on Build GRCh38
rs963654406 Chr.4: 107824555-107824555 on Build GRCh38
rs972265078 Chr.4: 107824561-107824561 on Build GRCh38
rs930554743 Chr.4: 107824493-107824493 on Build GRCh38
rs940488100 Chr.4: 107824465-107824465 on Build GRCh38
rs556257848 Chr.4: 107824566-107824566 on Build GRCh38
rs527595531 Chr.4: 107824796-107824796 on Build GRCh38
rs927833773 Chr.4: 107824428-107824428 on Build GRCh38
rs764280695 Chr.4: 107824802-107824802 on Build GRCh38
rs910613520 Chr.4: 107824586-107824586 on Build GRCh38
rs920552132 Chr.4: 107824479-107824479 on Build GRCh38
rs1008660531 Chr.4: 107824568-107824568 on Build GRCh38
rs969350871 Chr.4: 107824635-107824635 on Build GRCh38
rs930586047 Chr.4: 107824732-107824732 on Build GRCh38
rs987596471 Chr.4: 107824780-107824780 on Build GRCh38
rs982583008 Chr.4: 107824404-107824404 on Build GRCh38
rs753273988 Chr.4: 107824704-107824704 on Build GRCh38
rs1001838782 Chr.4: 107824517-107824522 on Build GRCh38
rs1011143279 Chr.4: 107824598-107824598 on Build GRCh38
rs1023658166 Chr.4: 107824620-107824620 on Build GRCh38
rs371254469 Chr.4: 107824669-107824670 on Build GRCh38
rs1000140530 Chr.4: 107824581-107824581 on Build GRCh38
rs1052625959 Chr.4: 107824808-107824808 on Build GRCh38
rs982227487 Chr.4: 107824648-107824648 on Build GRCh38
rs756626334 Chr.4: 107824764-107824764 on Build GRCh38
rs1031164498 Chr.4: 107824590-107824590 on Build GRCh38
rs765941692 Chr.4: 107824593-107824593 on Build GRCh38
rs949199891 Chr.4: 107824680-107824680 on Build GRCh38
rs373163406 Chr.4: 107824677-107824677 on Build GRCh38
rs1040036485 Chr.4: 107824789-107824789 on Build GRCh38
rs1050082398 Chr.4: 107824518-107824518 on Build GRCh38
rs919406312 Chr.4: 107824569-107824569 on Build GRCh38
rs899435316 Chr.4: 107824720-107824720 on Build GRCh38
rs749988817 Chr.4: 107824402-107824402 on Build GRCh38
rs892242094 Chr.4: 107824453-107824453 on Build GRCh38
rs979197425 Chr.4: 107824813-107824813 on Build GRCh38
rs910225280 Chr.4: 107824768-107824768 on Build GRCh38
rs993736789 Chr.4: 107824722-107824722 on Build GRCh38
rs1016936729 Chr.4: 107824534-107824534 on Build GRCh38
rs934411322 Chr.4: 107824800-107824800 on Build GRCh38
rs962151822 Chr.4: 107824690-107824690 on Build GRCh38

View Full Product Details