Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 147653710-147654231 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRMT9
Entrez Gene ID
90826
Gene Name
protein arginine methyltransferase 9
Gene Aliases
PRMT10
UniGene
Hs.591692
Gene Chromosome Location
Chr.4: 147635626-147684230 on Build GRCh38
Transcripts
NM_001304458.1 NM_138364.3 XM_017008823.1 XM_017008824.1 XM_017008825.1 XM_017008826.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4929744 c.1755A>T p.L585F Chr.4: 147654142-147654142 on Build GRCh38
6756256 c.1749C>T p.Y583Y Chr.4: 147654148-147654148 on Build GRCh38
4122626 c.1868C>T p.S623F Chr.4: 147654029-147654029 on Build GRCh38
4717867 c.2004G>C p.G668G Chr.4: 147653893-147653893 on Build GRCh38
1427431 c.1891G>T p.E631* Chr.4: 147654006-147654006 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs371580166 Chr.4: 147654147-147654147 on Build GRCh38
rs374569514 Chr.4: 147654168-147654168 on Build GRCh38
rs772640938 Chr.4: 147654163-147654163 on Build GRCh38
rs538790926 Chr.4: 147653871-147653871 on Build GRCh38
rs138969454 Chr.4: 147653920-147653920 on Build GRCh38
rs959482458 Chr.4: 147653734-147653734 on Build GRCh38
rs767676759 Chr.4: 147653919-147653919 on Build GRCh38
rs747020217 Chr.4: 147654169-147654169 on Build GRCh38
rs775818432 Chr.4: 147653980-147653981 on Build GRCh38
rs200063176 Chr.4: 147653880-147653880 on Build GRCh38
rs184937144 Chr.4: 147653772-147653772 on Build GRCh38
rs112218554 Chr.4: 147654198-147654198 on Build GRCh38
rs141544929 Chr.4: 147654057-147654057 on Build GRCh38
rs866338815 Chr.4: 147654122-147654122 on Build GRCh38
rs201607320 Chr.4: 147654107-147654107 on Build GRCh38
rs751553388 Chr.4: 147654111-147654111 on Build GRCh38
rs201067391 Chr.4: 147654148-147654148 on Build GRCh38
rs768326884 Chr.4: 147653828-147653828 on Build GRCh38
rs917726892 Chr.4: 147653995-147653995 on Build GRCh38
rs368548509 Chr.4: 147653810-147653811 on Build GRCh38
rs368438347 Chr.4: 147654194-147654194 on Build GRCh38
rs749570123 Chr.4: 147654048-147654048 on Build GRCh38
rs142877020 Chr.4: 147653897-147653897 on Build GRCh38
rs758383196 Chr.4: 147653939-147653939 on Build GRCh38
rs747773557 Chr.4: 147653873-147653873 on Build GRCh38
rs914716907 Chr.4: 147653950-147653950 on Build GRCh38
rs977783806 Chr.4: 147653907-147653907 on Build GRCh38
rs779628896 Chr.4: 147653821-147653821 on Build GRCh38
rs747860731 Chr.4: 147654015-147654015 on Build GRCh38
rs767015993 Chr.4: 147654119-147654119 on Build GRCh38
rs1015127152 Chr.4: 147653822-147653822 on Build GRCh38
rs1049965186 Chr.4: 147654016-147654016 on Build GRCh38
rs755818378 Chr.4: 147654132-147654132 on Build GRCh38
rs367655758 Chr.4: 147654138-147654138 on Build GRCh38
rs1053963742 Chr.4: 147653942-147653942 on Build GRCh38
rs932840710 Chr.4: 147653951-147653951 on Build GRCh38
rs780664768 Chr.4: 147654011-147654011 on Build GRCh38
rs775041237 Chr.4: 147654052-147654052 on Build GRCh38
rs779924735 Chr.4: 147653949-147653949 on Build GRCh38
rs370542261 Chr.4: 147653926-147653926 on Build GRCh38
rs775708248 Chr.4: 147654058-147654058 on Build GRCh38
rs766224844 Chr.4: 147654209-147654209 on Build GRCh38
rs923607161 Chr.4: 147653909-147653909 on Build GRCh38
rs80141801 Chr.4: 147653899-147653899 on Build GRCh38
rs773441385 Chr.4: 147653879-147653879 on Build GRCh38
rs936394458 Chr.4: 147653928-147653928 on Build GRCh38
rs772591492 Chr.4: 147653811-147653811 on Build GRCh38
rs768749114 Chr.4: 147654172-147654172 on Build GRCh38
rs150904021 Chr.4: 147654079-147654079 on Build GRCh38
rs760795444 Chr.4: 147653923-147653923 on Build GRCh38
rs776765372 Chr.4: 147654177-147654177 on Build GRCh38
rs111243466 Chr.4: 147654196-147654196 on Build GRCh38
rs778218066 Chr.4: 147654045-147654045 on Build GRCh38
rs777254663 Chr.4: 147654133-147654133 on Build GRCh38
rs746446242 Chr.4: 147654150-147654150 on Build GRCh38
rs760098991 Chr.4: 147654056-147654056 on Build GRCh38
rs754529740 Chr.4: 147654003-147654003 on Build GRCh38
rs890627316 Chr.4: 147653795-147653795 on Build GRCh38
rs199616123 Chr.4: 147653875-147653875 on Build GRCh38
rs771503214 Chr.4: 147654051-147654051 on Build GRCh38
rs751465462 Chr.4: 147653979-147653979 on Build GRCh38
rs139380764 Chr.4: 147653855-147653855 on Build GRCh38
rs748609949 Chr.4: 147653748-147653752 on Build GRCh38
rs769260235 Chr.4: 147654034-147654034 on Build GRCh38
rs541704391 Chr.4: 147654105-147654105 on Build GRCh38
rs752086737 Chr.4: 147654128-147654128 on Build GRCh38
rs990991506 Chr.4: 147653961-147653961 on Build GRCh38
rs758263957 Chr.4: 147653810-147653810 on Build GRCh38
rs374390428 Chr.4: 147654101-147654101 on Build GRCh38
rs556897380 Chr.4: 147653826-147653826 on Build GRCh38
rs750118595 Chr.4: 147653932-147653932 on Build GRCh38
rs139425779 Chr.4: 147654117-147654117 on Build GRCh38
rs763516738 Chr.4: 147653895-147653895 on Build GRCh38

View Full Product Details