Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 147654122-147654634 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRMT9
Entrez Gene ID
90826
Gene Name
protein arginine methyltransferase 9
Gene Aliases
PRMT10
UniGene
Hs.591692
Gene Chromosome Location
Chr.4: 147635626-147684230 on Build GRCh38
Transcripts
NM_001304458.1 NM_138364.3 XM_017008823.1 XM_017008824.1 XM_017008825.1 XM_017008826.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6756257 c.1614A>C p.K538N Chr.4: 147654283-147654283 on Build GRCh38
6756258 c.1540G>A p.V514I Chr.4: 147654357-147654357 on Build GRCh38
447460 c.1565C>G p.S522C Chr.4: 147654332-147654332 on Build GRCh38
447461 c.1563A>G p.E521E Chr.4: 147654334-147654334 on Build GRCh38
6756256 c.1749C>T p.Y583Y Chr.4: 147654148-147654148 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs371580166 Chr.4: 147654147-147654147 on Build GRCh38
rs768581346 Chr.4: 147654512-147654512 on Build GRCh38
rs774061559 Chr.4: 147654227-147654227 on Build GRCh38
rs878944258 Chr.4: 147654291-147654291 on Build GRCh38
rs199919200 Chr.4: 147654471-147654471 on Build GRCh38
rs771827266 Chr.4: 147654385-147654385 on Build GRCh38
rs374569514 Chr.4: 147654168-147654168 on Build GRCh38
rs772640938 Chr.4: 147654163-147654163 on Build GRCh38
rs1024888817 Chr.4: 147654281-147654281 on Build GRCh38
rs777819680 Chr.4: 147654602-147654602 on Build GRCh38
rs111341835 Chr.4: 147654523-147654523 on Build GRCh38
rs762688689 Chr.4: 147654581-147654581 on Build GRCh38
rs747020217 Chr.4: 147654169-147654169 on Build GRCh38
rs752287364 Chr.4: 147654242-147654242 on Build GRCh38
rs768674064 Chr.4: 147654339-147654339 on Build GRCh38
rs112470944 Chr.4: 147654365-147654365 on Build GRCh38
rs747171225 Chr.4: 147654508-147654509 on Build GRCh38
rs761571001 Chr.4: 147654404-147654404 on Build GRCh38
rs766654676 Chr.4: 147654583-147654583 on Build GRCh38
rs995070350 Chr.4: 147654411-147654411 on Build GRCh38
rs944272685 Chr.4: 147654422-147654422 on Build GRCh38
rs371609584 Chr.4: 147654321-147654321 on Build GRCh38
rs775341687 Chr.4: 147654387-147654387 on Build GRCh38
rs753108180 Chr.4: 147654597-147654597 on Build GRCh38
rs112218554 Chr.4: 147654198-147654198 on Build GRCh38
rs766002384 Chr.4: 147654270-147654270 on Build GRCh38
rs754447510 Chr.4: 147654553-147654553 on Build GRCh38
rs202062787 Chr.4: 147654282-147654282 on Build GRCh38
rs149240722 Chr.4: 147654342-147654342 on Build GRCh38
rs528315267 Chr.4: 147654421-147654421 on Build GRCh38
rs199827425 Chr.4: 147654266-147654266 on Build GRCh38
rs201067391 Chr.4: 147654148-147654148 on Build GRCh38
rs139529106 Chr.4: 147654483-147654483 on Build GRCh38
rs747526289 Chr.4: 147654324-147654324 on Build GRCh38
rs200858338 Chr.4: 147654488-147654488 on Build GRCh38
rs774566430 Chr.4: 147654322-147654322 on Build GRCh38
rs748371032 Chr.4: 147654359-147654359 on Build GRCh38
rs112950828 Chr.4: 147654233-147654233 on Build GRCh38
rs959307938 Chr.4: 147654425-147654427 on Build GRCh38
rs368438347 Chr.4: 147654194-147654194 on Build GRCh38
rs764934847 Chr.4: 147654416-147654416 on Build GRCh38
rs148661515 Chr.4: 147654554-147654554 on Build GRCh38
rs542882645 Chr.4: 147654384-147654384 on Build GRCh38
rs763639663 Chr.4: 147654255-147654255 on Build GRCh38
rs188327569 Chr.4: 147654236-147654236 on Build GRCh38
rs749581527 Chr.4: 147654605-147654605 on Build GRCh38
rs113446925 Chr.4: 147654273-147654273 on Build GRCh38
rs760650287 Chr.4: 147654388-147654388 on Build GRCh38
rs867679649 Chr.4: 147654529-147654529 on Build GRCh38
rs768973785 Chr.4: 147654588-147654588 on Build GRCh38
rs372927933 Chr.4: 147654528-147654528 on Build GRCh38
rs921916521 Chr.4: 147654599-147654599 on Build GRCh38
rs746178040 Chr.4: 147654301-147654301 on Build GRCh38
rs766224844 Chr.4: 147654209-147654209 on Build GRCh38
rs866781466 Chr.4: 147654253-147654253 on Build GRCh38
rs1037261522 Chr.4: 147654439-147654439 on Build GRCh38
rs750156332 Chr.4: 147654580-147654580 on Build GRCh38
rs548422720 Chr.4: 147654459-147654459 on Build GRCh38
rs1025134447 Chr.4: 147654480-147654480 on Build GRCh38
rs755457525 Chr.4: 147654596-147654596 on Build GRCh38
rs780451725 Chr.4: 147654442-147654442 on Build GRCh38
rs1012343545 Chr.4: 147654526-147654526 on Build GRCh38
rs774151090 Chr.4: 147654376-147654376 on Build GRCh38
rs781141997 Chr.4: 147654465-147654465 on Build GRCh38
rs768749114 Chr.4: 147654172-147654172 on Build GRCh38
rs769941986 Chr.4: 147654374-147654374 on Build GRCh38
rs17023638 Chr.4: 147654450-147654450 on Build GRCh38
rs111243466 Chr.4: 147654196-147654196 on Build GRCh38
rs759003872 Chr.4: 147654303-147654303 on Build GRCh38
rs776765372 Chr.4: 147654177-147654177 on Build GRCh38
rs753175170 Chr.4: 147654598-147654598 on Build GRCh38
rs145133883 Chr.4: 147654497-147654497 on Build GRCh38
rs746446242 Chr.4: 147654150-147654150 on Build GRCh38
rs761165977 Chr.4: 147654608-147654608 on Build GRCh38
rs779425651 Chr.4: 147654289-147654289 on Build GRCh38
rs147098467 Chr.4: 147654222-147654222 on Build GRCh38
rs759478496 Chr.4: 147654230-147654230 on Build GRCh38
rs761448742 Chr.4: 147654564-147654564 on Build GRCh38
rs771823588 Chr.4: 147654500-147654500 on Build GRCh38
rs767910171 Chr.4: 147654577-147654577 on Build GRCh38
rs751891437 Chr.4: 147654587-147654587 on Build GRCh38
rs147603635 Chr.4: 147654508-147654508 on Build GRCh38
rs756239657 Chr.4: 147654600-147654600 on Build GRCh38
rs753324013 Chr.4: 147654389-147654389 on Build GRCh38

View Full Product Details