Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 66995172-66995668 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SBDS
Entrez Gene ID
51119
Gene Name
SBDS ribosome assembly guanine nucleotide exchange factor
Gene Aliases
CGI-97, SDS, SWDS
UniGene
Hs.110445
Gene Chromosome Location
Chr.7: 66987703-66995601 on Build GRCh38
Transcripts
NM_016038.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5430191 c.27G>A p.Q9Q Chr.7: 66995391-66995391 on Build GRCh38
5407821 c.3G>A p.M1I Chr.7: 66995415-66995415 on Build GRCh38
4826086 c.98A>T p.K33I Chr.7: 66995320-66995320 on Build GRCh38
4463649 c.12C>T p.F4F Chr.7: 66995406-66995406 on Build GRCh38
4820653 c.72G>A p.G24G Chr.7: 66995346-66995346 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772797192 Chr.7: 66995405-66995405 on Build GRCh38
rs553080928 Chr.7: 66995374-66995374 on Build GRCh38
rs561588829 Chr.7: 66995581-66995581 on Build GRCh38
rs182989808 Chr.7: 66995433-66995433 on Build GRCh38
rs376037363 Chr.7: 66995572-66995572 on Build GRCh38
rs377469960 Chr.7: 66995445-66995445 on Build GRCh38
rs900288596 Chr.7: 66995528-66995528 on Build GRCh38
rs560159813 Chr.7: 66995251-66995252 on Build GRCh38
rs559213058 Chr.7: 66995570-66995570 on Build GRCh38
rs927409928 Chr.7: 66995390-66995390 on Build GRCh38
rs955974464 Chr.7: 66995361-66995361 on Build GRCh38
rs779874549 Chr.7: 66995243-66995243 on Build GRCh38
rs748342404 Chr.7: 66995344-66995344 on Build GRCh38
rs891388098 Chr.7: 66995627-66995627 on Build GRCh38
rs113993990 Chr.7: 66995299-66995299 on Build GRCh38
rs897405945 Chr.7: 66995450-66995450 on Build GRCh38
rs537763716 Chr.7: 66995282-66995282 on Build GRCh38
rs771720952 Chr.7: 66995395-66995395 on Build GRCh38
rs776303630 Chr.7: 66995307-66995307 on Build GRCh38
rs770801583 Chr.7: 66995461-66995461 on Build GRCh38
rs11557409 Chr.7: 66995386-66995386 on Build GRCh38
rs766277488 Chr.7: 66995377-66995377 on Build GRCh38
rs60712404 Chr.7: 66995210-66995210 on Build GRCh38
rs377748611 Chr.7: 66995250-66995250 on Build GRCh38
rs370287930 Chr.7: 66995468-66995468 on Build GRCh38
rs891263695 Chr.7: 66995512-66995512 on Build GRCh38
rs918613592 Chr.7: 66995619-66995619 on Build GRCh38
rs751329484 Chr.7: 66995378-66995378 on Build GRCh38
rs757728020 Chr.7: 66995403-66995403 on Build GRCh38
rs1044385030 Chr.7: 66995193-66995193 on Build GRCh38
rs536606303 Chr.7: 66995364-66995364 on Build GRCh38
rs988703244 Chr.7: 66995568-66995568 on Build GRCh38
rs1005565290 Chr.7: 66995213-66995213 on Build GRCh38
rs759370247 Chr.7: 66995379-66995379 on Build GRCh38
rs886039431 Chr.7: 66995298-66995298 on Build GRCh38
rs367806793 Chr.7: 66995615-66995615 on Build GRCh38
rs767281778 Chr.7: 66995381-66995381 on Build GRCh38
rs1051189433 Chr.7: 66995511-66995511 on Build GRCh38
rs147652512 Chr.7: 66995291-66995291 on Build GRCh38
rs930258806 Chr.7: 66995476-66995476 on Build GRCh38
rs774593246 Chr.7: 66995352-66995352 on Build GRCh38
rs971186617 Chr.7: 66995574-66995574 on Build GRCh38
rs763783384 Chr.7: 66995306-66995306 on Build GRCh38
rs760974918 Chr.7: 66995407-66995407 on Build GRCh38
rs551982113 Chr.7: 66995220-66995220 on Build GRCh38
rs766151644 Chr.7: 66995331-66995331 on Build GRCh38
rs1051249966 Chr.7: 66995625-66995625 on Build GRCh38
rs544584268 Chr.7: 66995580-66995580 on Build GRCh38
rs1038994411 Chr.7: 66995236-66995236 on Build GRCh38
rs771692204 Chr.7: 66995290-66995290 on Build GRCh38
rs780550927 Chr.7: 66995434-66995434 on Build GRCh38
rs371417670 Chr.7: 66995281-66995281 on Build GRCh38
rs879198009 Chr.7: 66995536-66995536 on Build GRCh38
rs781338918 Chr.7: 66995342-66995342 on Build GRCh38
rs749830607 Chr.7: 66995348-66995348 on Build GRCh38
rs548436732 Chr.7: 66995311-66995311 on Build GRCh38
rs756239598 Chr.7: 66995345-66995345 on Build GRCh38
rs777899269 Chr.7: 66995346-66995346 on Build GRCh38
rs187584171 Chr.7: 66995447-66995447 on Build GRCh38
rs1000261392 Chr.7: 66995533-66995533 on Build GRCh38
rs755203268 Chr.7: 66995341-66995341 on Build GRCh38
rs781250001 Chr.7: 66995276-66995276 on Build GRCh38
rs1032706368 Chr.7: 66995541-66995541 on Build GRCh38
rs1009037678 Chr.7: 66995310-66995310 on Build GRCh38
rs28942099 Chr.7: 66995394-66995394 on Build GRCh38
rs771403283 Chr.7: 66995349-66995349 on Build GRCh38
rs776462097 Chr.7: 66995362-66995362 on Build GRCh38
rs370999538 Chr.7: 66995577-66995577 on Build GRCh38
rs575024588 Chr.7: 66995587-66995587 on Build GRCh38
rs768457164 Chr.7: 66995247-66995248 on Build GRCh38
rs559698532 Chr.7: 66995624-66995624 on Build GRCh38
rs539111398 Chr.7: 66995524-66995524 on Build GRCh38
rs11557408 Chr.7: 66995369-66995369 on Build GRCh38
rs568696356 Chr.7: 66995234-66995234 on Build GRCh38
rs772772005 Chr.7: 66995372-66995372 on Build GRCh38
rs1017731057 Chr.7: 66995527-66995527 on Build GRCh38
rs755185140 Chr.7: 66995252-66995252 on Build GRCh38
rs145800188 Chr.7: 66995382-66995382 on Build GRCh38
rs768454020 Chr.7: 66995358-66995358 on Build GRCh38
rs756327627 Chr.7: 66995391-66995391 on Build GRCh38
rs772607723 Chr.7: 66995452-66995452 on Build GRCh38
rs761310052 Chr.7: 66995295-66995295 on Build GRCh38
rs753899308 Chr.7: 66995398-66995398 on Build GRCh38
rs746186710 Chr.7: 66995356-66995356 on Build GRCh38
rs777720287 Chr.7: 66995393-66995393 on Build GRCh38
rs771259728 Chr.7: 66995284-66995284 on Build GRCh38
rs751298432 Chr.7: 66995335-66995335 on Build GRCh38
rs955922561 Chr.7: 66995546-66995546 on Build GRCh38
rs761366712 Chr.7: 66995309-66995309 on Build GRCh38
rs762472602 Chr.7: 66995458-66995458 on Build GRCh38
rs758341690 Chr.7: 66995329-66995329 on Build GRCh38
rs146516249 Chr.7: 66995634-66995634 on Build GRCh38
rs1007064843 Chr.7: 66995518-66995518 on Build GRCh38
rs373730800 Chr.7: 66995320-66995320 on Build GRCh38
rs540271027 Chr.7: 66995591-66995591 on Build GRCh38
rs11557406 Chr.7: 66995499-66995499 on Build GRCh38
rs746346017 Chr.7: 66995429-66995429 on Build GRCh38

View Full Product Details