Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 104126653-104127152 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ORC5
Entrez Gene ID
5001
Gene Name
origin recognition complex subunit 5
Gene Aliases
ORC5L, ORC5P, ORC5T, PPP1R117
UniGene
Hs.432948
Gene Chromosome Location
Chr.7: 104126341-104208047 on Build GRCh38
Transcripts
NM_002553.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3084373 c.1307G>A p.*436* Chr.7: 104126849-104126849 on Build GRCh38
7381921 c.1305G>T p.L435F Chr.7: 104126851-104126851 on Build GRCh38
4773070 c.1269G>T p.V423V Chr.7: 104126887-104126887 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs888519338 Chr.7: 104127028-104127028 on Build GRCh38
rs1028411500 Chr.7: 104126752-104126752 on Build GRCh38
rs933048305 Chr.7: 104126866-104126866 on Build GRCh38
rs185550733 Chr.7: 104127008-104127008 on Build GRCh38
rs372461864 Chr.7: 104126805-104126805 on Build GRCh38
rs1003578782 Chr.7: 104126963-104126963 on Build GRCh38
rs775866356 Chr.7: 104126800-104126800 on Build GRCh38
rs552036789 Chr.7: 104127057-104127057 on Build GRCh38
rs1016006880 Chr.7: 104127077-104127077 on Build GRCh38
rs77261166 Chr.7: 104127091-104127092 on Build GRCh38
rs754653353 Chr.7: 104126903-104126903 on Build GRCh38
rs1042263162 Chr.7: 104126702-104126702 on Build GRCh38
rs777518705 Chr.7: 104126946-104126946 on Build GRCh38
rs61749914 Chr.7: 104126884-104126884 on Build GRCh38
rs141952883 Chr.7: 104126890-104126890 on Build GRCh38
rs868555419 Chr.7: 104126758-104126758 on Build GRCh38
rs752138068 Chr.7: 104126910-104126910 on Build GRCh38
rs747186349 Chr.7: 104126801-104126801 on Build GRCh38
rs372324427 Chr.7: 104126748-104126748 on Build GRCh38
rs902444904 Chr.7: 104126708-104126708 on Build GRCh38
rs200275033 Chr.7: 104126813-104126813 on Build GRCh38
rs755595702 Chr.7: 104126911-104126911 on Build GRCh38
rs543548100 Chr.7: 104126971-104126971 on Build GRCh38
rs79011570 Chr.7: 104127079-104127079 on Build GRCh38
rs777156623 Chr.7: 104126916-104126916 on Build GRCh38
rs181033863 Chr.7: 104126905-104126905 on Build GRCh38
rs751233901 Chr.7: 104126970-104126970 on Build GRCh38
rs561084708 Chr.7: 104126939-104126939 on Build GRCh38
rs776660982 Chr.7: 104126826-104126826 on Build GRCh38
rs2307411 Chr.7: 104126749-104126756 on Build GRCh38
rs969028671 Chr.7: 104127061-104127061 on Build GRCh38
rs959330766 Chr.7: 104127004-104127004 on Build GRCh38
rs567081465 Chr.7: 104127102-104127102 on Build GRCh38
rs758153083 Chr.7: 104126936-104126936 on Build GRCh38
rs74901297 Chr.7: 104127078-104127078 on Build GRCh38
rs369829260 Chr.7: 104126857-104126857 on Build GRCh38
rs61756141 Chr.7: 104126891-104126891 on Build GRCh38
rs547289518 Chr.7: 104127018-104127024 on Build GRCh38
rs768863016 Chr.7: 104126941-104126941 on Build GRCh38
rs531396615 Chr.7: 104126962-104126962 on Build GRCh38
rs897187605 Chr.7: 104127078-104127081 on Build GRCh38
rs746350561 Chr.7: 104126920-104126920 on Build GRCh38
rs528812315 Chr.7: 104126994-104126995 on Build GRCh38
rs372350893 Chr.7: 104127005-104127005 on Build GRCh38
rs553813959 Chr.7: 104126783-104126783 on Build GRCh38
rs762019851 Chr.7: 104126854-104126854 on Build GRCh38
rs977305857 Chr.7: 104127119-104127119 on Build GRCh38
rs375796375 Chr.7: 104126832-104126832 on Build GRCh38
rs545276866 Chr.7: 104126782-104126782 on Build GRCh38
rs946358973 Chr.7: 104126928-104126928 on Build GRCh38
rs914394142 Chr.7: 104126763-104126763 on Build GRCh38
rs765882428 Chr.7: 104126822-104126834 on Build GRCh38
rs958652835 Chr.7: 104126716-104126716 on Build GRCh38
rs775716038 Chr.7: 104126910-104126912 on Build GRCh38
rs537957416 Chr.7: 104126747-104126747 on Build GRCh38
rs772416964 Chr.7: 104126799-104126799 on Build GRCh38
rs773143254 Chr.7: 104126874-104126874 on Build GRCh38
rs1008847170 Chr.7: 104126730-104126730 on Build GRCh38
rs780374882 Chr.7: 104126938-104126938 on Build GRCh38
rs577993299 Chr.7: 104126750-104126750 on Build GRCh38

View Full Product Details