Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 151232407-151232909 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHPF2
Entrez Gene ID
54480
Gene Name
chondroitin polymerizing factor 2
Gene Aliases
CSGLCA-T, CSGlcAT, ChSy-3, chPF-2
UniGene
Hs.647084
Gene Chromosome Location
Chr.7: 151232489-151238827 on Build GRCh38
Transcripts
NM_001284295.1 NM_019015.2

SNP Detail

SNP ID Chromosome Location
rs1032398552 Chr.7: 151232735-151232735 on Build GRCh38
rs938578314 Chr.7: 151232549-151232549 on Build GRCh38
rs925294842 Chr.7: 151232536-151232536 on Build GRCh38
rs541293329 Chr.7: 151232433-151232433 on Build GRCh38
rs941677863 Chr.7: 151232588-151232588 on Build GRCh38
rs540073449 Chr.7: 151232869-151232869 on Build GRCh38
rs767896569 Chr.7: 151232746-151232746 on Build GRCh38
rs562135561 Chr.7: 151232575-151232575 on Build GRCh38
rs114879520 Chr.7: 151232880-151232880 on Build GRCh38
rs1039027116 Chr.7: 151232623-151232623 on Build GRCh38
rs979760286 Chr.7: 151232538-151232538 on Build GRCh38
rs958038544 Chr.7: 151232539-151232539 on Build GRCh38
rs865803453 Chr.7: 151232551-151232551 on Build GRCh38
rs576513829 Chr.7: 151232635-151232635 on Build GRCh38
rs900484540 Chr.7: 151232654-151232654 on Build GRCh38
rs550517268 Chr.7: 151232511-151232511 on Build GRCh38
rs998165966 Chr.7: 151232633-151232633 on Build GRCh38
rs764681672 Chr.7: 151232837-151232837 on Build GRCh38
rs926976375 Chr.7: 151232544-151232544 on Build GRCh38
rs1006181992 Chr.7: 151232472-151232472 on Build GRCh38
rs999629064 Chr.7: 151232685-151232685 on Build GRCh38
rs902665029 Chr.7: 151232658-151232658 on Build GRCh38
rs868080882 Chr.7: 151232677-151232677 on Build GRCh38
rs1030501339 Chr.7: 151232518-151232518 on Build GRCh38
rs374377569 Chr.7: 151232786-151232786 on Build GRCh38
rs968876758 Chr.7: 151232537-151232537 on Build GRCh38
rs929120056 Chr.7: 151232849-151232849 on Build GRCh38
rs1030583012 Chr.7: 151232717-151232717 on Build GRCh38
rs997784815 Chr.7: 151232515-151232515 on Build GRCh38
rs549452233 Chr.7: 151232567-151232567 on Build GRCh38
rs556475529 Chr.7: 151232750-151232758 on Build GRCh38
rs942122041 Chr.7: 151232606-151232606 on Build GRCh38
rs994431082 Chr.7: 151232447-151232447 on Build GRCh38
rs372900058 Chr.7: 151232458-151232458 on Build GRCh38
rs956177162 Chr.7: 151232520-151232520 on Build GRCh38
rs1045899690 Chr.7: 151232494-151232494 on Build GRCh38
rs993207258 Chr.7: 151232507-151232507 on Build GRCh38
rs905503052 Chr.7: 151232513-151232514 on Build GRCh38
rs929087285 Chr.7: 151232574-151232574 on Build GRCh38
rs1047799649 Chr.7: 151232455-151232455 on Build GRCh38
rs533633612 Chr.7: 151232479-151232479 on Build GRCh38
rs878856555 Chr.7: 151232842-151232842 on Build GRCh38
rs1034085493 Chr.7: 151232778-151232778 on Build GRCh38
rs529311095 Chr.7: 151232528-151232528 on Build GRCh38
rs534798730 Chr.7: 151232605-151232605 on Build GRCh38
rs929822847 Chr.7: 151232572-151232572 on Build GRCh38
rs752645441 Chr.7: 151232841-151232841 on Build GRCh38
rs968469118 Chr.7: 151232774-151232774 on Build GRCh38
rs997544913 Chr.7: 151232706-151232706 on Build GRCh38
rs905332592 Chr.7: 151232830-151232830 on Build GRCh38
rs112985049 Chr.7: 151232755-151232755 on Build GRCh38
rs183298564 Chr.7: 151232881-151232881 on Build GRCh38
rs563632120 Chr.7: 151232516-151232516 on Build GRCh38
rs906924650 Chr.7: 151232499-151232499 on Build GRCh38
rs892094227 Chr.7: 151232724-151232724 on Build GRCh38
rs766205044 Chr.7: 151232771-151232771 on Build GRCh38
rs900620412 Chr.7: 151232882-151232882 on Build GRCh38
rs754486091 Chr.7: 151232827-151232827 on Build GRCh38
rs139900977 Chr.7: 151232852-151232852 on Build GRCh38
rs566067599 Chr.7: 151232579-151232579 on Build GRCh38
rs948551507 Chr.7: 151232493-151232493 on Build GRCh38
rs573574420 Chr.7: 151232859-151232859 on Build GRCh38
rs1056959783 Chr.7: 151232566-151232566 on Build GRCh38
rs959687051 Chr.7: 151232782-151232782 on Build GRCh38
rs980278595 Chr.7: 151232775-151232775 on Build GRCh38
rs773502782 Chr.7: 151232748-151232748 on Build GRCh38
rs989420902 Chr.7: 151232524-151232524 on Build GRCh38
rs551125196 Chr.7: 151232653-151232653 on Build GRCh38
rs887431185 Chr.7: 151232512-151232512 on Build GRCh38
rs978156154 Chr.7: 151232532-151232532 on Build GRCh38
rs970034128 Chr.7: 151232794-151232794 on Build GRCh38
rs761971375 Chr.7: 151232509-151232509 on Build GRCh38
rs948564415 Chr.7: 151232553-151232553 on Build GRCh38

View Full Product Details