Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 21974314-21974804 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDKN2A
Entrez Gene ID
1029
Gene Name
cyclin dependent kinase inhibitor 2A
Gene Aliases
ARF, CDK4I, CDKN2, CMM2, INK4, INK4A, MLM, MTS-1, MTS1, P14, P14ARF, P16, P16-INK4A, P16INK4, P16INK4A, P19, P19ARF, TP16
UniGene
Hs.512599
Gene Chromosome Location
Chr.9: 21967752-21995043 on Build GRCh38
Transcripts
NM_000077.4 NM_001195132.1 NM_058195.3 NM_058197.4 XM_011517675.2 XM_011517676.2 XM_011517679.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
20969 c.131A>C p.Y44S Chr.9: 21974697-21974697 on Build GRCh38
1731785 c.150+1G>C p.? Chr.9: 21974677-21974677 on Build GRCh38
3395756 c.150G>T p.Q50H Chr.9: 21974678-21974678 on Build GRCh38
5628983 c.67G>A p.G23S Chr.9: 21974761-21974761 on Build GRCh38
13260 c.150+15T>C p.? Chr.9: 21974663-21974663 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs587782206 Chr.9: 21974778-21974781 on Build GRCh38
rs752731682 Chr.9: 21974713-21974713 on Build GRCh38
rs199907548 Chr.9: 21974682-21974682 on Build GRCh38
rs774140115 Chr.9: 21974434-21974434 on Build GRCh38
rs150201743 Chr.9: 21974398-21974398 on Build GRCh38
rs1057517587 Chr.9: 21974485-21974485 on Build GRCh38
rs781066926 Chr.9: 21974785-21974785 on Build GRCh38
rs147602781 Chr.9: 21974462-21974462 on Build GRCh38
rs766772030 Chr.9: 21974759-21974759 on Build GRCh38
rs781780454 Chr.9: 21974549-21974549 on Build GRCh38
rs745827714 Chr.9: 21974734-21974734 on Build GRCh38
rs779983201 Chr.9: 21974665-21974665 on Build GRCh38
rs757066045 Chr.9: 21974725-21974725 on Build GRCh38
rs369946603 Chr.9: 21974657-21974657 on Build GRCh38
rs762876108 Chr.9: 21974427-21974427 on Build GRCh38
rs201820199 Chr.9: 21974460-21974462 on Build GRCh38
rs557891718 Chr.9: 21974668-21974668 on Build GRCh38
rs780022223 Chr.9: 21974562-21974562 on Build GRCh38
rs141480348 Chr.9: 21974480-21974480 on Build GRCh38
rs730881673 Chr.9: 21974696-21974697 on Build GRCh38
rs1016841954 Chr.9: 21974717-21974717 on Build GRCh38
rs750771226 Chr.9: 21974634-21974634 on Build GRCh38
rs587778189 Chr.9: 21974679-21974679 on Build GRCh38
rs765985763 Chr.9: 21974485-21974488 on Build GRCh38
rs752389002 Chr.9: 21974629-21974629 on Build GRCh38
rs45456595 Chr.9: 21974641-21974641 on Build GRCh38
rs759140308 Chr.9: 21974536-21974536 on Build GRCh38
rs200504822 Chr.9: 21974452-21974457 on Build GRCh38
rs143013864 Chr.9: 21974633-21974633 on Build GRCh38
rs749488253 Chr.9: 21974472-21974472 on Build GRCh38
rs104894097 Chr.9: 21974757-21974757 on Build GRCh38
rs767404338 Chr.9: 21974438-21974438 on Build GRCh38
rs762508111 Chr.9: 21974651-21974651 on Build GRCh38
rs745999521 Chr.9: 21974466-21974466 on Build GRCh38
rs747994882 Chr.9: 21974574-21974574 on Build GRCh38
rs759239111 Chr.9: 21974595-21974598 on Build GRCh38
rs149117035 Chr.9: 21974548-21974548 on Build GRCh38
rs769762355 Chr.9: 21974467-21974467 on Build GRCh38
rs147705603 Chr.9: 21974388-21974388 on Build GRCh38
rs864622263 Chr.9: 21974781-21974781 on Build GRCh38
rs935044558 Chr.9: 21974362-21974362 on Build GRCh38
rs768877674 Chr.9: 21974476-21974476 on Build GRCh38
rs746834149 Chr.9: 21974724-21974724 on Build GRCh38
rs780580671 Chr.9: 21974639-21974639 on Build GRCh38
rs938648422 Chr.9: 21974491-21974491 on Build GRCh38
rs770859592 Chr.9: 21974599-21974599 on Build GRCh38
rs537813612 Chr.9: 21974647-21974647 on Build GRCh38
rs730881672 Chr.9: 21974781-21974782 on Build GRCh38
rs780780607 Chr.9: 21974726-21974726 on Build GRCh38
rs754366579 Chr.9: 21974705-21974705 on Build GRCh38
rs771851528 Chr.9: 21974581-21974581 on Build GRCh38
rs200129494 Chr.9: 21974464-21974464 on Build GRCh38
rs373407950 Chr.9: 21974706-21974706 on Build GRCh38
rs751809654 Chr.9: 21974778-21974778 on Build GRCh38
rs551479808 Chr.9: 21974451-21974451 on Build GRCh38
rs772933026 Chr.9: 21974585-21974585 on Build GRCh38
rs886457271 Chr.9: 21974463-21974463 on Build GRCh38
rs182946652 Chr.9: 21974425-21974425 on Build GRCh38
rs749350928 Chr.9: 21974565-21974565 on Build GRCh38
rs535365274 Chr.9: 21974588-21974588 on Build GRCh38
rs761079352 Chr.9: 21974700-21974700 on Build GRCh38
rs386733552 Chr.9: 21974452-21974464 on Build GRCh38
rs764362225 Chr.9: 21974777-21974777 on Build GRCh38
rs796315946 Chr.9: 21974696-21974697 on Build GRCh38
rs752308489 Chr.9: 21974611-21974611 on Build GRCh38
rs759061855 Chr.9: 21974615-21974615 on Build GRCh38
rs760065045 Chr.9: 21974770-21974770 on Build GRCh38
rs764592691 Chr.9: 21974356-21974356 on Build GRCh38
rs773755761 Chr.9: 21974689-21974689 on Build GRCh38
rs764909269 Chr.9: 21974538-21974538 on Build GRCh38
rs372058542 Chr.9: 21974523-21974523 on Build GRCh38
rs145143907 Chr.9: 21974439-21974439 on Build GRCh38
rs753316964 Chr.9: 21974507-21974508 on Build GRCh38
rs748780473 Chr.9: 21974754-21974754 on Build GRCh38
rs386833399 Chr.9: 21974602-21974602 on Build GRCh38
rs878853644 Chr.9: 21974677-21974677 on Build GRCh38
rs775558394 Chr.9: 21974426-21974426 on Build GRCh38
rs549407023 Chr.9: 21974400-21974400 on Build GRCh38
rs778019498 Chr.9: 21974461-21974461 on Build GRCh38
rs763804037 Chr.9: 21974685-21974685 on Build GRCh38
rs1057517604 Chr.9: 21974638-21974638 on Build GRCh38
rs878853650 Chr.9: 21974733-21974733 on Build GRCh38
rs777576283 Chr.9: 21974653-21974653 on Build GRCh38
rs139807099 Chr.9: 21974644-21974644 on Build GRCh38
rs1044987386 Chr.9: 21974406-21974406 on Build GRCh38
rs765702324 Chr.9: 21974774-21974774 on Build GRCh38
rs374921006 Chr.9: 21974764-21974764 on Build GRCh38
rs763804673 Chr.9: 21974428-21974428 on Build GRCh38
rs928711188 Chr.9: 21974483-21974483 on Build GRCh38
rs777948908 Chr.9: 21974722-21974722 on Build GRCh38
rs531646614 Chr.9: 21974608-21974608 on Build GRCh38
rs377665008 Chr.9: 21974484-21974484 on Build GRCh38
rs398123152 Chr.9: 21974721-21974722 on Build GRCh38
rs139116263 Chr.9: 21974380-21974380 on Build GRCh38
rs374087901 Chr.9: 21974469-21974469 on Build GRCh38
rs267602199 Chr.9: 21974555-21974555 on Build GRCh38
rs150447516 Chr.9: 21974525-21974525 on Build GRCh38
rs766113466 Chr.9: 21974440-21974440 on Build GRCh38
rs786204195 Chr.9: 21974686-21974686 on Build GRCh38
rs138424891 Chr.9: 21974632-21974632 on Build GRCh38
rs1025333664 Chr.9: 21974672-21974672 on Build GRCh38
rs200382984 Chr.9: 21974721-21974721 on Build GRCh38
rs764853658 Chr.9: 21974618-21974618 on Build GRCh38
rs864622484 Chr.9: 21974769-21974769 on Build GRCh38
rs752016743 Chr.9: 21974545-21974545 on Build GRCh38
rs761014328 Chr.9: 21974758-21974758 on Build GRCh38
rs864622439 Chr.9: 21974711-21974711 on Build GRCh38
rs965114521 Chr.9: 21974355-21974355 on Build GRCh38
rs758455611 Chr.9: 21974776-21974776 on Build GRCh38
rs775176191 Chr.9: 21974745-21974745 on Build GRCh38
rs779087809 Chr.9: 21974568-21974568 on Build GRCh38
rs540871544 Chr.9: 21974741-21974741 on Build GRCh38
rs779174268 Chr.9: 21974646-21974646 on Build GRCh38
rs373185790 Chr.9: 21974437-21974437 on Build GRCh38
rs200738474 Chr.9: 21974681-21974681 on Build GRCh38
rs145697272 Chr.9: 21974410-21974410 on Build GRCh38
rs876659638 Chr.9: 21974684-21974684 on Build GRCh38
rs140984614 Chr.9: 21974395-21974395 on Build GRCh38
rs75256133 Chr.9: 21974526-21974526 on Build GRCh38
rs138677674 Chr.9: 21974783-21974783 on Build GRCh38
rs761567237 Chr.9: 21974502-21974502 on Build GRCh38
rs876658556 Chr.9: 21974784-21974784 on Build GRCh38
rs190519792 Chr.9: 21974370-21974370 on Build GRCh38
rs560518923 Chr.9: 21974775-21974775 on Build GRCh38
rs539257071 Chr.9: 21974520-21974520 on Build GRCh38
rs879254078 Chr.9: 21974739-21974739 on Build GRCh38
rs189126115 Chr.9: 21974464-21974464 on Build GRCh38
rs898369498 Chr.9: 21974424-21974424 on Build GRCh38
rs761763765 Chr.9: 21974436-21974436 on Build GRCh38
rs753508262 Chr.9: 21974602-21974607 on Build GRCh38
rs864622636 Chr.9: 21974680-21974680 on Build GRCh38
rs980682752 Chr.9: 21974753-21974753 on Build GRCh38
rs767492367 Chr.9: 21974508-21974508 on Build GRCh38
rs762288239 Chr.9: 21974438-21974440 on Build GRCh38
rs766854048 Chr.9: 21974701-21974701 on Build GRCh38
rs776810546 Chr.9: 21974762-21974762 on Build GRCh38
rs876658895 Chr.9: 21974746-21974746 on Build GRCh38
rs1057517608 Chr.9: 21974573-21974574 on Build GRCh38
rs768802553 Chr.9: 21974567-21974567 on Build GRCh38
rs746788029 Chr.9: 21974656-21974656 on Build GRCh38
rs765321737 Chr.9: 21974707-21974707 on Build GRCh38
rs144845109 Chr.9: 21974637-21974637 on Build GRCh38
rs550846229 Chr.9: 21974658-21974658 on Build GRCh38
rs750973473 Chr.9: 21974550-21974550 on Build GRCh38

View Full Product Details