Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 32418036-32418545 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACO1
Entrez Gene ID
48
Gene Name
aconitase 1
Gene Aliases
ACONS, HEL60, IREB1, IREBP, IREBP1, IRP1
UniGene
Hs.567229
Gene Chromosome Location
Chr.9: 32384603-32450836 on Build GRCh38
Transcripts
NM_001278352.1 NM_002197.2 XM_005251476.1 XM_011517888.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5583241 c.488C>T p.A163V Chr.9: 32418341-32418341 on Build GRCh38
1108086 c.423G>T p.K141N Chr.9: 32418146-32418146 on Build GRCh38
3703538 c.530T>C p.I177T Chr.9: 32418383-32418383 on Build GRCh38
4919207 c.499A>G p.M167V Chr.9: 32418352-32418352 on Build GRCh38
3906787 c.501G>A p.M167I Chr.9: 32418354-32418354 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766152770 Chr.9: 32418379-32418379 on Build GRCh38
rs879141690 Chr.9: 32418277-32418277 on Build GRCh38
rs768708549 Chr.9: 32418446-32418446 on Build GRCh38
rs373332582 Chr.9: 32418311-32418311 on Build GRCh38
rs778052946 Chr.9: 32418293-32418293 on Build GRCh38
rs767756764 Chr.9: 32418197-32418198 on Build GRCh38
rs755996483 Chr.9: 32418123-32418123 on Build GRCh38
rs2026739 Chr.9: 32418239-32418239 on Build GRCh38
rs904683578 Chr.9: 32418272-32418272 on Build GRCh38
rs779706002 Chr.9: 32418144-32418144 on Build GRCh38
rs778537345 Chr.9: 32418174-32418174 on Build GRCh38
rs749679991 Chr.9: 32418432-32418432 on Build GRCh38
rs1036093594 Chr.9: 32418457-32418457 on Build GRCh38
rs888997704 Chr.9: 32418478-32418478 on Build GRCh38
rs544613435 Chr.9: 32418362-32418362 on Build GRCh38
rs749513351 Chr.9: 32418351-32418351 on Build GRCh38
rs771535995 Chr.9: 32418190-32418190 on Build GRCh38
rs139223145 Chr.9: 32418456-32418456 on Build GRCh38
rs371287650 Chr.9: 32418418-32418418 on Build GRCh38
rs983881085 Chr.9: 32418075-32418075 on Build GRCh38
rs774234584 Chr.9: 32418356-32418356 on Build GRCh38
rs762956887 Chr.9: 32418082-32418086 on Build GRCh38
rs766204427 Chr.9: 32418245-32418245 on Build GRCh38
rs770259136 Chr.9: 32418298-32418298 on Build GRCh38
rs375356805 Chr.9: 32418129-32418129 on Build GRCh38
rs780495634 Chr.9: 32418427-32418427 on Build GRCh38
rs764307499 Chr.9: 32418111-32418111 on Build GRCh38
rs751429417 Chr.9: 32418117-32418117 on Build GRCh38
rs148577484 Chr.9: 32418132-32418132 on Build GRCh38
rs150497498 Chr.9: 32418198-32418198 on Build GRCh38
rs143260336 Chr.9: 32418388-32418388 on Build GRCh38
rs748759131 Chr.9: 32418153-32418153 on Build GRCh38
rs753470136 Chr.9: 32418248-32418248 on Build GRCh38
rs2274721 Chr.9: 32418279-32418279 on Build GRCh38
rs1028410753 Chr.9: 32418330-32418330 on Build GRCh38
rs774553670 Chr.9: 32418234-32418234 on Build GRCh38
rs201517506 Chr.9: 32418499-32418499 on Build GRCh38
rs377699256 Chr.9: 32418411-32418411 on Build GRCh38
rs978139256 Chr.9: 32418257-32418257 on Build GRCh38
rs763154719 Chr.9: 32418089-32418089 on Build GRCh38
rs371235086 Chr.9: 32418471-32418471 on Build GRCh38
rs759044295 Chr.9: 32418391-32418391 on Build GRCh38
rs776354014 Chr.9: 32418497-32418497 on Build GRCh38
rs61753543 Chr.9: 32418475-32418475 on Build GRCh38
rs573614040 Chr.9: 32418205-32418205 on Build GRCh38
rs747457843 Chr.9: 32418179-32418179 on Build GRCh38
rs898816838 Chr.9: 32418065-32418065 on Build GRCh38
rs760657982 Chr.9: 32418371-32418371 on Build GRCh38
rs764066830 Chr.9: 32418217-32418217 on Build GRCh38
rs772171891 Chr.9: 32418474-32418474 on Build GRCh38
rs1028885005 Chr.9: 32418087-32418087 on Build GRCh38
rs746415598 Chr.9: 32418295-32418295 on Build GRCh38
rs754687260 Chr.9: 32418249-32418249 on Build GRCh38
rs182129295 Chr.9: 32418236-32418236 on Build GRCh38
rs771866685 Chr.9: 32418365-32418365 on Build GRCh38
rs750952732 Chr.9: 32418420-32418420 on Build GRCh38
rs41305321 Chr.9: 32418355-32418355 on Build GRCh38
rs530451988 Chr.9: 32418421-32418421 on Build GRCh38
rs760606540 Chr.9: 32418485-32418485 on Build GRCh38
rs770860316 Chr.9: 32418491-32418491 on Build GRCh38
rs142015075 Chr.9: 32418180-32418180 on Build GRCh38
rs750556772 Chr.9: 32418474-32418474 on Build GRCh38
rs940448475 Chr.9: 32418409-32418409 on Build GRCh38
rs537053975 Chr.9: 32418085-32418085 on Build GRCh38
rs763315403 Chr.9: 32418213-32418213 on Build GRCh38
rs372495915 Chr.9: 32418082-32418082 on Build GRCh38
rs545140455 Chr.9: 32418292-32418292 on Build GRCh38
rs775822687 Chr.9: 32418208-32418208 on Build GRCh38
rs185730378 Chr.9: 32418294-32418294 on Build GRCh38
rs779854970 Chr.9: 32418238-32418238 on Build GRCh38
rs771023115 Chr.9: 32418328-32418328 on Build GRCh38
rs199823143 Chr.9: 32418181-32418181 on Build GRCh38
rs561386458 Chr.9: 32418417-32418417 on Build GRCh38
rs773106532 Chr.9: 32418366-32418366 on Build GRCh38
rs752101251 Chr.9: 32418397-32418397 on Build GRCh38
rs556949534 Chr.9: 32418119-32418119 on Build GRCh38
rs965472539 Chr.9: 32418250-32418250 on Build GRCh38
rs770099446 Chr.9: 32418206-32418206 on Build GRCh38
rs948785101 Chr.9: 32418256-32418256 on Build GRCh38
rs776053905 Chr.9: 32418350-32418350 on Build GRCh38
rs146705506 Chr.9: 32418453-32418453 on Build GRCh38
rs200265624 Chr.9: 32418314-32418314 on Build GRCh38

View Full Product Details