Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 244850215-244850715 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HNRNPU
Entrez Gene ID
3192
Gene Name
heterogeneous nuclear ribonucleoprotein U
Gene Aliases
HNRNPU-AS1, HNRPU, SAF-A, SAFA, U21.1, hnRNP U
UniGene
Hs.106212
Gene Chromosome Location
Chr.1: 244842123-244864720 on Build GRCh38
Transcripts
NM_004501.3 NM_031844.2 XM_017001115.1 XM_017001116.1 XM_017001117.1

SNP Detail

SNP ID Chromosome Location
rs572017176 Chr.1: 244850555-244850555 on Build GRCh38
rs376405322 Chr.1: 244850473-244850473 on Build GRCh38
rs375411009 Chr.1: 244850475-244850476 on Build GRCh38
rs770360524 Chr.1: 244850472-244850475 on Build GRCh38
rs920441634 Chr.1: 244850329-244850329 on Build GRCh38
rs371360474 Chr.1: 244850399-244850399 on Build GRCh38
rs760973058 Chr.1: 244850473-244850474 on Build GRCh38
rs1051888671 Chr.1: 244850365-244850365 on Build GRCh38
rs796869947 Chr.1: 244850474-244850475 on Build GRCh38
rs111230124 Chr.1: 244850475-244850475 on Build GRCh38
rs973436442 Chr.1: 244850308-244850308 on Build GRCh38
rs148844946 Chr.1: 244850247-244850249 on Build GRCh38
rs979751208 Chr.1: 244850482-244850482 on Build GRCh38
rs796822696 Chr.1: 244850474-244850475 on Build GRCh38
rs760415505 Chr.1: 244850473-244850475 on Build GRCh38
rs989067834 Chr.1: 244850289-244850289 on Build GRCh38
rs935808242 Chr.1: 244850279-244850279 on Build GRCh38
rs961990884 Chr.1: 244850506-244850506 on Build GRCh38
rs969733911 Chr.1: 244850245-244850245 on Build GRCh38
rs925900899 Chr.1: 244850270-244850270 on Build GRCh38
rs886645191 Chr.1: 244850464-244850464 on Build GRCh38
rs886587370 Chr.1: 244850633-244850633 on Build GRCh38
rs902956469 Chr.1: 244850431-244850431 on Build GRCh38
rs1495946 Chr.1: 244850505-244850505 on Build GRCh38
rs1054175039 Chr.1: 244850461-244850461 on Build GRCh38
rs1045427784 Chr.1: 244850682-244850682 on Build GRCh38
rs1026907516 Chr.1: 244850458-244850458 on Build GRCh38
rs1048015654 Chr.1: 244850611-244850611 on Build GRCh38
rs933865112 Chr.1: 244850515-244850515 on Build GRCh38
rs949779228 Chr.1: 244850382-244850382 on Build GRCh38
rs987949582 Chr.1: 244850529-244850529 on Build GRCh38
rs899434438 Chr.1: 244850425-244850425 on Build GRCh38
rs1027655864 Chr.1: 244850545-244850545 on Build GRCh38
rs991722578 Chr.1: 244850590-244850590 on Build GRCh38
rs996747927 Chr.1: 244850478-244850478 on Build GRCh38
rs890690517 Chr.1: 244850483-244850483 on Build GRCh38
rs533555019 Chr.1: 244850375-244850375 on Build GRCh38
rs899381913 Chr.1: 244850604-244850604 on Build GRCh38
rs755202859 Chr.1: 244850285-244850285 on Build GRCh38
rs890445354 Chr.1: 244850372-244850372 on Build GRCh38
rs200921383 Chr.1: 244850484-244850484 on Build GRCh38
rs570452837 Chr.1: 244850239-244850239 on Build GRCh38
rs57229249 Chr.1: 244850475-244850475 on Build GRCh38
rs138995227 Chr.1: 244850433-244850433 on Build GRCh38
rs1024080506 Chr.1: 244850479-244850479 on Build GRCh38
rs969682819 Chr.1: 244850480-244850480 on Build GRCh38
rs201646006 Chr.1: 244850473-244850473 on Build GRCh38
rs371496907 Chr.1: 244850475-244850477 on Build GRCh38
rs756758297 Chr.1: 244850337-244850337 on Build GRCh38
rs1041761231 Chr.1: 244850393-244850393 on Build GRCh38
rs957286091 Chr.1: 244850485-244850485 on Build GRCh38
rs945458931 Chr.1: 244850359-244850359 on Build GRCh38
rs35952469 Chr.1: 244850483-244850484 on Build GRCh38
rs796209846 Chr.1: 244850484-244850485 on Build GRCh38
rs913048175 Chr.1: 244850513-244850513 on Build GRCh38
rs930885074 Chr.1: 244850610-244850610 on Build GRCh38
rs540917842 Chr.1: 244850561-244850561 on Build GRCh38
rs758465529 Chr.1: 244850600-244850601 on Build GRCh38
rs780838733 Chr.1: 244850474-244850476 on Build GRCh38
rs1015040502 Chr.1: 244850268-244850268 on Build GRCh38
rs1039056693 Chr.1: 244850602-244850602 on Build GRCh38
rs913946747 Chr.1: 244850307-244850307 on Build GRCh38
rs35791152 Chr.1: 244850290-244850290 on Build GRCh38
rs749743527 Chr.1: 244850560-244850560 on Build GRCh38
rs917150694 Chr.1: 244850376-244850376 on Build GRCh38
rs1014417180 Chr.1: 244850477-244850477 on Build GRCh38
rs146061226 Chr.1: 244850266-244850266 on Build GRCh38
rs1029622726 Chr.1: 244850258-244850258 on Build GRCh38
rs560158144 Chr.1: 244850641-244850641 on Build GRCh38
rs112842127 Chr.1: 244850472-244850473 on Build GRCh38

View Full Product Details