Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 86891866-86892384 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BMPR1A
Entrez Gene ID
657
Gene Name
bone morphogenetic protein receptor type 1A
Gene Aliases
10q23del, ACVRLK3, ALK3, CD292, SKR5
UniGene
Hs.524477
Gene Chromosome Location
Chr.10: 86755786-86927969 on Build GRCh38
Transcripts
NM_004329.2 XM_011540103.2 XM_011540104.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1967769 c.329G>A p.C110Y Chr.10: 86892225-86892225 on Build GRCh38
6130295 c.314G>T p.G105V Chr.10: 86892210-86892210 on Build GRCh38
1559325 c.253A>C p.I85L Chr.10: 86892149-86892149 on Build GRCh38
7408959 c.295G>T p.G99W Chr.10: 86892191-86892191 on Build GRCh38
5636370 c.233C>G p.T78S Chr.10: 86892129-86892129 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs533037068 Chr.10: 86891973-86891973 on Build GRCh38
rs756856268 Chr.10: 86892083-86892083 on Build GRCh38
rs928754647 Chr.10: 86892276-86892276 on Build GRCh38
rs876658891 Chr.10: 86892205-86892205 on Build GRCh38
rs960339590 Chr.10: 86892057-86892057 on Build GRCh38
rs902827224 Chr.10: 86892320-86892320 on Build GRCh38
rs759444659 Chr.10: 86892258-86892261 on Build GRCh38
rs183257916 Chr.10: 86891977-86891977 on Build GRCh38
rs730881431 Chr.10: 86892158-86892158 on Build GRCh38
rs997691584 Chr.10: 86892348-86892348 on Build GRCh38
rs894407881 Chr.10: 86892027-86892027 on Build GRCh38
rs746986965 Chr.10: 86892266-86892266 on Build GRCh38
rs756944246 Chr.10: 86892228-86892228 on Build GRCh38
rs748971979 Chr.10: 86892080-86892080 on Build GRCh38
rs115583244 Chr.10: 86892070-86892070 on Build GRCh38
rs938513328 Chr.10: 86892284-86892284 on Build GRCh38
rs786204187 Chr.10: 86892139-86892143 on Build GRCh38
rs75039078 Chr.10: 86892274-86892274 on Build GRCh38
rs757943183 Chr.10: 86892253-86892253 on Build GRCh38
rs759168207 Chr.10: 86892315-86892315 on Build GRCh38
rs772231659 Chr.10: 86892094-86892098 on Build GRCh38
rs759647230 Chr.10: 86892142-86892142 on Build GRCh38
rs947259814 Chr.10: 86892290-86892290 on Build GRCh38
rs547039886 Chr.10: 86891995-86891995 on Build GRCh38
rs779367077 Chr.10: 86892262-86892262 on Build GRCh38
rs756541429 Chr.10: 86892189-86892189 on Build GRCh38
rs78693667 Chr.10: 86891993-86891993 on Build GRCh38
rs746866866 Chr.10: 86892104-86892104 on Build GRCh38
rs928430084 Chr.10: 86891994-86891994 on Build GRCh38
rs938532958 Chr.10: 86892001-86892001 on Build GRCh38
rs560251165 Chr.10: 86891982-86891982 on Build GRCh38
rs368240372 Chr.10: 86891957-86891957 on Build GRCh38
rs192599431 Chr.10: 86892257-86892257 on Build GRCh38
rs778536321 Chr.10: 86892244-86892244 on Build GRCh38
rs188083803 Chr.10: 86892047-86892047 on Build GRCh38
rs778034897 Chr.10: 86892093-86892094 on Build GRCh38
rs953391509 Chr.10: 86891906-86891906 on Build GRCh38
rs980633434 Chr.10: 86892010-86892010 on Build GRCh38
rs772271554 Chr.10: 86891985-86891985 on Build GRCh38
rs768578338 Chr.10: 86892110-86892110 on Build GRCh38
rs569319386 Chr.10: 86892084-86892084 on Build GRCh38
rs764466269 Chr.10: 86892190-86892190 on Build GRCh38
rs900946136 Chr.10: 86892337-86892337 on Build GRCh38
rs745431046 Chr.10: 86892248-86892248 on Build GRCh38
rs869312783 Chr.10: 86892185-86892186 on Build GRCh38
rs754180012 Chr.10: 86892196-86892196 on Build GRCh38
rs766739759 Chr.10: 86891953-86891953 on Build GRCh38
rs768690540 Chr.10: 86892268-86892268 on Build GRCh38
rs1029227040 Chr.10: 86892098-86892098 on Build GRCh38
rs140782702 Chr.10: 86892349-86892349 on Build GRCh38
rs927382260 Chr.10: 86892019-86892019 on Build GRCh38
rs768406007 Chr.10: 86892324-86892324 on Build GRCh38
rs759961048 Chr.10: 86892288-86892289 on Build GRCh38
rs534150993 Chr.10: 86892323-86892323 on Build GRCh38
rs1043150548 Chr.10: 86892069-86892069 on Build GRCh38
rs746810642 Chr.10: 86892088-86892088 on Build GRCh38
rs771974755 Chr.10: 86892091-86892091 on Build GRCh38
rs184442043 Chr.10: 86892301-86892301 on Build GRCh38
rs753250496 Chr.10: 86892163-86892163 on Build GRCh38
rs876659498 Chr.10: 86892153-86892153 on Build GRCh38
rs1008321903 Chr.10: 86892286-86892286 on Build GRCh38
rs876659086 Chr.10: 86892160-86892162 on Build GRCh38
rs774357141 Chr.10: 86892125-86892125 on Build GRCh38
rs372358552 Chr.10: 86892102-86892102 on Build GRCh38
rs745967033 Chr.10: 86891978-86891978 on Build GRCh38
rs953015884 Chr.10: 86892355-86892355 on Build GRCh38
rs878854667 Chr.10: 86892167-86892167 on Build GRCh38
rs1042211540 Chr.10: 86892121-86892121 on Build GRCh38
rs763313220 Chr.10: 86892118-86892118 on Build GRCh38
rs889491867 Chr.10: 86891941-86891941 on Build GRCh38
rs7912862 Chr.10: 86892005-86892005 on Build GRCh38
rs953797046 Chr.10: 86892137-86892137 on Build GRCh38
rs747206781 Chr.10: 86892110-86892111 on Build GRCh38
rs776276672 Chr.10: 86892258-86892259 on Build GRCh38
rs374739820 Chr.10: 86892149-86892149 on Build GRCh38

View Full Product Details