Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 223967186-223967751 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MRPL44
Entrez Gene ID
65080
Gene Name
mitochondrial ribosomal protein L44
Gene Aliases
COXPD16, L44MT, MRP-L44
UniGene
Hs.203559
Gene Chromosome Location
Chr.2: 223943224-223967714 on Build GRCh38
Transcripts
NM_022915.3 XM_011511668.2 XM_017004730.1 XM_017004731.1

SNP Detail

SNP ID Chromosome Location
rs1015155092 Chr.2: 223967523-223967523 on Build GRCh38
rs181615975 Chr.2: 223967613-223967613 on Build GRCh38
rs988689371 Chr.2: 223967434-223967434 on Build GRCh38
rs548236986 Chr.2: 223967398-223967398 on Build GRCh38
rs879054805 Chr.2: 223967254-223967254 on Build GRCh38
rs1039080276 Chr.2: 223967400-223967400 on Build GRCh38
rs151128420 Chr.2: 223967318-223967318 on Build GRCh38
rs556505754 Chr.2: 223967271-223967271 on Build GRCh38
rs764923761 Chr.2: 223967235-223967235 on Build GRCh38
rs1027934633 Chr.2: 223967235-223967237 on Build GRCh38
rs374731067 Chr.2: 223967302-223967302 on Build GRCh38
rs890735443 Chr.2: 223967504-223967504 on Build GRCh38
rs979730206 Chr.2: 223967489-223967489 on Build GRCh38
rs747998905 Chr.2: 223967691-223967691 on Build GRCh38
rs143020958 Chr.2: 223967296-223967296 on Build GRCh38
rs190593678 Chr.2: 223967381-223967381 on Build GRCh38
rs750911754 Chr.2: 223967391-223967391 on Build GRCh38
rs76326303 Chr.2: 223967697-223967697 on Build GRCh38
rs1039029966 Chr.2: 223967640-223967640 on Build GRCh38
rs10980 Chr.2: 223967629-223967629 on Build GRCh38
rs572847979 Chr.2: 223967229-223967229 on Build GRCh38
rs1142417 Chr.2: 223967444-223967444 on Build GRCh38
rs535470442 Chr.2: 223967251-223967251 on Build GRCh38
rs1005560857 Chr.2: 223967513-223967513 on Build GRCh38
rs1034471027 Chr.2: 223967490-223967490 on Build GRCh38
rs571033902 Chr.2: 223967479-223967479 on Build GRCh38
rs539377026 Chr.2: 223967437-223967437 on Build GRCh38
rs575262289 Chr.2: 223967295-223967295 on Build GRCh38
rs762405120 Chr.2: 223967244-223967244 on Build GRCh38
rs1045575372 Chr.2: 223967701-223967701 on Build GRCh38
rs938580855 Chr.2: 223967423-223967423 on Build GRCh38
rs1027740473 Chr.2: 223967442-223967442 on Build GRCh38
rs978581492 Chr.2: 223967713-223967713 on Build GRCh38
rs1001367901 Chr.2: 223967714-223967714 on Build GRCh38
rs963550395 Chr.2: 223967343-223967343 on Build GRCh38
rs1030594450 Chr.2: 223967313-223967313 on Build GRCh38
rs555408563 Chr.2: 223967294-223967294 on Build GRCh38
rs533621698 Chr.2: 223967486-223967486 on Build GRCh38
rs979764354 Chr.2: 223967333-223967333 on Build GRCh38
rs114000585 Chr.2: 223967552-223967552 on Build GRCh38
rs558063547 Chr.2: 223967692-223967692 on Build GRCh38
rs912992443 Chr.2: 223967528-223967528 on Build GRCh38
rs973479139 Chr.2: 223967359-223967359 on Build GRCh38
rs143136727 Chr.2: 223967663-223967663 on Build GRCh38
rs900605067 Chr.2: 223967234-223967235 on Build GRCh38
rs1023709498 Chr.2: 223967584-223967584 on Build GRCh38
rs886550691 Chr.2: 223967694-223967694 on Build GRCh38
rs1138616 Chr.2: 223967419-223967419 on Build GRCh38
rs978278972 Chr.2: 223967565-223967565 on Build GRCh38
rs899333917 Chr.2: 223967526-223967526 on Build GRCh38
rs1042633706 Chr.2: 223967382-223967382 on Build GRCh38
rs150253706 Chr.2: 223967463-223967463 on Build GRCh38
rs765834403 Chr.2: 223967301-223967301 on Build GRCh38
rs540034755 Chr.2: 223967335-223967335 on Build GRCh38
rs915357102 Chr.2: 223967358-223967358 on Build GRCh38
rs537826978 Chr.2: 223967664-223967664 on Build GRCh38
rs1042798927 Chr.2: 223967288-223967288 on Build GRCh38
rs567011103 Chr.2: 223967568-223967568 on Build GRCh38
rs559848771 Chr.2: 223967342-223967342 on Build GRCh38
rs564339832 Chr.2: 223967310-223967310 on Build GRCh38
rs769288506 Chr.2: 223967661-223967662 on Build GRCh38
rs754670952 Chr.2: 223967493-223967493 on Build GRCh38
rs982102290 Chr.2: 223967619-223967619 on Build GRCh38
rs756737769 Chr.2: 223967240-223967240 on Build GRCh38

View Full Product Details