Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 80845649-80846161 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LIN7A
Entrez Gene ID
8825
Gene Name
lin-7 homolog A, crumbs cell polarity complex component
Gene Aliases
LIN-7A, LIN7, MALS-1, TIP-33, VELI1
UniGene
Hs.144333
Gene Chromosome Location
Chr.12: 80792520-80937915 on Build GRCh38
Transcripts
NM_001324423.1 NM_004664.3 NR_136887.1 NR_136888.1 XM_011538928.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6701377 c.402C>A p.I134I Chr.12: 80845811-80845811 on Build GRCh38
468967 c.354T>A p.L118L Chr.12: 80845859-80845859 on Build GRCh38
4564050 c.388_389TC>AT p.S130I Chr.12: 80845824-80845825 on Build GRCh38
4600259 c.425T>A p.V142E Chr.12: 80845788-80845788 on Build GRCh38
6701375 c.338A>C p.K113T Chr.12: 80845875-80845875 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745516494 Chr.12: 80845695-80845695 on Build GRCh38
rs757814174 Chr.12: 80845813-80845813 on Build GRCh38
rs748669258 Chr.12: 80845873-80845873 on Build GRCh38
rs919422830 Chr.12: 80846123-80846123 on Build GRCh38
rs770345364 Chr.12: 80845874-80845874 on Build GRCh38
rs963925423 Chr.12: 80846077-80846077 on Build GRCh38
rs759761824 Chr.12: 80845888-80845888 on Build GRCh38
rs984162906 Chr.12: 80846024-80846024 on Build GRCh38
rs1050665675 Chr.12: 80846130-80846131 on Build GRCh38
rs753828551 Chr.12: 80845795-80845795 on Build GRCh38
rs763169948 Chr.12: 80845675-80845675 on Build GRCh38
rs765767089 Chr.12: 80845943-80845943 on Build GRCh38
rs768582140 Chr.12: 80845894-80845894 on Build GRCh38
rs754673628 Chr.12: 80845943-80845946 on Build GRCh38
rs758125235 Chr.12: 80845707-80845707 on Build GRCh38
rs377032979 Chr.12: 80845881-80845881 on Build GRCh38
rs750378476 Chr.12: 80845942-80845943 on Build GRCh38
rs756442179 Chr.12: 80845968-80845968 on Build GRCh38
rs752598180 Chr.12: 80845678-80845678 on Build GRCh38
rs866830191 Chr.12: 80845798-80845798 on Build GRCh38
rs573393665 Chr.12: 80845677-80845677 on Build GRCh38
rs761781509 Chr.12: 80845796-80845796 on Build GRCh38
rs369054902 Chr.12: 80846019-80846019 on Build GRCh38
rs755860046 Chr.12: 80845680-80845680 on Build GRCh38
rs188937687 Chr.12: 80846028-80846028 on Build GRCh38
rs766269625 Chr.12: 80845943-80845945 on Build GRCh38
rs145813467 Chr.12: 80845919-80845919 on Build GRCh38
rs774408530 Chr.12: 80845784-80845784 on Build GRCh38
rs753130057 Chr.12: 80845967-80845967 on Build GRCh38
rs561591477 Chr.12: 80845941-80845941 on Build GRCh38
rs758376918 Chr.12: 80845943-80845944 on Build GRCh38
rs770897474 Chr.12: 80845877-80845877 on Build GRCh38
rs909567397 Chr.12: 80846071-80846071 on Build GRCh38
rs765304610 Chr.12: 80845924-80845924 on Build GRCh38
rs756855265 Chr.12: 80845686-80845686 on Build GRCh38
rs183376972 Chr.12: 80845775-80845775 on Build GRCh38
rs767232112 Chr.12: 80845677-80845677 on Build GRCh38
rs774898360 Chr.12: 80845676-80845677 on Build GRCh38
rs369120651 Chr.12: 80845745-80845745 on Build GRCh38
rs751228960 Chr.12: 80845951-80845951 on Build GRCh38
rs79919291 Chr.12: 80845955-80845957 on Build GRCh38
rs763937415 Chr.12: 80845792-80845792 on Build GRCh38
rs776623278 Chr.12: 80845908-80845908 on Build GRCh38
rs541511733 Chr.12: 80845677-80845678 on Build GRCh38
rs539927453 Chr.12: 80845862-80845862 on Build GRCh38
rs150476261 Chr.12: 80845736-80845736 on Build GRCh38
rs779804958 Chr.12: 80845817-80845817 on Build GRCh38
rs763863477 Chr.12: 80845702-80845702 on Build GRCh38
rs773221131 Chr.12: 80845772-80845772 on Build GRCh38
rs1022013918 Chr.12: 80845690-80845690 on Build GRCh38
rs780810275 Chr.12: 80845960-80845960 on Build GRCh38
rs778140268 Chr.12: 80845875-80845875 on Build GRCh38
rs751123231 Chr.12: 80845844-80845844 on Build GRCh38
rs776089847 Chr.12: 80845726-80845727 on Build GRCh38
rs909276256 Chr.12: 80846001-80846001 on Build GRCh38
rs933561917 Chr.12: 80846051-80846053 on Build GRCh38
rs753808468 Chr.12: 80845685-80845685 on Build GRCh38
rs749428556 Chr.12: 80845876-80845876 on Build GRCh38
rs200552580 Chr.12: 80845709-80845709 on Build GRCh38
rs761390051 Chr.12: 80845908-80845911 on Build GRCh38
rs764170464 Chr.12: 80845684-80845684 on Build GRCh38
rs755367037 Chr.12: 80845861-80845861 on Build GRCh38
rs949556377 Chr.12: 80846124-80846124 on Build GRCh38
rs750015465 Chr.12: 80845812-80845812 on Build GRCh38
rs972342454 Chr.12: 80846099-80846099 on Build GRCh38
rs948046789 Chr.12: 80845890-80845890 on Build GRCh38
rs754568568 Chr.12: 80845957-80845957 on Build GRCh38
rs568837790 Chr.12: 80846083-80846083 on Build GRCh38
rs187508405 Chr.12: 80845694-80845694 on Build GRCh38
rs774126564 Chr.12: 80845943-80845943 on Build GRCh38
rs778396294 Chr.12: 80845982-80845982 on Build GRCh38
rs750304433 Chr.12: 80845940-80845940 on Build GRCh38
rs528221716 Chr.12: 80845789-80845789 on Build GRCh38
rs765251435 Chr.12: 80845805-80845805 on Build GRCh38
rs762931661 Chr.12: 80845774-80845774 on Build GRCh38
rs190432616 Chr.12: 80845735-80845735 on Build GRCh38
rs529042334 Chr.12: 80846020-80846020 on Build GRCh38

View Full Product Details