Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 15721237-15721743 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLEKHM2
Entrez Gene ID
23207
Gene Name
pleckstrin homology and RUN domain containing M2
Gene Aliases
SKIP
UniGene
Hs.646775
Gene Chromosome Location
Chr.1: 15681506-15734769 on Build GRCh38
Transcripts
NM_015164.2 XM_005245790.3 XM_005245791.4 XM_017000757.1 XM_017000758.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6843286 c.1007G>A p.S336N Chr.1: 15721374-15721374 on Build GRCh38
6843285 c.698G>A p.S233N Chr.1: 15721374-15721374 on Build GRCh38
6223994 c.1006A>G p.S336G Chr.1: 15721373-15721373 on Build GRCh38
4442611 c.1019A>G p.E340G Chr.1: 15721386-15721386 on Build GRCh38
6405036 c.685C>T p.P229S Chr.1: 15721361-15721361 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758769572 Chr.1: 15721382-15721382 on Build GRCh38
rs778300334 Chr.1: 15721402-15721402 on Build GRCh38
rs765614073 Chr.1: 15721363-15721363 on Build GRCh38
rs189607714 Chr.1: 15721693-15721693 on Build GRCh38
rs751822963 Chr.1: 15721403-15721403 on Build GRCh38
rs887589197 Chr.1: 15721595-15721595 on Build GRCh38
rs1033356681 Chr.1: 15721462-15721462 on Build GRCh38
rs556149633 Chr.1: 15721495-15721495 on Build GRCh38
rs776621339 Chr.1: 15721555-15721555 on Build GRCh38
rs76395188 Chr.1: 15721644-15721644 on Build GRCh38
rs763826102 Chr.1: 15721712-15721712 on Build GRCh38
rs987618660 Chr.1: 15721506-15721506 on Build GRCh38
rs529429914 Chr.1: 15721692-15721692 on Build GRCh38
rs117390718 Chr.1: 15721696-15721696 on Build GRCh38
rs375501738 Chr.1: 15721334-15721334 on Build GRCh38
rs955813301 Chr.1: 15721690-15721690 on Build GRCh38
rs114598853 Chr.1: 15721475-15721475 on Build GRCh38
rs781303887 Chr.1: 15721430-15721430 on Build GRCh38
rs975110795 Chr.1: 15721524-15721524 on Build GRCh38
rs926016340 Chr.1: 15721532-15721532 on Build GRCh38
rs993566379 Chr.1: 15721554-15721554 on Build GRCh38
rs116699906 Chr.1: 15721709-15721709 on Build GRCh38
rs1022848282 Chr.1: 15721615-15721615 on Build GRCh38
rs542815793 Chr.1: 15721476-15721476 on Build GRCh38
rs1023073561 Chr.1: 15721293-15721293 on Build GRCh38
rs553546149 Chr.1: 15721427-15721427 on Build GRCh38
rs1046274185 Chr.1: 15721643-15721643 on Build GRCh38
rs760055434 Chr.1: 15721346-15721346 on Build GRCh38
rs12739222 Chr.1: 15721514-15721514 on Build GRCh38
rs12091326 Chr.1: 15721510-15721510 on Build GRCh38
rs761466899 Chr.1: 15721325-15721325 on Build GRCh38
rs578068746 Chr.1: 15721612-15721612 on Build GRCh38
rs562546054 Chr.1: 15721287-15721287 on Build GRCh38
rs866799799 Chr.1: 15721374-15721374 on Build GRCh38
rs963418171 Chr.1: 15721472-15721472 on Build GRCh38
rs1040771912 Chr.1: 15721523-15721523 on Build GRCh38
rs142928865 Chr.1: 15721292-15721292 on Build GRCh38
rs928365875 Chr.1: 15721359-15721359 on Build GRCh38
rs775095007 Chr.1: 15721621-15721621 on Build GRCh38
rs1009473437 Chr.1: 15721318-15721318 on Build GRCh38
rs560643171 Chr.1: 15721677-15721677 on Build GRCh38
rs773922799 Chr.1: 15721324-15721324 on Build GRCh38
rs372350931 Chr.1: 15721331-15721331 on Build GRCh38
rs369758650 Chr.1: 15721459-15721459 on Build GRCh38
rs184985321 Chr.1: 15721411-15721411 on Build GRCh38
rs943343359 Chr.1: 15721519-15721519 on Build GRCh38
rs753290058 Chr.1: 15721371-15721371 on Build GRCh38
rs945167036 Chr.1: 15721636-15721636 on Build GRCh38
rs537418406 Chr.1: 15721259-15721259 on Build GRCh38
rs12092485 Chr.1: 15721543-15721543 on Build GRCh38
rs1054024330 Chr.1: 15721268-15721268 on Build GRCh38
rs12065705 Chr.1: 15721619-15721619 on Build GRCh38
rs981608101 Chr.1: 15721343-15721343 on Build GRCh38
rs1033757881 Chr.1: 15721654-15721654 on Build GRCh38
rs570990579 Chr.1: 15721319-15721319 on Build GRCh38
rs987566914 Chr.1: 15721711-15721711 on Build GRCh38

View Full Product Details