Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 41848281-41848775 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPTBN5
Entrez Gene ID
51332
Gene Name
spectrin beta, non-erythrocytic 5
Gene Aliases
BSPECV, HUBSPECV, HUSPECV
UniGene
Hs.709819
Gene Chromosome Location
Chr.15: 41848146-41894077 on Build GRCh38
Transcripts
NM_016642.3 XM_017022299.1 XM_017022300.1 XM_017022301.1 XM_017022302.1

SNP Detail

SNP ID Chromosome Location
rs749453019 Chr.15: 41848629-41848629 on Build GRCh38
rs375340369 Chr.15: 41848671-41848671 on Build GRCh38
rs116152856 Chr.15: 41848426-41848426 on Build GRCh38
rs146466158 Chr.15: 41848576-41848576 on Build GRCh38
rs202059071 Chr.15: 41848668-41848668 on Build GRCh38
rs765283742 Chr.15: 41848654-41848654 on Build GRCh38
rs146672510 Chr.15: 41848613-41848613 on Build GRCh38
rs748539119 Chr.15: 41848615-41848615 on Build GRCh38
rs149904045 Chr.15: 41848575-41848575 on Build GRCh38
rs922267446 Chr.15: 41848409-41848409 on Build GRCh38
rs540870962 Chr.15: 41848744-41848744 on Build GRCh38
rs971141105 Chr.15: 41848343-41848343 on Build GRCh38
rs1046936566 Chr.15: 41848554-41848554 on Build GRCh38
rs372821744 Chr.15: 41848374-41848374 on Build GRCh38
rs761641786 Chr.15: 41848646-41848646 on Build GRCh38
rs967298323 Chr.15: 41848596-41848596 on Build GRCh38
rs139805178 Chr.15: 41848584-41848584 on Build GRCh38
rs746124727 Chr.15: 41848641-41848641 on Build GRCh38
rs1022974707 Chr.15: 41848514-41848514 on Build GRCh38
rs566099939 Chr.15: 41848425-41848425 on Build GRCh38
rs777703654 Chr.15: 41848618-41848618 on Build GRCh38
rs1011089838 Chr.15: 41848725-41848725 on Build GRCh38
rs34619464 Chr.15: 41848621-41848622 on Build GRCh38
rs966452160 Chr.15: 41848388-41848388 on Build GRCh38
rs766056588 Chr.15: 41848601-41848601 on Build GRCh38
rs972650212 Chr.15: 41848663-41848663 on Build GRCh38
rs958311692 Chr.15: 41848372-41848372 on Build GRCh38
rs570270012 Chr.15: 41848320-41848322 on Build GRCh38
rs920433356 Chr.15: 41848740-41848740 on Build GRCh38
rs751895220 Chr.15: 41848602-41848602 on Build GRCh38
rs981923447 Chr.15: 41848524-41848524 on Build GRCh38
rs1055827 Chr.15: 41848327-41848327 on Build GRCh38
rs753785974 Chr.15: 41848585-41848585 on Build GRCh38
rs991515926 Chr.15: 41848344-41848344 on Build GRCh38
rs551901108 Chr.15: 41848459-41848459 on Build GRCh38
rs529534042 Chr.15: 41848385-41848385 on Build GRCh38
rs888843506 Chr.15: 41848440-41848440 on Build GRCh38
rs757979710 Chr.15: 41848595-41848595 on Build GRCh38
rs1055013966 Chr.15: 41848712-41848712 on Build GRCh38
rs369402065 Chr.15: 41848689-41848689 on Build GRCh38
rs1021173305 Chr.15: 41848729-41848729 on Build GRCh38
rs1033301655 Chr.15: 41848333-41848333 on Build GRCh38
rs957620569 Chr.15: 41848502-41848502 on Build GRCh38
rs1002437258 Chr.15: 41848346-41848346 on Build GRCh38
rs962166438 Chr.15: 41848552-41848552 on Build GRCh38
rs779948726 Chr.15: 41848606-41848606 on Build GRCh38
rs879848917 Chr.15: 41848380-41848380 on Build GRCh38
rs993559533 Chr.15: 41848546-41848546 on Build GRCh38
rs972287801 Chr.15: 41848728-41848728 on Build GRCh38
rs1036860261 Chr.15: 41848517-41848517 on Build GRCh38
rs906505992 Chr.15: 41848557-41848557 on Build GRCh38
rs748387353 Chr.15: 41848591-41848592 on Build GRCh38
rs763591918 Chr.15: 41848549-41848550 on Build GRCh38
rs776580312 Chr.15: 41848643-41848643 on Build GRCh38
rs985120954 Chr.15: 41848632-41848632 on Build GRCh38
rs781367632 Chr.15: 41848582-41848583 on Build GRCh38
rs372651861 Chr.15: 41848696-41848696 on Build GRCh38
rs971383384 Chr.15: 41848682-41848683 on Build GRCh38
rs909780462 Chr.15: 41848476-41848476 on Build GRCh38
rs757112348 Chr.15: 41848479-41848479 on Build GRCh38
rs915969322 Chr.15: 41848359-41848359 on Build GRCh38
rs951931488 Chr.15: 41848743-41848743 on Build GRCh38
rs910226901 Chr.15: 41848589-41848592 on Build GRCh38
rs139360469 Chr.15: 41848401-41848401 on Build GRCh38
rs535233880 Chr.15: 41848446-41848446 on Build GRCh38
rs770100039 Chr.15: 41848606-41848606 on Build GRCh38
rs963653079 Chr.15: 41848378-41848379 on Build GRCh38
rs938142841 Chr.15: 41848707-41848707 on Build GRCh38
rs750383749 Chr.15: 41848591-41848606 on Build GRCh38
rs774763439 Chr.15: 41848640-41848640 on Build GRCh38
rs755092999 Chr.15: 41848570-41848571 on Build GRCh38
rs761813223 Chr.15: 41848586-41848586 on Build GRCh38
rs371907105 Chr.15: 41848616-41848616 on Build GRCh38
rs571810260 Chr.15: 41848535-41848535 on Build GRCh38
rs893810648 Chr.15: 41848713-41848713 on Build GRCh38
rs563987667 Chr.15: 41848373-41848373 on Build GRCh38
rs950130473 Chr.15: 41848363-41848363 on Build GRCh38
rs1024943932 Chr.15: 41848326-41848326 on Build GRCh38
rs772562636 Chr.15: 41848658-41848658 on Build GRCh38
rs779334777 Chr.15: 41848518-41848518 on Build GRCh38
rs199626756 Chr.15: 41848589-41848589 on Build GRCh38
rs548669718 Chr.15: 41848642-41848642 on Build GRCh38
rs1033666040 Chr.15: 41848481-41848481 on Build GRCh38
rs1033907138 Chr.15: 41848365-41848365 on Build GRCh38
rs1012899004 Chr.15: 41848513-41848513 on Build GRCh38
rs143197259 Chr.15: 41848590-41848590 on Build GRCh38
rs746022249 Chr.15: 41848520-41848520 on Build GRCh38
rs532471796 Chr.15: 41848473-41848474 on Build GRCh38
rs760426645 Chr.15: 41848659-41848659 on Build GRCh38
rs771248218 Chr.15: 41848631-41848631 on Build GRCh38

View Full Product Details