Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 29588548-29589060 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OR2H2
Entrez Gene ID
7932
Gene Name
olfactory receptor family 2 subfamily H member 2
Gene Aliases
FAT11, OLFR2, OLFR42B, OR2H3, dJ271M21.2, hs6M1-12
UniGene
Hs.529493
Gene Chromosome Location
Chr.6: 29586524-29591022 on Build GRCh38
Transcripts
NM_007160.3 XM_011514912.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5939572 c.662G>A p.W221* Chr.6: 29588606-29588606 on Build GRCh38
3928290 c.818G>C p.G273A Chr.6: 29588762-29588762 on Build GRCh38
5931870 c.650G>A p.G217E Chr.6: 29588594-29588594 on Build GRCh38
3948946 c.818G>A p.G273D Chr.6: 29588762-29588762 on Build GRCh38
255534 c.933A>G p.Q311Q Chr.6: 29588877-29588877 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768170738 Chr.6: 29588609-29588609 on Build GRCh38
rs919047687 Chr.6: 29588699-29588699 on Build GRCh38
rs573382937 Chr.6: 29588957-29588957 on Build GRCh38
rs768807040 Chr.6: 29588928-29588928 on Build GRCh38
rs752171782 Chr.6: 29588880-29588880 on Build GRCh38
rs3025647 Chr.6: 29588942-29588942 on Build GRCh38
rs188661866 Chr.6: 29588908-29588908 on Build GRCh38
rs73404743 Chr.6: 29588915-29588915 on Build GRCh38
rs146052995 Chr.6: 29588621-29588621 on Build GRCh38
rs763172729 Chr.6: 29588808-29588808 on Build GRCh38
rs950347294 Chr.6: 29588985-29588985 on Build GRCh38
rs369423849 Chr.6: 29588918-29588918 on Build GRCh38
rs147023039 Chr.6: 29588710-29588710 on Build GRCh38
rs771780279 Chr.6: 29588629-29588629 on Build GRCh38
rs760024528 Chr.6: 29588770-29588770 on Build GRCh38
rs362521 Chr.6: 29588982-29588982 on Build GRCh38
rs752832233 Chr.6: 29588678-29588678 on Build GRCh38
rs368234343 Chr.6: 29588896-29588896 on Build GRCh38
rs780084053 Chr.6: 29588601-29588601 on Build GRCh38
rs781618723 Chr.6: 29588911-29588911 on Build GRCh38
rs764376870 Chr.6: 29588816-29588816 on Build GRCh38
rs771593201 Chr.6: 29588740-29588741 on Build GRCh38
rs766137846 Chr.6: 29588846-29588846 on Build GRCh38
rs201155078 Chr.6: 29588749-29588749 on Build GRCh38
rs147725189 Chr.6: 29588764-29588764 on Build GRCh38
rs765268885 Chr.6: 29588576-29588576 on Build GRCh38
rs776876582 Chr.6: 29588766-29588766 on Build GRCh38
rs778418654 Chr.6: 29588732-29588732 on Build GRCh38
rs770274270 Chr.6: 29588772-29588772 on Build GRCh38
rs150027625 Chr.6: 29588761-29588761 on Build GRCh38
rs777441816 Chr.6: 29588760-29588760 on Build GRCh38
rs200676901 Chr.6: 29588715-29588715 on Build GRCh38
rs866100954 Chr.6: 29588663-29588663 on Build GRCh38
rs778873283 Chr.6: 29588859-29588859 on Build GRCh38
rs565097625 Chr.6: 29588983-29588983 on Build GRCh38
rs756103358 Chr.6: 29588593-29588593 on Build GRCh38
rs199537451 Chr.6: 29588691-29588691 on Build GRCh38
rs372781964 Chr.6: 29588575-29588575 on Build GRCh38
rs772801930 Chr.6: 29588842-29588860 on Build GRCh38
rs142456697 Chr.6: 29588652-29588652 on Build GRCh38
rs765306153 Chr.6: 29588675-29588675 on Build GRCh38
rs530760862 Chr.6: 29589020-29589020 on Build GRCh38
rs746696528 Chr.6: 29588762-29588762 on Build GRCh38
rs763259959 Chr.6: 29588685-29588685 on Build GRCh38
rs747716141 Chr.6: 29588733-29588733 on Build GRCh38
rs1009659948 Chr.6: 29588834-29588834 on Build GRCh38
rs758710486 Chr.6: 29588585-29588585 on Build GRCh38
rs984523952 Chr.6: 29588988-29588988 on Build GRCh38
rs769914406 Chr.6: 29588677-29588677 on Build GRCh38
rs908925617 Chr.6: 29589024-29589024 on Build GRCh38
rs138283057 Chr.6: 29588592-29588592 on Build GRCh38
rs557670549 Chr.6: 29588856-29588856 on Build GRCh38
rs377015654 Chr.6: 29588888-29588888 on Build GRCh38
rs775268392 Chr.6: 29588714-29588714 on Build GRCh38
rs764466653 Chr.6: 29588689-29588689 on Build GRCh38
rs761963062 Chr.6: 29588827-29588827 on Build GRCh38
rs892586474 Chr.6: 29588829-29588829 on Build GRCh38
rs141430371 Chr.6: 29588602-29588602 on Build GRCh38
rs944417775 Chr.6: 29588671-29588671 on Build GRCh38
rs746610944 Chr.6: 29588900-29588900 on Build GRCh38
rs773530562 Chr.6: 29588638-29588638 on Build GRCh38
rs73744761 Chr.6: 29588639-29588639 on Build GRCh38
rs751790552 Chr.6: 29588824-29588824 on Build GRCh38
rs948535478 Chr.6: 29588719-29588719 on Build GRCh38
rs537325173 Chr.6: 29588781-29588781 on Build GRCh38
rs766969982 Chr.6: 29588572-29588572 on Build GRCh38
rs759987091 Chr.6: 29588669-29588669 on Build GRCh38
rs753579202 Chr.6: 29588855-29588855 on Build GRCh38
rs752853758 Chr.6: 29589030-29589030 on Build GRCh38
rs191884729 Chr.6: 29589035-29589035 on Build GRCh38
rs771312590 Chr.6: 29588642-29588642 on Build GRCh38
rs749839621 Chr.6: 29588920-29588920 on Build GRCh38
rs752919216 Chr.6: 29588580-29588580 on Build GRCh38
rs1044331668 Chr.6: 29588725-29588725 on Build GRCh38

View Full Product Details