Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 128754178-128754685 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CALU
Entrez Gene ID
813
Gene Name
calumenin
Gene Aliases
-
UniGene
Hs.743262
Gene Chromosome Location
Chr.7: 128739292-128773423 on Build GRCh38
Transcripts
NM_001130674.2 NM_001199671.1 NM_001199672.1 NM_001199673.1 NM_001219.4 NR_074086.1 XM_017012659.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4971296 c.257G>A p.G86E Chr.7: 128754297-128754297 on Build GRCh38
5026825 c.354C>T p.L118L Chr.7: 128754394-128754394 on Build GRCh38
2769023 c.360A>G p.Q120Q Chr.7: 128754643-128754643 on Build GRCh38
6619154 c.307C>T p.R103C Chr.7: 128754347-128754347 on Build GRCh38
3831829 c.397A>C p.T133P Chr.7: 128754437-128754437 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764975079 Chr.7: 128754247-128754247 on Build GRCh38
rs753507913 Chr.7: 128754211-128754211 on Build GRCh38
rs150565977 Chr.7: 128754348-128754348 on Build GRCh38
rs746265584 Chr.7: 128754572-128754572 on Build GRCh38
rs139447748 Chr.7: 128754449-128754449 on Build GRCh38
rs1003977596 Chr.7: 128754565-128754565 on Build GRCh38
rs767524055 Chr.7: 128754376-128754376 on Build GRCh38
rs749313719 Chr.7: 128754319-128754319 on Build GRCh38
rs147388270 Chr.7: 128754410-128754410 on Build GRCh38
rs377059578 Chr.7: 128754573-128754573 on Build GRCh38
rs755183703 Chr.7: 128754520-128754520 on Build GRCh38
rs761108072 Chr.7: 128754357-128754357 on Build GRCh38
rs771362389 Chr.7: 128754451-128754451 on Build GRCh38
rs142351036 Chr.7: 128754509-128754509 on Build GRCh38
rs773787640 Chr.7: 128754448-128754448 on Build GRCh38
rs951148503 Chr.7: 128754265-128754265 on Build GRCh38
rs772416007 Chr.7: 128754446-128754446 on Build GRCh38
rs993921435 Chr.7: 128754580-128754580 on Build GRCh38
rs990305798 Chr.7: 128754363-128754363 on Build GRCh38
rs745704766 Chr.7: 128754298-128754298 on Build GRCh38
rs558268751 Chr.7: 128754536-128754536 on Build GRCh38
rs373909155 Chr.7: 128754316-128754316 on Build GRCh38
rs368808641 Chr.7: 128754550-128754550 on Build GRCh38
rs780108940 Chr.7: 128754404-128754404 on Build GRCh38
rs995297915 Chr.7: 128754235-128754235 on Build GRCh38
rs371458036 Chr.7: 128754605-128754605 on Build GRCh38
rs769839774 Chr.7: 128754299-128754299 on Build GRCh38
rs199523748 Chr.7: 128754296-128754296 on Build GRCh38
rs149687314 Chr.7: 128754409-128754409 on Build GRCh38
rs919493924 Chr.7: 128754661-128754661 on Build GRCh38
rs764741419 Chr.7: 128754358-128754358 on Build GRCh38
rs750416117 Chr.7: 128754500-128754500 on Build GRCh38
rs767336502 Chr.7: 128754489-128754489 on Build GRCh38
rs776598260 Chr.7: 128754396-128754396 on Build GRCh38
rs759064275 Chr.7: 128754231-128754231 on Build GRCh38
rs759295892 Chr.7: 128754442-128754442 on Build GRCh38
rs762414185 Chr.7: 128754362-128754362 on Build GRCh38
rs554351552 Chr.7: 128754209-128754209 on Build GRCh38
rs374762041 Chr.7: 128754477-128754477 on Build GRCh38
rs770954063 Chr.7: 128754295-128754295 on Build GRCh38
rs139501806 Chr.7: 128754351-128754351 on Build GRCh38
rs2307040 Chr.7: 128754552-128754552 on Build GRCh38
rs752084720 Chr.7: 128754251-128754251 on Build GRCh38
rs774040896 Chr.7: 128754574-128754574 on Build GRCh38
rs372734856 Chr.7: 128754347-128754347 on Build GRCh38
rs933551601 Chr.7: 128754541-128754541 on Build GRCh38
rs761336524 Chr.7: 128754484-128754484 on Build GRCh38
rs866287484 Chr.7: 128754417-128754417 on Build GRCh38
rs775274113 Chr.7: 128754361-128754361 on Build GRCh38
rs772306972 Chr.7: 128754577-128754577 on Build GRCh38
rs763656173 Chr.7: 128754364-128754364 on Build GRCh38
rs768923765 Chr.7: 128754332-128754332 on Build GRCh38
rs569487134 Chr.7: 128754372-128754372 on Build GRCh38
rs771359612 Chr.7: 128754557-128754557 on Build GRCh38
rs776917035 Chr.7: 128754560-128754560 on Build GRCh38
rs762538757 Chr.7: 128754472-128754472 on Build GRCh38
rs769613756 Chr.7: 128754443-128754443 on Build GRCh38
rs751087084 Chr.7: 128754371-128754371 on Build GRCh38
rs990379085 Chr.7: 128754647-128754647 on Build GRCh38
rs376487937 Chr.7: 128754466-128754466 on Build GRCh38
rs1008160047 Chr.7: 128754198-128754198 on Build GRCh38
rs753697698 Chr.7: 128754512-128754512 on Build GRCh38
rs117820033 Chr.7: 128754630-128754630 on Build GRCh38
rs773009689 Chr.7: 128754659-128754659 on Build GRCh38
rs751533093 Chr.7: 128754287-128754287 on Build GRCh38
rs768757548 Chr.7: 128754518-128754522 on Build GRCh38
rs151188778 Chr.7: 128754553-128754553 on Build GRCh38
rs777678590 Chr.7: 128754280-128754280 on Build GRCh38
rs779081471 Chr.7: 128754254-128754254 on Build GRCh38
rs755857759 Chr.7: 128754508-128754508 on Build GRCh38
rs746969372 Chr.7: 128754267-128754267 on Build GRCh38
rs763744180 Chr.7: 128754473-128754473 on Build GRCh38
rs981695469 Chr.7: 128754340-128754340 on Build GRCh38
rs946173161 Chr.7: 128754385-128754385 on Build GRCh38
rs748187816 Chr.7: 128754540-128754540 on Build GRCh38
rs757921265 Chr.7: 128754271-128754271 on Build GRCh38
rs1046361375 Chr.7: 128754619-128754619 on Build GRCh38
rs755986867 Chr.7: 128754403-128754403 on Build GRCh38
rs1034632098 Chr.7: 128754596-128754596 on Build GRCh38

View Full Product Details