Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 40154833-40155339 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C8orf4
Entrez Gene ID
56892
Gene Name
chromosome 8 open reading frame 4
Gene Aliases
TC-1, TC1
UniGene
Hs.591849
Gene Chromosome Location
Chr.8: 40153468-40155308 on Build GRCh38
Transcripts
NM_020130.4

SNP Detail

SNP ID Chromosome Location
rs559788059 Chr.8: 40155275-40155275 on Build GRCh38
rs570189531 Chr.8: 40155137-40155137 on Build GRCh38
rs907283438 Chr.8: 40155153-40155153 on Build GRCh38
rs914302371 Chr.8: 40155030-40155030 on Build GRCh38
rs28672910 Chr.8: 40155165-40155165 on Build GRCh38
rs975829264 Chr.8: 40155285-40155285 on Build GRCh38
rs970934938 Chr.8: 40154950-40154950 on Build GRCh38
rs928994825 Chr.8: 40155310-40155310 on Build GRCh38
rs924270266 Chr.8: 40154967-40154967 on Build GRCh38
rs546192578 Chr.8: 40155258-40155258 on Build GRCh38
rs910328592 Chr.8: 40154987-40154987 on Build GRCh38
rs929557995 Chr.8: 40155048-40155048 on Build GRCh38
rs985937329 Chr.8: 40155216-40155216 on Build GRCh38
rs569293235 Chr.8: 40154962-40154962 on Build GRCh38
rs759716584 Chr.8: 40155023-40155023 on Build GRCh38
rs563841541 Chr.8: 40155190-40155190 on Build GRCh38
rs1037825165 Chr.8: 40155233-40155233 on Build GRCh38
rs999748062 Chr.8: 40155284-40155284 on Build GRCh38
rs558150263 Chr.8: 40154975-40154975 on Build GRCh38
rs139621652 Chr.8: 40155311-40155311 on Build GRCh38
rs933198463 Chr.8: 40155132-40155132 on Build GRCh38
rs895221353 Chr.8: 40155252-40155252 on Build GRCh38
rs1044535685 Chr.8: 40155151-40155151 on Build GRCh38
rs1031630657 Chr.8: 40154935-40154935 on Build GRCh38
rs191144811 Chr.8: 40155196-40155196 on Build GRCh38
rs923204662 Chr.8: 40155128-40155128 on Build GRCh38
rs1031755743 Chr.8: 40155303-40155303 on Build GRCh38
rs956853139 Chr.8: 40154940-40154940 on Build GRCh38
rs1032100664 Chr.8: 40155176-40155176 on Build GRCh38
rs535961755 Chr.8: 40154920-40154920 on Build GRCh38
rs150597368 Chr.8: 40154892-40154892 on Build GRCh38
rs943215080 Chr.8: 40155099-40155099 on Build GRCh38
rs956066952 Chr.8: 40155183-40155183 on Build GRCh38
rs918989151 Chr.8: 40155291-40155291 on Build GRCh38
rs73606287 Chr.8: 40154886-40154886 on Build GRCh38
rs989874973 Chr.8: 40155029-40155029 on Build GRCh38
rs541324837 Chr.8: 40155067-40155067 on Build GRCh38
rs944439103 Chr.8: 40155001-40155001 on Build GRCh38
rs543531387 Chr.8: 40155167-40155167 on Build GRCh38
rs1022211182 Chr.8: 40155195-40155195 on Build GRCh38
rs187836628 Chr.8: 40155143-40155143 on Build GRCh38
rs538212174 Chr.8: 40154966-40154966 on Build GRCh38
rs981856568 Chr.8: 40154932-40154932 on Build GRCh38
rs1038871735 Chr.8: 40155108-40155108 on Build GRCh38
rs115939299 Chr.8: 40154929-40154929 on Build GRCh38
rs887917353 Chr.8: 40154863-40154863 on Build GRCh38
rs909921814 Chr.8: 40155148-40155148 on Build GRCh38
rs888026853 Chr.8: 40155133-40155133 on Build GRCh38
rs934412261 Chr.8: 40155016-40155016 on Build GRCh38
rs1040447567 Chr.8: 40155012-40155017 on Build GRCh38
rs1012393411 Chr.8: 40155261-40155261 on Build GRCh38
rs1025074772 Chr.8: 40155274-40155274 on Build GRCh38
rs571959769 Chr.8: 40155015-40155015 on Build GRCh38
rs752952465 Chr.8: 40155171-40155171 on Build GRCh38
rs1011516353 Chr.8: 40155185-40155185 on Build GRCh38

View Full Product Details