Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 47396255-47396774 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPIDR
Entrez Gene ID
23514
Gene Name
scaffolding protein involved in DNA repair
Gene Aliases
KIAA0146
UniGene
Hs.381058
Gene Chromosome Location
Chr.8: 47260878-47736001 on Build GRCh38
Transcripts
NM_001080394.3 NM_001282916.1 NM_001282919.1 NR_104581.1 XM_005251189.3 XM_005251191.3 XM_005251193.3 XM_005251195.4 XM_005251198.4 XM_005251199.4 XM_006716443.3 XM_006716444.3 XM_011517497.2 XM_011517500.2 XM_017013268.1 XM_017013269.1 XM_017013270.1 XM_017013271.1 XM_017013272.1 XM_017013273.1 XM_017013274.1 XM_017013275.1 XM_017013276.1 XM_017013277.1 XM_017013278.1 XM_017013279.1 XM_017013280.1 XM_017013281.1 XM_017013282.1 XM_017013283.1 XM_017013284.1 XM_017013285.1 XM_017013286.1 XM_017013287.1 XM_017013288.1 XM_017013289.1 XM_017013290.1 XM_017013291.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
486490 c.703T>C p.L235L Chr.8: 47396553-47396553 on Build GRCh38
751377 c.715C>G p.Q239E Chr.8: 47396565-47396565 on Build GRCh38
605599 c.756A>T p.S252S Chr.8: 47396606-47396606 on Build GRCh38
1162101 c.611C>G p.S204C Chr.8: 47396461-47396461 on Build GRCh38
3900369 c.713C>A p.P238H Chr.8: 47396563-47396563 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782598746 Chr.8: 47396583-47396583 on Build GRCh38
rs781848107 Chr.8: 47396527-47396527 on Build GRCh38
rs782104342 Chr.8: 47396631-47396631 on Build GRCh38
rs782808722 Chr.8: 47396661-47396661 on Build GRCh38
rs933936479 Chr.8: 47396681-47396681 on Build GRCh38
rs782549220 Chr.8: 47396639-47396639 on Build GRCh38
rs781920757 Chr.8: 47396472-47396472 on Build GRCh38
rs782193292 Chr.8: 47396595-47396595 on Build GRCh38
rs782718440 Chr.8: 47396489-47396489 on Build GRCh38
rs782485125 Chr.8: 47396468-47396469 on Build GRCh38
rs933847212 Chr.8: 47396315-47396315 on Build GRCh38
rs781963884 Chr.8: 47396344-47396344 on Build GRCh38
rs782244112 Chr.8: 47396578-47396578 on Build GRCh38
rs782769636 Chr.8: 47396526-47396526 on Build GRCh38
rs782059196 Chr.8: 47396651-47396651 on Build GRCh38
rs782098742 Chr.8: 47396492-47396492 on Build GRCh38
rs564570072 Chr.8: 47396365-47396365 on Build GRCh38
rs922542674 Chr.8: 47396657-47396657 on Build GRCh38
rs782267850 Chr.8: 47396444-47396444 on Build GRCh38
rs199811662 Chr.8: 47396413-47396413 on Build GRCh38
rs782129147 Chr.8: 47396510-47396510 on Build GRCh38
rs781850801 Chr.8: 47396452-47396453 on Build GRCh38
rs377584458 Chr.8: 47396676-47396676 on Build GRCh38
rs782796743 Chr.8: 47396505-47396505 on Build GRCh38
rs1039660189 Chr.8: 47396299-47396299 on Build GRCh38
rs376651593 Chr.8: 47396409-47396409 on Build GRCh38
rs956427830 Chr.8: 47396504-47396504 on Build GRCh38
rs781794960 Chr.8: 47396580-47396580 on Build GRCh38
rs782463319 Chr.8: 47396387-47396387 on Build GRCh38
rs782566278 Chr.8: 47396563-47396563 on Build GRCh38
rs942575544 Chr.8: 47396281-47396281 on Build GRCh38
rs782672626 Chr.8: 47396702-47396702 on Build GRCh38
rs200652874 Chr.8: 47396625-47396625 on Build GRCh38
rs782368097 Chr.8: 47396306-47396306 on Build GRCh38
rs782663481 Chr.8: 47396432-47396432 on Build GRCh38
rs75731959 Chr.8: 47396535-47396535 on Build GRCh38
rs781807952 Chr.8: 47396643-47396643 on Build GRCh38
rs908395285 Chr.8: 47396737-47396737 on Build GRCh38
rs782142138 Chr.8: 47396615-47396615 on Build GRCh38
rs201866039 Chr.8: 47396371-47396371 on Build GRCh38
rs144058164 Chr.8: 47396353-47396353 on Build GRCh38
rs782085725 Chr.8: 47396376-47396376 on Build GRCh38
rs782701257 Chr.8: 47396364-47396364 on Build GRCh38
rs781882965 Chr.8: 47396508-47396508 on Build GRCh38
rs782476635 Chr.8: 47396530-47396530 on Build GRCh38
rs373621549 Chr.8: 47396448-47396448 on Build GRCh38
rs373900377 Chr.8: 47396562-47396562 on Build GRCh38
rs914886573 Chr.8: 47396518-47396518 on Build GRCh38
rs1013096153 Chr.8: 47396458-47396458 on Build GRCh38
rs781985363 Chr.8: 47396491-47396491 on Build GRCh38
rs782523926 Chr.8: 47396364-47396365 on Build GRCh38
rs782322153 Chr.8: 47396467-47396467 on Build GRCh38
rs782616390 Chr.8: 47396455-47396455 on Build GRCh38
rs782306854 Chr.8: 47396596-47396596 on Build GRCh38
rs781932865 Chr.8: 47396496-47396496 on Build GRCh38
rs369135241 Chr.8: 47396478-47396478 on Build GRCh38
rs370208493 Chr.8: 47396453-47396453 on Build GRCh38
rs782040070 Chr.8: 47396368-47396368 on Build GRCh38
rs201205083 Chr.8: 47396607-47396607 on Build GRCh38
rs139083896 Chr.8: 47396303-47396303 on Build GRCh38
rs782218741 Chr.8: 47396459-47396459 on Build GRCh38
rs556111257 Chr.8: 47396338-47396338 on Build GRCh38
rs199888365 Chr.8: 47396494-47396494 on Build GRCh38
rs1046880151 Chr.8: 47396684-47396684 on Build GRCh38
rs782299758 Chr.8: 47396477-47396477 on Build GRCh38
rs781987881 Chr.8: 47396627-47396627 on Build GRCh38
rs782720277 Chr.8: 47396390-47396390 on Build GRCh38
rs781840199 Chr.8: 47396375-47396375 on Build GRCh38
rs199971445 Chr.8: 47396488-47396488 on Build GRCh38
rs782660975 Chr.8: 47396575-47396575 on Build GRCh38
rs782043026 Chr.8: 47396498-47396498 on Build GRCh38
rs781966856 Chr.8: 47396545-47396545 on Build GRCh38
rs782601266 Chr.8: 47396534-47396534 on Build GRCh38
rs782067492 Chr.8: 47396348-47396348 on Build GRCh38

View Full Product Details