Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 56446333-56446875 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC101929415
Entrez Gene ID
101929415
Gene Name
uncharacterized LOC101929415
Gene Aliases
-
UniGene
Hs.701520
Gene Chromosome Location
Chr.8: 56445807-56552067 on Build GRCh38
Transcripts
NR_125813.1 NM_001135690.2

Gene Symbol
PENK
Entrez Gene ID
5179
Gene Name
proenkephalin
Gene Aliases
PE, PENK-A
UniGene
Hs.339831
Gene Chromosome Location
Chr.8: 56440954-56446734 on Build GRCh38
Transcripts
NR_125813.1 NM_001135690.2

SNP Detail

SNP ID Chromosome Location
rs967378601 Chr.8: 56446698-56446698 on Build GRCh38
rs978000146 Chr.8: 56446454-56446454 on Build GRCh38
rs1056607633 Chr.8: 56446385-56446385 on Build GRCh38
rs1032154484 Chr.8: 56446570-56446570 on Build GRCh38
rs908328299 Chr.8: 56446389-56446389 on Build GRCh38
rs572782869 Chr.8: 56446625-56446625 on Build GRCh38
rs949176062 Chr.8: 56446783-56446783 on Build GRCh38
rs939800409 Chr.8: 56446679-56446679 on Build GRCh38
rs1014216586 Chr.8: 56446801-56446801 on Build GRCh38
rs1038070245 Chr.8: 56446463-56446463 on Build GRCh38
rs536068284 Chr.8: 56446505-56446505 on Build GRCh38
rs949123326 Chr.8: 56446622-56446622 on Build GRCh38
rs1052970664 Chr.8: 56446800-56446800 on Build GRCh38
rs2609999 Chr.8: 56446541-56446541 on Build GRCh38
rs970242970 Chr.8: 56446633-56446633 on Build GRCh38
rs1049665733 Chr.8: 56446731-56446731 on Build GRCh38
rs148194717 Chr.8: 56446387-56446387 on Build GRCh38
rs919595305 Chr.8: 56446694-56446694 on Build GRCh38
rs11545965 Chr.8: 56446433-56446433 on Build GRCh38
rs1020309536 Chr.8: 56446697-56446697 on Build GRCh38
rs898794145 Chr.8: 56446823-56446823 on Build GRCh38
rs188156999 Chr.8: 56446842-56446842 on Build GRCh38
rs373077341 Chr.8: 56446605-56446605 on Build GRCh38
rs939068494 Chr.8: 56446375-56446375 on Build GRCh38
rs943855127 Chr.8: 56446780-56446780 on Build GRCh38
rs564150739 Chr.8: 56446825-56446825 on Build GRCh38
rs936534304 Chr.8: 56446520-56446520 on Build GRCh38
rs996793753 Chr.8: 56446680-56446680 on Build GRCh38
rs754553374 Chr.8: 56446818-56446818 on Build GRCh38
rs1020358383 Chr.8: 56446853-56446854 on Build GRCh38
rs1011962188 Chr.8: 56446624-56446624 on Build GRCh38
rs1011662526 Chr.8: 56446813-56446813 on Build GRCh38
rs939665879 Chr.8: 56446423-56446423 on Build GRCh38
rs891849538 Chr.8: 56446806-56446806 on Build GRCh38
rs954820503 Chr.8: 56446377-56446377 on Build GRCh38
rs1021544699 Chr.8: 56446824-56446824 on Build GRCh38
rs891087949 Chr.8: 56446767-56446767 on Build GRCh38
rs890649957 Chr.8: 56446837-56446837 on Build GRCh38
rs1046203213 Chr.8: 56446791-56446791 on Build GRCh38
rs540989841 Chr.8: 56446807-56446807 on Build GRCh38
rs960479063 Chr.8: 56446640-56446640 on Build GRCh38
rs898388478 Chr.8: 56446478-56446478 on Build GRCh38
rs908277503 Chr.8: 56446674-56446674 on Build GRCh38
rs1001526639 Chr.8: 56446827-56446827 on Build GRCh38
rs1037375240 Chr.8: 56446822-56446822 on Build GRCh38
rs866088096 Chr.8: 56446755-56446755 on Build GRCh38
rs916429832 Chr.8: 56446782-56446782 on Build GRCh38
rs891907503 Chr.8: 56446606-56446606 on Build GRCh38
rs991109165 Chr.8: 56446776-56446776 on Build GRCh38
rs899413421 Chr.8: 56446670-56446670 on Build GRCh38
rs932522667 Chr.8: 56446710-56446710 on Build GRCh38
rs913250172 Chr.8: 56446402-56446402 on Build GRCh38
rs973712143 Chr.8: 56446686-56446686 on Build GRCh38
rs1035935913 Chr.8: 56446636-56446636 on Build GRCh38
rs978722640 Chr.8: 56446705-56446705 on Build GRCh38
rs925208372 Chr.8: 56446487-56446487 on Build GRCh38
rs932574955 Chr.8: 56446492-56446492 on Build GRCh38
rs983664266 Chr.8: 56446651-56446651 on Build GRCh38
rs960350782 Chr.8: 56446854-56446854 on Build GRCh38
rs980328326 Chr.8: 56446634-56446634 on Build GRCh38
rs1008096228 Chr.8: 56446547-56446547 on Build GRCh38

View Full Product Details