Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 70734200-70734727 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
XKR9
Entrez Gene ID
389668
Gene Name
XK related 9
Gene Aliases
XRG9
UniGene
Hs.458938
Gene Chromosome Location
Chr.8: 70669365-71066635 on Build GRCh38
Transcripts
NM_001011720.1 NM_001287258.1 NM_001287259.1 NM_001287260.1 XM_006716447.3 XM_011517524.2 XM_011517525.2 XM_011517527.2 XM_017013404.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6870858 c.1116G>A p.M372I Chr.8: 70734418-70734418 on Build GRCh38
3650699 c.1022G>A p.G341E Chr.8: 70734324-70734324 on Build GRCh38
1101443 c.933T>C p.T311T Chr.8: 70734235-70734235 on Build GRCh38
3650698 c.1000C>T p.L334F Chr.8: 70734302-70734302 on Build GRCh38
1101445 c.995G>T p.G332V Chr.8: 70734297-70734297 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767319845 Chr.8: 70734460-70734460 on Build GRCh38
rs753971526 Chr.8: 70734414-70734414 on Build GRCh38
rs939121681 Chr.8: 70734282-70734282 on Build GRCh38
rs1051167679 Chr.8: 70734628-70734628 on Build GRCh38
rs146626082 Chr.8: 70734267-70734267 on Build GRCh38
rs758111736 Chr.8: 70734624-70734624 on Build GRCh38
rs748344269 Chr.8: 70734232-70734232 on Build GRCh38
rs936294374 Chr.8: 70734647-70734648 on Build GRCh38
rs989135414 Chr.8: 70734649-70734649 on Build GRCh38
rs751042925 Chr.8: 70734560-70734562 on Build GRCh38
rs761190463 Chr.8: 70734356-70734356 on Build GRCh38
rs945020654 Chr.8: 70734572-70734572 on Build GRCh38
rs778737522 Chr.8: 70734546-70734546 on Build GRCh38
rs777705878 Chr.8: 70734236-70734236 on Build GRCh38
rs369664634 Chr.8: 70734394-70734394 on Build GRCh38
rs771871778 Chr.8: 70734261-70734261 on Build GRCh38
rs888409121 Chr.8: 70734515-70734515 on Build GRCh38
rs982802223 Chr.8: 70734434-70734434 on Build GRCh38
rs1039403902 Chr.8: 70734620-70734620 on Build GRCh38
rs114543585 Chr.8: 70734419-70734419 on Build GRCh38
rs116289101 Chr.8: 70734313-70734313 on Build GRCh38
rs770786913 Chr.8: 70734451-70734451 on Build GRCh38
rs1015601622 Chr.8: 70734530-70734530 on Build GRCh38
rs377096936 Chr.8: 70734274-70734274 on Build GRCh38
rs143177895 Chr.8: 70734606-70734606 on Build GRCh38
rs772607792 Chr.8: 70734323-70734323 on Build GRCh38
rs747748695 Chr.8: 70734334-70734334 on Build GRCh38
rs755201189 Chr.8: 70734228-70734228 on Build GRCh38
rs372770211 Chr.8: 70734273-70734273 on Build GRCh38
rs410716 Chr.8: 70734312-70734312 on Build GRCh38
rs193050616 Chr.8: 70734445-70734445 on Build GRCh38
rs922873907 Chr.8: 70734317-70734317 on Build GRCh38
rs932900183 Chr.8: 70734321-70734321 on Build GRCh38
rs759931681 Chr.8: 70734369-70734369 on Build GRCh38
rs768138534 Chr.8: 70734347-70734347 on Build GRCh38
rs754048195 Chr.8: 70734461-70734461 on Build GRCh38
rs746192962 Chr.8: 70734245-70734245 on Build GRCh38
rs766499229 Chr.8: 70734233-70734233 on Build GRCh38
rs111440841 Chr.8: 70734399-70734399 on Build GRCh38
rs750164085 Chr.8: 70734227-70734227 on Build GRCh38
rs771283965 Chr.8: 70734242-70734242 on Build GRCh38
rs769357697 Chr.8: 70734443-70734443 on Build GRCh38
rs945146198 Chr.8: 70734666-70734666 on Build GRCh38
rs545697570 Chr.8: 70734623-70734623 on Build GRCh38
rs564887609 Chr.8: 70734620-70734624 on Build GRCh38
rs207469334 Chr.8: 70734523-70734523 on Build GRCh38
rs903799152 Chr.8: 70734366-70734366 on Build GRCh38
rs543122360 Chr.8: 70734496-70734496 on Build GRCh38
rs765470942 Chr.8: 70734467-70734467 on Build GRCh38
rs372837042 Chr.8: 70734453-70734453 on Build GRCh38
rs961470396 Chr.8: 70734566-70734566 on Build GRCh38
rs148334859 Chr.8: 70734390-70734390 on Build GRCh38
rs926335767 Chr.8: 70734280-70734280 on Build GRCh38
rs998279633 Chr.8: 70734406-70734406 on Build GRCh38
rs527883763 Chr.8: 70734688-70734688 on Build GRCh38
rs534554881 Chr.8: 70734454-70734454 on Build GRCh38
rs766128266 Chr.8: 70734296-70734296 on Build GRCh38
rs761827600 Chr.8: 70734287-70734287 on Build GRCh38
rs757050196 Chr.8: 70734499-70734499 on Build GRCh38
rs554754600 Chr.8: 70734464-70734464 on Build GRCh38
rs752979133 Chr.8: 70734302-70734302 on Build GRCh38
rs751039777 Chr.8: 70734431-70734431 on Build GRCh38
rs746718072 Chr.8: 70734336-70734336 on Build GRCh38
rs751293760 Chr.8: 70734398-70734398 on Build GRCh38
rs199630915 Chr.8: 70734230-70734230 on Build GRCh38
rs796953041 Chr.8: 70734640-70734641 on Build GRCh38
rs775995806 Chr.8: 70734286-70734286 on Build GRCh38
rs775992547 Chr.8: 70734326-70734326 on Build GRCh38
rs757137997 Chr.8: 70734225-70734225 on Build GRCh38
rs1007319569 Chr.8: 70734386-70734386 on Build GRCh38
rs774782700 Chr.8: 70734243-70734243 on Build GRCh38
rs373034048 Chr.8: 70734432-70734432 on Build GRCh38
rs754443897 Chr.8: 70734226-70734226 on Build GRCh38
rs775332328 Chr.8: 70734263-70734263 on Build GRCh38
rs779106012 Chr.8: 70734421-70734421 on Build GRCh38
rs765019886 Chr.8: 70734291-70734291 on Build GRCh38
rs778425756 Chr.8: 70734293-70734293 on Build GRCh38
rs375996539 Chr.8: 70734440-70734440 on Build GRCh38
rs115479275 Chr.8: 70734269-70734269 on Build GRCh38
rs549479673 Chr.8: 70734378-70734378 on Build GRCh38
rs183419561 Chr.8: 70734568-70734568 on Build GRCh38
rs368667695 Chr.8: 70734293-70734293 on Build GRCh38
rs769796311 Chr.8: 70734360-70734360 on Build GRCh38
rs541227946 Chr.8: 70734702-70734702 on Build GRCh38
rs375243 Chr.8: 70734309-70734309 on Build GRCh38
rs747273885 Chr.8: 70734436-70734436 on Build GRCh38
rs746356541 Chr.8: 70734314-70734314 on Build GRCh38
rs453136 Chr.8: 70734379-70734379 on Build GRCh38
rs2290703 Chr.8: 70734241-70734241 on Build GRCh38
rs925169060 Chr.8: 70734601-70734601 on Build GRCh38
rs199532081 Chr.8: 70734652-70734652 on Build GRCh38
rs762887719 Chr.8: 70734446-70734446 on Build GRCh38
rs985673457 Chr.8: 70734611-70734611 on Build GRCh38
rs910220247 Chr.8: 70734631-70734631 on Build GRCh38
rs867404674 Chr.8: 70734403-70734403 on Build GRCh38
rs1043386956 Chr.8: 70734578-70734578 on Build GRCh38
rs952832987 Chr.8: 70734285-70734285 on Build GRCh38
rs138237314 Chr.8: 70734673-70734673 on Build GRCh38
rs771690019 Chr.8: 70734438-70734439 on Build GRCh38
rs574008710 Chr.8: 70734492-70734492 on Build GRCh38
rs538526509 Chr.8: 70734442-70734442 on Build GRCh38
rs764181796 Chr.8: 70734410-70734410 on Build GRCh38
rs1005611985 Chr.8: 70734524-70734524 on Build GRCh38
rs74975207 Chr.8: 70734226-70734226 on Build GRCh38

View Full Product Details