Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 99216604-99217152 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALG2
Entrez Gene ID
85365
Gene Name
ALG2, alpha-1,3/1,6-mannosyltransferase
Gene Aliases
CDGIi, CMS14, CMSTA3, NET38, hALPG2
UniGene
Hs.40919
Gene Chromosome Location
Chr.9: 99216425-99221964 on Build GRCh38
Transcripts
NM_033087.3 NR_024532.1

SNP Detail

SNP ID Chromosome Location
rs931880508 Chr.9: 99217102-99217102 on Build GRCh38
rs776181833 Chr.9: 99216688-99216688 on Build GRCh38
rs934205997 Chr.9: 99216869-99216869 on Build GRCh38
rs1017047451 Chr.9: 99216685-99216685 on Build GRCh38
rs35668168 Chr.9: 99216850-99216851 on Build GRCh38
rs960579368 Chr.9: 99216718-99216718 on Build GRCh38
rs1021790815 Chr.9: 99216915-99216917 on Build GRCh38
rs889777883 Chr.9: 99216916-99216916 on Build GRCh38
rs990700861 Chr.9: 99216723-99216723 on Build GRCh38
rs577121693 Chr.9: 99216997-99216997 on Build GRCh38
rs764853786 Chr.9: 99216991-99216991 on Build GRCh38
rs780178144 Chr.9: 99217078-99217078 on Build GRCh38
rs956441367 Chr.9: 99217084-99217084 on Build GRCh38
rs916226511 Chr.9: 99216730-99216730 on Build GRCh38
rs537194394 Chr.9: 99216945-99216945 on Build GRCh38
rs190332411 Chr.9: 99216827-99216827 on Build GRCh38
rs938467868 Chr.9: 99216963-99216963 on Build GRCh38
rs377344326 Chr.9: 99216992-99216992 on Build GRCh38
rs777372125 Chr.9: 99217008-99217008 on Build GRCh38
rs886063259 Chr.9: 99216671-99216674 on Build GRCh38
rs752478970 Chr.9: 99216959-99216959 on Build GRCh38
rs376220185 Chr.9: 99216834-99216834 on Build GRCh38
rs373676804 Chr.9: 99216826-99216826 on Build GRCh38
rs546196040 Chr.9: 99217029-99217029 on Build GRCh38
rs996897017 Chr.9: 99216752-99216752 on Build GRCh38
rs889808918 Chr.9: 99216665-99216665 on Build GRCh38
rs768961491 Chr.9: 99216706-99216708 on Build GRCh38
rs1047623728 Chr.9: 99216704-99216704 on Build GRCh38
rs558535455 Chr.9: 99216632-99216632 on Build GRCh38
rs921906752 Chr.9: 99217098-99217098 on Build GRCh38
rs979826316 Chr.9: 99216947-99216947 on Build GRCh38
rs765889573 Chr.9: 99216793-99216793 on Build GRCh38
rs562811725 Chr.9: 99217042-99217042 on Build GRCh38
rs898502002 Chr.9: 99216746-99216747 on Build GRCh38
rs934815161 Chr.9: 99216811-99216811 on Build GRCh38
rs541388782 Chr.9: 99216977-99216977 on Build GRCh38
rs749447273 Chr.9: 99216658-99216659 on Build GRCh38
rs979581050 Chr.9: 99216776-99216776 on Build GRCh38
rs565984629 Chr.9: 99216732-99216732 on Build GRCh38
rs777002843 Chr.9: 99217126-99217126 on Build GRCh38
rs994055204 Chr.9: 99216691-99216691 on Build GRCh38
rs960548356 Chr.9: 99217019-99217019 on Build GRCh38
rs754650118 Chr.9: 99216864-99216864 on Build GRCh38
rs923607704 Chr.9: 99217068-99217068 on Build GRCh38
rs1049777115 Chr.9: 99217116-99217117 on Build GRCh38
rs182511989 Chr.9: 99217034-99217034 on Build GRCh38
rs768809942 Chr.9: 99217125-99217125 on Build GRCh38
rs967671965 Chr.9: 99217038-99217038 on Build GRCh38
rs1005610603 Chr.9: 99216678-99216678 on Build GRCh38
rs747200262 Chr.9: 99217108-99217108 on Build GRCh38
rs940201082 Chr.9: 99217088-99217088 on Build GRCh38
rs749978807 Chr.9: 99216773-99216773 on Build GRCh38
rs907718068 Chr.9: 99217059-99217059 on Build GRCh38
rs1005579643 Chr.9: 99216918-99216918 on Build GRCh38
rs879473723 Chr.9: 99217038-99217039 on Build GRCh38
rs185819399 Chr.9: 99216780-99216780 on Build GRCh38
rs796462499 Chr.9: 99216713-99216715 on Build GRCh38
rs573607342 Chr.9: 99217079-99217079 on Build GRCh38
rs956177731 Chr.9: 99216796-99216796 on Build GRCh38
rs948377098 Chr.9: 99216640-99216640 on Build GRCh38
rs554374279 Chr.9: 99216817-99216817 on Build GRCh38
rs1050016500 Chr.9: 99216910-99216910 on Build GRCh38
rs542429917 Chr.9: 99217096-99217096 on Build GRCh38
rs979152273 Chr.9: 99217044-99217044 on Build GRCh38
rs1035342058 Chr.9: 99216699-99216699 on Build GRCh38
rs941941566 Chr.9: 99216713-99216713 on Build GRCh38
rs774467853 Chr.9: 99216720-99216721 on Build GRCh38
rs1038858547 Chr.9: 99216926-99216926 on Build GRCh38
rs1031820712 Chr.9: 99216782-99216782 on Build GRCh38
rs764575207 Chr.9: 99216788-99216788 on Build GRCh38
rs967702873 Chr.9: 99216731-99216731 on Build GRCh38
rs765685860 Chr.9: 99216679-99216679 on Build GRCh38
rs751225116 Chr.9: 99216852-99216852 on Build GRCh38

View Full Product Details