Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 126614297-126614833 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LMX1B
Entrez Gene ID
4010
Gene Name
LIM homeobox transcription factor 1 beta
Gene Aliases
LMX1.2, NPS1
UniGene
Hs.129133
Gene Chromosome Location
Chr.9: 126614443-126701032 on Build GRCh38
Transcripts
NM_001174146.1 NM_001174147.1 NM_002316.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7327902 c.86A>G p.K29R Chr.9: 126614535-126614535 on Build GRCh38
6702032 c.99C>T p.H33H Chr.9: 126614548-126614548 on Build GRCh38
4152063 c.16G>T p.G6C Chr.9: 126614465-126614465 on Build GRCh38
7280165 c.100G>A p.A34T Chr.9: 126614549-126614549 on Build GRCh38
3730489 c.63C>T p.C21C Chr.9: 126614512-126614512 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs2235055 Chr.9: 126614797-126614797 on Build GRCh38
rs374168272 Chr.9: 126614800-126614800 on Build GRCh38
rs544660411 Chr.9: 126614623-126614623 on Build GRCh38
rs756001518 Chr.9: 126614471-126614471 on Build GRCh38
rs761427758 Chr.9: 126614561-126614561 on Build GRCh38
rs766398015 Chr.9: 126614494-126614494 on Build GRCh38
rs1017526258 Chr.9: 126614579-126614579 on Build GRCh38
rs767281663 Chr.9: 126614468-126614468 on Build GRCh38
rs768237865 Chr.9: 126614552-126614552 on Build GRCh38
rs566318362 Chr.9: 126614649-126614649 on Build GRCh38
rs1003965777 Chr.9: 126614581-126614581 on Build GRCh38
rs367661538 Chr.9: 126614565-126614565 on Build GRCh38
rs774854659 Chr.9: 126614421-126614421 on Build GRCh38
rs1053815959 Chr.9: 126614776-126614776 on Build GRCh38
rs780146183 Chr.9: 126614347-126614347 on Build GRCh38
rs1056718084 Chr.9: 126614331-126614331 on Build GRCh38
rs557891117 Chr.9: 126614704-126614704 on Build GRCh38
rs376265443 Chr.9: 126614506-126614506 on Build GRCh38
rs776418737 Chr.9: 126614751-126614752 on Build GRCh38
rs2235053 Chr.9: 126614600-126614600 on Build GRCh38
rs552619001 Chr.9: 126614647-126614647 on Build GRCh38
rs753803889 Chr.9: 126614499-126614499 on Build GRCh38
rs752863245 Chr.9: 126614609-126614609 on Build GRCh38
rs781727698 Chr.9: 126614531-126614531 on Build GRCh38
rs987042553 Chr.9: 126614712-126614712 on Build GRCh38
rs750589984 Chr.9: 126614505-126614505 on Build GRCh38
rs748676035 Chr.9: 126614543-126614543 on Build GRCh38
rs902707880 Chr.9: 126614750-126614750 on Build GRCh38
rs757642890 Chr.9: 126614626-126614626 on Build GRCh38
rs763592126 Chr.9: 126614446-126614446 on Build GRCh38
rs778194415 Chr.9: 126614622-126614622 on Build GRCh38
rs945510169 Chr.9: 126614743-126614743 on Build GRCh38
rs1020068974 Chr.9: 126614788-126614788 on Build GRCh38
rs945477694 Chr.9: 126614376-126614376 on Build GRCh38
rs771509217 Chr.9: 126614418-126614418 on Build GRCh38
rs114614195 Chr.9: 126614787-126614787 on Build GRCh38
rs778251500 Chr.9: 126614509-126614509 on Build GRCh38
rs1009587773 Chr.9: 126614538-126614538 on Build GRCh38
rs920923026 Chr.9: 126614680-126614680 on Build GRCh38
rs1012810702 Chr.9: 126614558-126614558 on Build GRCh38
rs1042419929 Chr.9: 126614444-126614444 on Build GRCh38
rs747468994 Chr.9: 126614513-126614513 on Build GRCh38
rs143148278 Chr.9: 126614804-126614804 on Build GRCh38
rs1024155383 Chr.9: 126614563-126614563 on Build GRCh38
rs374538710 Chr.9: 126614611-126614611 on Build GRCh38
rs761549192 Chr.9: 126614467-126614467 on Build GRCh38
rs559931842 Chr.9: 126614640-126614640 on Build GRCh38
rs528488015 Chr.9: 126614703-126614703 on Build GRCh38
rs773994008 Chr.9: 126614458-126614458 on Build GRCh38
rs771325027 Chr.9: 126614529-126614529 on Build GRCh38
rs771784052 Chr.9: 126614564-126614564 on Build GRCh38
rs889829876 Chr.9: 126614782-126614782 on Build GRCh38
rs1001307100 Chr.9: 126614475-126614475 on Build GRCh38
rs948650749 Chr.9: 126614778-126614778 on Build GRCh38
rs912706824 Chr.9: 126614349-126614349 on Build GRCh38
rs758689701 Chr.9: 126614616-126614616 on Build GRCh38
rs563572406 Chr.9: 126614438-126614438 on Build GRCh38
rs1055119894 Chr.9: 126614528-126614528 on Build GRCh38
rs937662726 Chr.9: 126614701-126614701 on Build GRCh38
rs932908131 Chr.9: 126614336-126614336 on Build GRCh38
rs892693352 Chr.9: 126614428-126614428 on Build GRCh38
rs1000957670 Chr.9: 126614762-126614762 on Build GRCh38
rs1042624704 Chr.9: 126614747-126614747 on Build GRCh38
rs750263789 Chr.9: 126614469-126614469 on Build GRCh38
rs773902469 Chr.9: 126614560-126614560 on Build GRCh38
rs908934937 Chr.9: 126614690-126614690 on Build GRCh38
rs367777645 Chr.9: 126614809-126614809 on Build GRCh38
rs949707721 Chr.9: 126614728-126614728 on Build GRCh38
rs371424668 Chr.9: 126614607-126614607 on Build GRCh38
rs925399661 Chr.9: 126614756-126614756 on Build GRCh38
rs966323272 Chr.9: 126614582-126614582 on Build GRCh38
rs760533573 Chr.9: 126614567-126614567 on Build GRCh38
rs764059478 Chr.9: 126614427-126614433 on Build GRCh38
rs1055551660 Chr.9: 126614775-126614775 on Build GRCh38
rs976020713 Chr.9: 126614585-126614585 on Build GRCh38
rs112076964 Chr.9: 126614378-126614378 on Build GRCh38
rs758741262 Chr.9: 126614507-126614507 on Build GRCh38
rs965407244 Chr.9: 126614796-126614796 on Build GRCh38
rs532531403 Chr.9: 126614598-126614598 on Build GRCh38
rs984568982 Chr.9: 126614673-126614673 on Build GRCh38
rs759483901 Chr.9: 126614602-126614602 on Build GRCh38

View Full Product Details