Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 49830268-49830813 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PAGE4
Entrez Gene ID
9506
Gene Name
PAGE family member 4
Gene Aliases
CT16.7, GAGE-9, GAGEC1, JM-27, JM27, PAGE-1, PAGE-4
UniGene
Hs.441038
Gene Chromosome Location
Chr.X: 49829260-49834264 on Build GRCh38
Transcripts
NM_001318877.1 NM_007003.4

SNP Detail

SNP ID Chromosome Location
rs376908380 Chr.X: 49830516-49830516 on Build GRCh38
rs147290739 Chr.X: 49830428-49830428 on Build GRCh38
rs202162113 Chr.X: 49830445-49830445 on Build GRCh38
rs144490430 Chr.X: 49830297-49830297 on Build GRCh38
rs144908613 Chr.X: 49830703-49830703 on Build GRCh38
rs187637725 Chr.X: 49830733-49830733 on Build GRCh38
rs146339676 Chr.X: 49830475-49830475 on Build GRCh38
rs782223328 Chr.X: 49830555-49830555 on Build GRCh38
rs371171667 Chr.X: 49830558-49830558 on Build GRCh38
rs782237820 Chr.X: 49830483-49830483 on Build GRCh38
rs377646297 Chr.X: 49830392-49830392 on Build GRCh38
rs782730860 Chr.X: 49830372-49830372 on Build GRCh38
rs782276157 Chr.X: 49830504-49830504 on Build GRCh38
rs934930764 Chr.X: 49830302-49830302 on Build GRCh38
rs994578732 Chr.X: 49830591-49830591 on Build GRCh38
rs782763871 Chr.X: 49830387-49830387 on Build GRCh38
rs968809241 Chr.X: 49830760-49830760 on Build GRCh38
rs193098098 Chr.X: 49830634-49830634 on Build GRCh38
rs886234124 Chr.X: 49830675-49830675 on Build GRCh38
rs781986324 Chr.X: 49830520-49830520 on Build GRCh38
rs782163029 Chr.X: 49830386-49830386 on Build GRCh38
rs782523800 Chr.X: 49830427-49830427 on Build GRCh38
rs782358703 Chr.X: 49830535-49830535 on Build GRCh38
rs782528972 Chr.X: 49830470-49830470 on Build GRCh38
rs782472245 Chr.X: 49830352-49830352 on Build GRCh38
rs782018404 Chr.X: 49830383-49830383 on Build GRCh38
rs61052303 Chr.X: 49830755-49830755 on Build GRCh38
rs782652173 Chr.X: 49830714-49830714 on Build GRCh38
rs782098435 Chr.X: 49830394-49830394 on Build GRCh38
rs782578883 Chr.X: 49830500-49830500 on Build GRCh38
rs781798953 Chr.X: 49830411-49830411 on Build GRCh38
rs141683827 Chr.X: 49830410-49830410 on Build GRCh38
rs782459998 Chr.X: 49830666-49830666 on Build GRCh38
rs148418362 Chr.X: 49830298-49830298 on Build GRCh38
rs140934858 Chr.X: 49830681-49830681 on Build GRCh38
rs184275334 Chr.X: 49830698-49830698 on Build GRCh38
rs561287708 Chr.X: 49830534-49830534 on Build GRCh38
rs782443437 Chr.X: 49830485-49830485 on Build GRCh38
rs781845006 Chr.X: 49830446-49830446 on Build GRCh38
rs781839555 Chr.X: 49830376-49830376 on Build GRCh38
rs898954038 Chr.X: 49830576-49830576 on Build GRCh38
rs782168771 Chr.X: 49830654-49830654 on Build GRCh38

View Full Product Details