Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 112433140-112433646 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZDHHC6
Entrez Gene ID
64429
Gene Name
zinc finger DHHC-type containing 6
Gene Aliases
DHHC-6, ZNF376
UniGene
Hs.196990
Gene Chromosome Location
Chr.10: 112424411-112448335 on Build GRCh38
Transcripts
NM_001303134.1 NM_022494.2 XM_005270051.4 XM_006717948.3 XM_006717949.3 XM_011540091.2 XM_011540092.2 XM_011540093.1 XM_011540094.2 XM_011540095.2 XM_017016561.1 XM_017016562.1 XM_017016563.1 XM_017016564.1 XM_017016565.1 XM_017016566.1 XM_017016567.1 XM_017016568.1 XM_017016569.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6423859 c.936A>T p.R312S Chr.10: 112433249-112433249 on Build GRCh38
5041886 c.945+1G>A p.? Chr.10: 112433239-112433239 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1004864407 Chr.10: 112433380-112433380 on Build GRCh38
rs181476084 Chr.10: 112433373-112433373 on Build GRCh38
rs775820604 Chr.10: 112433573-112433573 on Build GRCh38
rs12776683 Chr.10: 112433610-112433610 on Build GRCh38
rs753458056 Chr.10: 112433259-112433259 on Build GRCh38
rs781288541 Chr.10: 112433277-112433280 on Build GRCh38
rs761637823 Chr.10: 112433226-112433226 on Build GRCh38
rs78738653 Chr.10: 112433216-112433216 on Build GRCh38
rs751622547 Chr.10: 112433310-112433310 on Build GRCh38
rs567275458 Chr.10: 112433507-112433507 on Build GRCh38
rs746105548 Chr.10: 112433323-112433323 on Build GRCh38
rs753154186 Chr.10: 112433292-112433292 on Build GRCh38
rs747527365 Chr.10: 112433206-112433206 on Build GRCh38
rs184200984 Chr.10: 112433490-112433490 on Build GRCh38
rs757422596 Chr.10: 112433311-112433311 on Build GRCh38
rs1001707263 Chr.10: 112433433-112433433 on Build GRCh38
rs759905947 Chr.10: 112433282-112433282 on Build GRCh38
rs778195885 Chr.10: 112433205-112433206 on Build GRCh38
rs75057632 Chr.10: 112433214-112433214 on Build GRCh38
rs778488730 Chr.10: 112433302-112433302 on Build GRCh38
rs1049750900 Chr.10: 112433471-112433472 on Build GRCh38
rs925197301 Chr.10: 112433551-112433551 on Build GRCh38
rs966388539 Chr.10: 112433213-112433213 on Build GRCh38
rs781511638 Chr.10: 112433320-112433320 on Build GRCh38
rs921799152 Chr.10: 112433597-112433597 on Build GRCh38
rs137910285 Chr.10: 112433548-112433548 on Build GRCh38
rs935748942 Chr.10: 112433339-112433339 on Build GRCh38
rs534289409 Chr.10: 112433452-112433452 on Build GRCh38
rs367577093 Chr.10: 112433330-112433330 on Build GRCh38
rs771446133 Chr.10: 112433244-112433245 on Build GRCh38
rs897256363 Chr.10: 112433536-112433536 on Build GRCh38
rs551930617 Chr.10: 112433338-112433338 on Build GRCh38
rs772707200 Chr.10: 112433232-112433232 on Build GRCh38
rs1041237272 Chr.10: 112433405-112433405 on Build GRCh38
rs960186800 Chr.10: 112433488-112433488 on Build GRCh38
rs1051845500 Chr.10: 112433358-112433358 on Build GRCh38
rs12356966 Chr.10: 112433387-112433387 on Build GRCh38
rs772060043 Chr.10: 112433229-112433229 on Build GRCh38
rs35836735 Chr.10: 112433515-112433516 on Build GRCh38
rs758817814 Chr.10: 112433298-112433298 on Build GRCh38
rs933137834 Chr.10: 112433622-112433622 on Build GRCh38
rs374531292 Chr.10: 112433492-112433492 on Build GRCh38
rs781566733 Chr.10: 112433215-112433215 on Build GRCh38
rs112383515 Chr.10: 112433221-112433221 on Build GRCh38
rs3214901 Chr.10: 112433310-112433311 on Build GRCh38
rs765488792 Chr.10: 112433284-112433284 on Build GRCh38
rs749051138 Chr.10: 112433218-112433218 on Build GRCh38
rs549111796 Chr.10: 112433496-112433496 on Build GRCh38
rs1038372954 Chr.10: 112433399-112433399 on Build GRCh38
rs1034558003 Chr.10: 112433477-112433477 on Build GRCh38
rs957908940 Chr.10: 112433562-112433562 on Build GRCh38
rs1014464543 Chr.10: 112433537-112433537 on Build GRCh38
rs771272767 Chr.10: 112433210-112433210 on Build GRCh38
rs970002922 Chr.10: 112433286-112433286 on Build GRCh38
rs557983659 Chr.10: 112433302-112433303 on Build GRCh38
rs980139677 Chr.10: 112433303-112433303 on Build GRCh38
rs201056784 Chr.10: 112433248-112433248 on Build GRCh38
rs756396006 Chr.10: 112433327-112433327 on Build GRCh38
rs185965398 Chr.10: 112433550-112433550 on Build GRCh38
rs901385028 Chr.10: 112433431-112433431 on Build GRCh38
rs775700604 Chr.10: 112433333-112433333 on Build GRCh38
rs932927932 Chr.10: 112433468-112433468 on Build GRCh38
rs745976946 Chr.10: 112433300-112433302 on Build GRCh38
rs188698236 Chr.10: 112433178-112433178 on Build GRCh38
rs575513280 Chr.10: 112433571-112433571 on Build GRCh38
rs747141454 Chr.10: 112433198-112433198 on Build GRCh38
rs11421522 Chr.10: 112433303-112433304 on Build GRCh38
rs899381002 Chr.10: 112433414-112433414 on Build GRCh38
rs149446941 Chr.10: 112433572-112433572 on Build GRCh38
rs767947824 Chr.10: 112433491-112433491 on Build GRCh38

View Full Product Details