Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 42531566-42532086 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LRRC57
Entrez Gene ID
255252
Gene Name
leucine rich repeat containing 57
Gene Aliases
-
UniGene
Hs.234681
Gene Chromosome Location
Chr.15: 42528332-42548804 on Build GRCh38
Transcripts
XM_011521423.2 NM_003825.3 NM_130798.2 XM_006720725.2

Gene Symbol
SNAP23
Entrez Gene ID
8773
Gene Name
synaptosome associated protein 23
Gene Aliases
HsT17016, SNAP-23, SNAP23A, SNAP23B
UniGene
Hs.511149
Gene Chromosome Location
Chr.15: 42491244-42533061 on Build GRCh38
Transcripts
XM_011521423.2 NM_003825.3 NM_130798.2 XM_006720725.2

SNP Detail

SNP ID Chromosome Location
rs889859116 Chr.15: 42531830-42531832 on Build GRCh38
rs866393586 Chr.15: 42531949-42531949 on Build GRCh38
rs920529472 Chr.15: 42531736-42531736 on Build GRCh38
rs11549246 Chr.15: 42531724-42531724 on Build GRCh38
rs989875362 Chr.15: 42531633-42531633 on Build GRCh38
rs185545020 Chr.15: 42532028-42532028 on Build GRCh38
rs969772794 Chr.15: 42532007-42532007 on Build GRCh38
rs956079255 Chr.15: 42531945-42531945 on Build GRCh38
rs772769603 Chr.15: 42531671-42531671 on Build GRCh38
rs534175283 Chr.15: 42531786-42531786 on Build GRCh38
rs576729451 Chr.15: 42531975-42531975 on Build GRCh38
rs904801383 Chr.15: 42531972-42531972 on Build GRCh38
rs527265901 Chr.15: 42531602-42531602 on Build GRCh38
rs938339134 Chr.15: 42532044-42532044 on Build GRCh38
rs1000221779 Chr.15: 42531630-42531630 on Build GRCh38
rs547108563 Chr.15: 42531607-42531607 on Build GRCh38
rs907524483 Chr.15: 42531772-42531772 on Build GRCh38
rs974515658 Chr.15: 42531709-42531709 on Build GRCh38
rs989927787 Chr.15: 42531954-42531954 on Build GRCh38
rs895983138 Chr.15: 42531593-42531593 on Build GRCh38
rs188966860 Chr.15: 42531694-42531694 on Build GRCh38
rs117476614 Chr.15: 42531750-42531750 on Build GRCh38
rs748610970 Chr.15: 42531651-42531651 on Build GRCh38
rs1044561933 Chr.15: 42531940-42531940 on Build GRCh38
rs139914618 Chr.15: 42532023-42532025 on Build GRCh38
rs553923610 Chr.15: 42531818-42531818 on Build GRCh38
rs539558497 Chr.15: 42531939-42531939 on Build GRCh38
rs532927058 Chr.15: 42531619-42531619 on Build GRCh38
rs373411749 Chr.15: 42531599-42531609 on Build GRCh38
rs564207384 Chr.15: 42531612-42531612 on Build GRCh38
rs577256948 Chr.15: 42531938-42531938 on Build GRCh38
rs914345878 Chr.15: 42531965-42531965 on Build GRCh38
rs377127825 Chr.15: 42531675-42531675 on Build GRCh38
rs201881933 Chr.15: 42531804-42531805 on Build GRCh38
rs1032074593 Chr.15: 42531650-42531650 on Build GRCh38
rs146015128 Chr.15: 42531597-42531607 on Build GRCh38
rs886696923 Chr.15: 42531986-42531986 on Build GRCh38
rs762587193 Chr.15: 42531646-42531646 on Build GRCh38
rs916298241 Chr.15: 42531882-42531882 on Build GRCh38
rs914293374 Chr.15: 42531659-42531659 on Build GRCh38
rs535338135 Chr.15: 42531744-42531744 on Build GRCh38
rs575278112 Chr.15: 42531624-42531626 on Build GRCh38
rs997843555 Chr.15: 42531929-42531929 on Build GRCh38
rs948417963 Chr.15: 42531678-42531678 on Build GRCh38
rs776268336 Chr.15: 42531807-42531807 on Build GRCh38
rs569518773 Chr.15: 42531698-42531698 on Build GRCh38
rs1044674025 Chr.15: 42531609-42531609 on Build GRCh38
rs1019616889 Chr.15: 42531883-42531883 on Build GRCh38
rs1056617531 Chr.15: 42531860-42531860 on Build GRCh38
rs56125021 Chr.15: 42531633-42531634 on Build GRCh38
rs1044021346 Chr.15: 42531681-42531681 on Build GRCh38
rs1006938017 Chr.15: 42531836-42531839 on Build GRCh38
rs548880570 Chr.15: 42531747-42531747 on Build GRCh38
rs181654094 Chr.15: 42531944-42531944 on Build GRCh38
rs965371654 Chr.15: 42531888-42531888 on Build GRCh38

View Full Product Details