Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 150628016-150628512 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GIMAP6
Entrez Gene ID
474344
Gene Name
GTPase, IMAP family member 6
Gene Aliases
IAN-2, IAN-6, IAN2, IAN6
UniGene
Hs.647105
Gene Chromosome Location
Chr.7: 150625375-150632648 on Build GRCh38
Transcripts
NM_001244071.1 NM_001244072.1 NM_024711.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6671012 c.490C>T p.R164W Chr.7: 150628108-150628108 on Build GRCh38
5463606 c.506C>T p.A169V Chr.7: 150628092-150628092 on Build GRCh38
394045 c.250C>T p.R84* Chr.7: 150628348-150628348 on Build GRCh38
3636040 c.265A>C p.K89Q Chr.7: 150628333-150628333 on Build GRCh38
1600139 c.549G>T p.Q183H Chr.7: 150628049-150628049 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs191817741 Chr.7: 150628275-150628275 on Build GRCh38
rs767036573 Chr.7: 150628174-150628174 on Build GRCh38
rs747155481 Chr.7: 150628056-150628056 on Build GRCh38
rs377194794 Chr.7: 150628245-150628245 on Build GRCh38
rs369422272 Chr.7: 150628414-150628414 on Build GRCh38
rs768604637 Chr.7: 150628397-150628397 on Build GRCh38
rs766120224 Chr.7: 150628226-150628226 on Build GRCh38
rs904002938 Chr.7: 150628307-150628307 on Build GRCh38
rs376869789 Chr.7: 150628107-150628107 on Build GRCh38
rs776876140 Chr.7: 150628472-150628472 on Build GRCh38
rs769649860 Chr.7: 150628324-150628324 on Build GRCh38
rs771698284 Chr.7: 150628292-150628292 on Build GRCh38
rs778202220 Chr.7: 150628117-150628117 on Build GRCh38
rs753596703 Chr.7: 150628241-150628241 on Build GRCh38
rs747067495 Chr.7: 150628125-150628125 on Build GRCh38
rs752898935 Chr.7: 150628106-150628106 on Build GRCh38
rs1002411269 Chr.7: 150628147-150628147 on Build GRCh38
rs773953391 Chr.7: 150628227-150628227 on Build GRCh38
rs867854300 Chr.7: 150628124-150628124 on Build GRCh38
rs146766461 Chr.7: 150628073-150628073 on Build GRCh38
rs746468152 Chr.7: 150628296-150628296 on Build GRCh38
rs762901733 Chr.7: 150628336-150628336 on Build GRCh38
rs760288329 Chr.7: 150628240-150628240 on Build GRCh38
rs769844868 Chr.7: 150628378-150628378 on Build GRCh38
rs975707950 Chr.7: 150628367-150628367 on Build GRCh38
rs775156654 Chr.7: 150628080-150628080 on Build GRCh38
rs751062815 Chr.7: 150628086-150628086 on Build GRCh38
rs62000976 Chr.7: 150628284-150628284 on Build GRCh38
rs144748380 Chr.7: 150628366-150628366 on Build GRCh38
rs773075886 Chr.7: 150628183-150628183 on Build GRCh38
rs144096765 Chr.7: 150628283-150628283 on Build GRCh38
rs766503943 Chr.7: 150628348-150628348 on Build GRCh38
rs138521615 Chr.7: 150628222-150628222 on Build GRCh38
rs560730769 Chr.7: 150628216-150628216 on Build GRCh38
rs748577330 Chr.7: 150628291-150628291 on Build GRCh38
rs532823075 Chr.7: 150628411-150628411 on Build GRCh38
rs766102408 Chr.7: 150628094-150628094 on Build GRCh38
rs369430612 Chr.7: 150628128-150628128 on Build GRCh38
rs777528459 Chr.7: 150628294-150628294 on Build GRCh38
rs750699094 Chr.7: 150628343-150628343 on Build GRCh38
rs754231907 Chr.7: 150628353-150628353 on Build GRCh38
rs868564539 Chr.7: 150628156-150628156 on Build GRCh38
rs774787796 Chr.7: 150628217-150628217 on Build GRCh38
rs777330596 Chr.7: 150628062-150628062 on Build GRCh38
rs375893282 Chr.7: 150628399-150628399 on Build GRCh38
rs375860624 Chr.7: 150628267-150628267 on Build GRCh38
rs746395431 Chr.7: 150628138-150628138 on Build GRCh38
rs146066712 Chr.7: 150628403-150628403 on Build GRCh38
rs780101306 Chr.7: 150628463-150628463 on Build GRCh38
rs759570307 Chr.7: 150628479-150628479 on Build GRCh38
rs922532694 Chr.7: 150628369-150628369 on Build GRCh38
rs767832540 Chr.7: 150628341-150628341 on Build GRCh38
rs138029142 Chr.7: 150628191-150628191 on Build GRCh38
rs775796331 Chr.7: 150628210-150628211 on Build GRCh38
rs148187154 Chr.7: 150628164-150628164 on Build GRCh38
rs553694055 Chr.7: 150628211-150628211 on Build GRCh38
rs372777404 Chr.7: 150628375-150628375 on Build GRCh38
rs149739807 Chr.7: 150628065-150628065 on Build GRCh38
rs923245283 Chr.7: 150628482-150628482 on Build GRCh38
rs768179141 Chr.7: 150628154-150628154 on Build GRCh38
rs372474469 Chr.7: 150628198-150628198 on Build GRCh38
rs145423565 Chr.7: 150628088-150628088 on Build GRCh38
rs775719953 Chr.7: 150628149-150628149 on Build GRCh38
rs749594773 Chr.7: 150628223-150628223 on Build GRCh38
rs867177978 Chr.7: 150628481-150628482 on Build GRCh38
rs867223043 Chr.7: 150628189-150628189 on Build GRCh38
rs1044027422 Chr.7: 150628108-150628108 on Build GRCh38
rs751778115 Chr.7: 150628264-150628264 on Build GRCh38
rs757367483 Chr.7: 150628209-150628209 on Build GRCh38
rs150982153 Chr.7: 150628441-150628441 on Build GRCh38
rs770058331 Chr.7: 150628070-150628071 on Build GRCh38
rs563027953 Chr.7: 150628467-150628467 on Build GRCh38
rs544332792 Chr.7: 150628303-150628303 on Build GRCh38
rs369994995 Chr.7: 150628268-150628268 on Build GRCh38
rs540666412 Chr.7: 150628143-150628143 on Build GRCh38
rs772703164 Chr.7: 150628236-150628236 on Build GRCh38
rs543061135 Chr.7: 150628406-150628406 on Build GRCh38
rs144184976 Chr.7: 150628394-150628394 on Build GRCh38
rs868828789 Chr.7: 150628392-150628392 on Build GRCh38
rs762523041 Chr.7: 150628082-150628082 on Build GRCh38
rs763603064 Chr.7: 150628470-150628470 on Build GRCh38
rs755727555 Chr.7: 150628462-150628462 on Build GRCh38
rs753036235 Chr.7: 150628356-150628356 on Build GRCh38
rs372190402 Chr.7: 150628055-150628055 on Build GRCh38
rs201648540 Chr.7: 150628165-150628165 on Build GRCh38
rs913688634 Chr.7: 150628393-150628393 on Build GRCh38
rs149049253 Chr.7: 150628134-150628134 on Build GRCh38
rs902947593 Chr.7: 150628203-150628203 on Build GRCh38
rs776667788 Chr.7: 150628250-150628250 on Build GRCh38
rs756267501 Chr.7: 150628276-150628276 on Build GRCh38
rs775620356 Chr.7: 150628382-150628382 on Build GRCh38
rs200071507 Chr.7: 150628085-150628085 on Build GRCh38
rs770135610 Chr.7: 150628140-150628140 on Build GRCh38
rs770284699 Chr.7: 150628370-150628370 on Build GRCh38
rs764376871 Chr.7: 150628202-150628202 on Build GRCh38
rs73727364 Chr.7: 150628190-150628190 on Build GRCh38
rs373004176 Chr.7: 150628304-150628304 on Build GRCh38
rs146934553 Chr.7: 150628213-150628213 on Build GRCh38
rs1010565765 Chr.7: 150628093-150628093 on Build GRCh38
rs759536442 Chr.7: 150628322-150628322 on Build GRCh38
rs751857426 Chr.7: 150628207-150628207 on Build GRCh38
rs751110665 Chr.7: 150628440-150628440 on Build GRCh38
rs747812177 Chr.7: 150628230-150628230 on Build GRCh38
rs964736814 Chr.7: 150628446-150628446 on Build GRCh38
rs759208114 Chr.7: 150628254-150628254 on Build GRCh38
rs148503988 Chr.7: 150628376-150628376 on Build GRCh38
rs114540180 Chr.7: 150628290-150628290 on Build GRCh38
rs777248371 Chr.7: 150628364-150628364 on Build GRCh38
rs780395938 Chr.7: 150628279-150628279 on Build GRCh38
rs760532667 Chr.7: 150628184-150628184 on Build GRCh38
rs143101732 Chr.7: 150628153-150628153 on Build GRCh38
rs138777121 Chr.7: 150628457-150628457 on Build GRCh38
rs17173519 Chr.7: 150628405-150628405 on Build GRCh38
rs775424203 Chr.7: 150628327-150628327 on Build GRCh38
rs772134650 Chr.7: 150628052-150628052 on Build GRCh38
rs139277843 Chr.7: 150628058-150628058 on Build GRCh38
rs147307195 Chr.7: 150628421-150628421 on Build GRCh38
rs201051041 Chr.7: 150628249-150628249 on Build GRCh38
rs758799907 Chr.7: 150628358-150628358 on Build GRCh38
rs149376163 Chr.7: 150628354-150628354 on Build GRCh38
rs529942970 Chr.7: 150628436-150628436 on Build GRCh38
rs368001704 Chr.7: 150628047-150628047 on Build GRCh38
rs199567311 Chr.7: 150628400-150628400 on Build GRCh38
rs11974345 Chr.7: 150628089-150628089 on Build GRCh38
rs202157054 Chr.7: 150628098-150628098 on Build GRCh38
rs13234724 Chr.7: 150628087-150628087 on Build GRCh38
rs951964110 Chr.7: 150628412-150628412 on Build GRCh38
rs771502406 Chr.7: 150628232-150628232 on Build GRCh38
rs765141539 Chr.7: 150628258-150628258 on Build GRCh38
rs867002814 Chr.7: 150628307-150628307 on Build GRCh38
rs758709854 Chr.7: 150628415-150628415 on Build GRCh38
rs140856543 Chr.7: 150628468-150628468 on Build GRCh38
rs745352182 Chr.7: 150628461-150628461 on Build GRCh38
rs574696492 Chr.7: 150628151-150628151 on Build GRCh38
rs376322381 Chr.7: 150628347-150628347 on Build GRCh38

View Full Product Details