Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 32746445-32746954 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HLA-DQA2
Entrez Gene ID
3118
Gene Name
major histocompatibility complex, class II, DQ alpha 2
Gene Aliases
DX-ALPHA, HLA-DXA
UniGene
Hs.591798
Gene Chromosome Location
Chr.6: 32741386-32746887 on Build GRCh38
Transcripts
NM_020056.4

SNP Detail

SNP ID Chromosome Location
rs890074507 Chr.6: 32746839-32746847 on Build GRCh38
rs902448594 Chr.6: 32746642-32746642 on Build GRCh38
rs572242231 Chr.6: 32746626-32746626 on Build GRCh38
rs114318456 Chr.6: 32746545-32746545 on Build GRCh38
rs996412741 Chr.6: 32746791-32746791 on Build GRCh38
rs566459845 Chr.6: 32746725-32746725 on Build GRCh38
rs972617183 Chr.6: 32746531-32746531 on Build GRCh38
rs1055658982 Chr.6: 32746558-32746558 on Build GRCh38
rs766849640 Chr.6: 32746597-32746597 on Build GRCh38
rs191319831 Chr.6: 32746825-32746825 on Build GRCh38
rs557518705 Chr.6: 32746720-32746720 on Build GRCh38
rs939546411 Chr.6: 32746919-32746919 on Build GRCh38
rs116570528 Chr.6: 32746913-32746913 on Build GRCh38
rs984883045 Chr.6: 32746827-32746827 on Build GRCh38
rs9276439 Chr.6: 32746826-32746826 on Build GRCh38
rs995932266 Chr.6: 32746806-32746806 on Build GRCh38
rs915907651 Chr.6: 32746664-32746664 on Build GRCh38
rs928136595 Chr.6: 32746918-32746918 on Build GRCh38
rs554449129 Chr.6: 32746768-32746768 on Build GRCh38
rs549884406 Chr.6: 32746527-32746527 on Build GRCh38
rs573580167 Chr.6: 32746800-32746800 on Build GRCh38
rs749056852 Chr.6: 32746847-32746847 on Build GRCh38
rs1037176530 Chr.6: 32746475-32746475 on Build GRCh38
rs895138528 Chr.6: 32746580-32746580 on Build GRCh38
rs879068561 Chr.6: 32746892-32746892 on Build GRCh38
rs2071798 Chr.6: 32746815-32746815 on Build GRCh38
rs142826528 Chr.6: 32746539-32746539 on Build GRCh38
rs376372673 Chr.6: 32746600-32746600 on Build GRCh38
rs529093089 Chr.6: 32746761-32746761 on Build GRCh38
rs9282272 Chr.6: 32746888-32746889 on Build GRCh38
rs9276438 Chr.6: 32746739-32746739 on Build GRCh38
rs780877469 Chr.6: 32746766-32746766 on Build GRCh38
rs200417972 Chr.6: 32746473-32746473 on Build GRCh38
rs186864820 Chr.6: 32746474-32746474 on Build GRCh38
rs74558919 Chr.6: 32746634-32746634 on Build GRCh38
rs879192320 Chr.6: 32746622-32746622 on Build GRCh38
rs369022900 Chr.6: 32746745-32746745 on Build GRCh38
rs117094765 Chr.6: 32746608-32746608 on Build GRCh38
rs112600646 Chr.6: 32746887-32746888 on Build GRCh38
rs1017113693 Chr.6: 32746840-32746840 on Build GRCh38
rs939435748 Chr.6: 32746553-32746553 on Build GRCh38
rs745796914 Chr.6: 32746685-32746685 on Build GRCh38
rs1010652651 Chr.6: 32746845-32746845 on Build GRCh38
rs371491889 Chr.6: 32746815-32746816 on Build GRCh38
rs796343070 Chr.6: 32746699-32746699 on Build GRCh38
rs1043622940 Chr.6: 32746611-32746611 on Build GRCh38
rs555605406 Chr.6: 32746747-32746747 on Build GRCh38
rs1022009643 Chr.6: 32746873-32746873 on Build GRCh38
rs531697297 Chr.6: 32746509-32746509 on Build GRCh38
rs868507392 Chr.6: 32746802-32746802 on Build GRCh38

View Full Product Details