Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 76881596-76882182 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCTD12
Entrez Gene ID
115207
Gene Name
potassium channel tetramerization domain containing 12
Gene Aliases
C13orf2, PFET1, PFETIN
UniGene
Hs.644125
Gene Chromosome Location
Chr.13: 76880169-76886405 on Build GRCh38
Transcripts
NM_138444.3

SNP Detail

SNP ID Chromosome Location
rs932125475 Chr.13: 76882035-76882035 on Build GRCh38
rs34157208 Chr.13: 76881788-76881789 on Build GRCh38
rs778511636 Chr.13: 76882127-76882127 on Build GRCh38
rs570922831 Chr.13: 76881763-76881763 on Build GRCh38
rs920672869 Chr.13: 76881947-76881947 on Build GRCh38
rs78535252 Chr.13: 76881792-76881792 on Build GRCh38
rs868197140 Chr.13: 76881742-76881742 on Build GRCh38
rs6732 Chr.13: 76881991-76881991 on Build GRCh38
rs34242725 Chr.13: 76881836-76881837 on Build GRCh38
rs80240140 Chr.13: 76881679-76881679 on Build GRCh38
rs796557857 Chr.13: 76881672-76881672 on Build GRCh38
rs949706721 Chr.13: 76881904-76881904 on Build GRCh38
rs4052607 Chr.13: 76881791-76881792 on Build GRCh38
rs56981893 Chr.13: 76881665-76881666 on Build GRCh38
rs970283260 Chr.13: 76881916-76881916 on Build GRCh38
rs13337 Chr.13: 76882048-76882048 on Build GRCh38
rs763865485 Chr.13: 76881666-76881667 on Build GRCh38
rs1032786990 Chr.13: 76881845-76881845 on Build GRCh38
rs7705 Chr.13: 76881871-76881871 on Build GRCh38
rs891099526 Chr.13: 76881631-76881631 on Build GRCh38
rs551918083 Chr.13: 76881717-76881717 on Build GRCh38
rs991284668 Chr.13: 76882119-76882119 on Build GRCh38
rs34132489 Chr.13: 76881837-76881837 on Build GRCh38
rs1033758252 Chr.13: 76882095-76882095 on Build GRCh38
rs1013957774 Chr.13: 76882152-76882152 on Build GRCh38
rs991356908 Chr.13: 76881867-76881867 on Build GRCh38
rs955692061 Chr.13: 76881826-76881826 on Build GRCh38
rs971708206 Chr.13: 76882137-76882137 on Build GRCh38
rs958569810 Chr.13: 76881862-76881862 on Build GRCh38
rs968548225 Chr.13: 76881816-76881816 on Build GRCh38
rs377115297 Chr.13: 76881660-76881661 on Build GRCh38
rs369288876 Chr.13: 76881677-76881678 on Build GRCh38
rs374626148 Chr.13: 76881677-76881677 on Build GRCh38
rs572053622 Chr.13: 76881998-76881998 on Build GRCh38
rs188908763 Chr.13: 76881978-76881978 on Build GRCh38
rs900655981 Chr.13: 76881701-76881701 on Build GRCh38
rs200046768 Chr.13: 76881787-76881788 on Build GRCh38
rs1053418 Chr.13: 76881961-76881961 on Build GRCh38
rs192562899 Chr.13: 76881989-76881989 on Build GRCh38
rs143424640 Chr.13: 76881789-76881790 on Build GRCh38
rs56991465 Chr.13: 76881681-76881682 on Build GRCh38
rs1000268193 Chr.13: 76881793-76881793 on Build GRCh38
rs376228901 Chr.13: 76881672-76881673 on Build GRCh38
rs904264448 Chr.13: 76882075-76882075 on Build GRCh38
rs543377730 Chr.13: 76882093-76882093 on Build GRCh38
rs746172475 Chr.13: 76881838-76881838 on Build GRCh38
rs57833605 Chr.13: 76881851-76881852 on Build GRCh38
rs370650437 Chr.13: 76881661-76881662 on Build GRCh38
rs2301578 Chr.13: 76881831-76881831 on Build GRCh38
rs554176071 Chr.13: 76882055-76882055 on Build GRCh38
rs1029955086 Chr.13: 76881626-76881626 on Build GRCh38
rs56743549 Chr.13: 76881674-76881675 on Build GRCh38
rs891404257 Chr.13: 76882026-76882026 on Build GRCh38
rs778603911 Chr.13: 76881837-76881838 on Build GRCh38
rs958496225 Chr.13: 76882104-76882104 on Build GRCh38
rs908230832 Chr.13: 76881944-76881944 on Build GRCh38
rs1031306302 Chr.13: 76881807-76881807 on Build GRCh38
rs1053433 Chr.13: 76881756-76881756 on Build GRCh38
rs982868899 Chr.13: 76881906-76881906 on Build GRCh38
rs60493446 Chr.13: 76881678-76881679 on Build GRCh38
rs386772488 Chr.13: 76881666-76881679 on Build GRCh38
rs952195466 Chr.13: 76881955-76881955 on Build GRCh38
rs184425964 Chr.13: 76882037-76882037 on Build GRCh38
rs371871726 Chr.13: 76881664-76881665 on Build GRCh38
rs560776562 Chr.13: 76881755-76881755 on Build GRCh38
rs869297843 Chr.13: 76881792-76881793 on Build GRCh38
rs796470805 Chr.13: 76881852-76881852 on Build GRCh38
rs567657949 Chr.13: 76881869-76881869 on Build GRCh38
rs57776381 Chr.13: 76881668-76881669 on Build GRCh38
rs979982793 Chr.13: 76881939-76881939 on Build GRCh38
rs199666914 Chr.13: 76881679-76881680 on Build GRCh38
rs74986456 Chr.13: 76881850-76881852 on Build GRCh38
rs944727948 Chr.13: 76881968-76881968 on Build GRCh38
rs138604266 Chr.13: 76881667-76881668 on Build GRCh38
rs60090589 Chr.13: 76881671-76881672 on Build GRCh38
rs1024238729 Chr.13: 76881874-76881874 on Build GRCh38
rs575609515 Chr.13: 76882091-76882091 on Build GRCh38
rs202247283 Chr.13: 76881667-76881667 on Build GRCh38
rs368116241 Chr.13: 76881795-76881795 on Build GRCh38
rs200970314 Chr.13: 76881666-76881666 on Build GRCh38
rs907842949 Chr.13: 76881962-76881962 on Build GRCh38
rs373098847 Chr.13: 76881670-76881671 on Build GRCh38

View Full Product Details